Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103324654-103325199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD6
Entrez Gene ID
8323
Gene Name
frizzled class receptor 6
Gene Aliases
FZ-6, FZ6, HFZ6, NDNC10
UniGene
Hs.591863
Gene Chromosome Location
Chr.8: 103298433-103332866 on Build GRCh38
Transcripts
NM_001164615.1 NM_001164616.1 NM_001317796.1 NM_003506.3 NR_133921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668174 c.657delT p.L221fs*1 Chr.8: 103324763-103324763 on Build GRCh38
1094462 c.938G>T p.R313I Chr.8: 103325044-103325044 on Build GRCh38
5428981 c.608G>C p.G203A Chr.8: 103324714-103324714 on Build GRCh38
1094460 c.766G>A p.D256N Chr.8: 103324872-103324872 on Build GRCh38
1094461 c.868T>C p.Y290H Chr.8: 103324974-103324974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765244859 Chr.8: 103324817-103324818 on Build GRCh38
rs1029258122 Chr.8: 103324797-103324797 on Build GRCh38
rs780187587 Chr.8: 103324888-103324888 on Build GRCh38
rs3808554 Chr.8: 103324868-103324868 on Build GRCh38
rs762587938 Chr.8: 103324951-103324951 on Build GRCh38
rs537987365 Chr.8: 103325015-103325015 on Build GRCh38
rs750807474 Chr.8: 103325031-103325031 on Build GRCh38
rs768709636 Chr.8: 103324919-103324919 on Build GRCh38
rs767102317 Chr.8: 103324986-103324986 on Build GRCh38
rs942373154 Chr.8: 103325156-103325156 on Build GRCh38
rs748090004 Chr.8: 103324929-103324929 on Build GRCh38
rs765626207 Chr.8: 103325030-103325030 on Build GRCh38
rs191400696 Chr.8: 103325128-103325128 on Build GRCh38
rs371618519 Chr.8: 103325140-103325140 on Build GRCh38
rs368892362 Chr.8: 103324763-103324763 on Build GRCh38
rs775573797 Chr.8: 103324711-103324711 on Build GRCh38
rs747106870 Chr.8: 103324912-103324912 on Build GRCh38
rs375091015 Chr.8: 103324947-103324947 on Build GRCh38
rs142626394 Chr.8: 103324871-103324871 on Build GRCh38
rs150261268 Chr.8: 103325009-103325009 on Build GRCh38
rs765077597 Chr.8: 103324756-103324756 on Build GRCh38
rs762687532 Chr.8: 103324799-103324799 on Build GRCh38
rs766701271 Chr.8: 103325155-103325155 on Build GRCh38
rs771311097 Chr.8: 103324683-103324683 on Build GRCh38
rs757599522 Chr.8: 103324851-103324851 on Build GRCh38
rs1004883584 Chr.8: 103325117-103325117 on Build GRCh38
rs751829856 Chr.8: 103325175-103325175 on Build GRCh38
rs759181459 Chr.8: 103324984-103324984 on Build GRCh38
rs766142758 Chr.8: 103324953-103324953 on Build GRCh38
rs759239566 Chr.8: 103325123-103325123 on Build GRCh38
rs966018925 Chr.8: 103324739-103324739 on Build GRCh38
rs201820118 Chr.8: 103324872-103324872 on Build GRCh38
rs879007257 Chr.8: 103324839-103324839 on Build GRCh38
rs141253426 Chr.8: 103324956-103324956 on Build GRCh38
rs372850452 Chr.8: 103324812-103324812 on Build GRCh38
rs769774212 Chr.8: 103325080-103325080 on Build GRCh38
rs536022156 Chr.8: 103325067-103325067 on Build GRCh38
rs551735338 Chr.8: 103324748-103324748 on Build GRCh38
rs35547794 Chr.8: 103324727-103324728 on Build GRCh38
rs763375328 Chr.8: 103325145-103325145 on Build GRCh38
rs149396895 Chr.8: 103325066-103325066 on Build GRCh38
rs900910164 Chr.8: 103324763-103324763 on Build GRCh38
rs144742285 Chr.8: 103325092-103325092 on Build GRCh38
rs751554375 Chr.8: 103324818-103324821 on Build GRCh38
rs749463825 Chr.8: 103324955-103324955 on Build GRCh38
rs748134351 Chr.8: 103325070-103325070 on Build GRCh38
rs146924761 Chr.8: 103324725-103324725 on Build GRCh38
rs976398573 Chr.8: 103325162-103325162 on Build GRCh38
rs554103695 Chr.8: 103325099-103325099 on Build GRCh38
rs770841418 Chr.8: 103325102-103325102 on Build GRCh38
rs1011584414 Chr.8: 103324802-103324802 on Build GRCh38
rs758705643 Chr.8: 103325040-103325040 on Build GRCh38
rs786205672 Chr.8: 103324975-103324975 on Build GRCh38
rs893947591 Chr.8: 103324768-103324768 on Build GRCh38
rs1048216110 Chr.8: 103325071-103325071 on Build GRCh38
rs751119249 Chr.8: 103324813-103324813 on Build GRCh38
rs138895095 Chr.8: 103325056-103325056 on Build GRCh38
rs754354607 Chr.8: 103325002-103325002 on Build GRCh38
rs370601506 Chr.8: 103324936-103324936 on Build GRCh38
rs747229738 Chr.8: 103325061-103325061 on Build GRCh38
rs377161583 Chr.8: 103324846-103324846 on Build GRCh38
rs369434689 Chr.8: 103324745-103324745 on Build GRCh38
rs746074508 Chr.8: 103324696-103324696 on Build GRCh38
rs1048279178 Chr.8: 103324713-103324713 on Build GRCh38
rs772095488 Chr.8: 103324700-103324700 on Build GRCh38
rs774304075 Chr.8: 103325168-103325168 on Build GRCh38
rs780313256 Chr.8: 103325042-103325042 on Build GRCh38
rs779068115 Chr.8: 103324684-103324684 on Build GRCh38
rs776618074 Chr.8: 103324926-103324926 on Build GRCh38
rs764751010 Chr.8: 103324842-103324842 on Build GRCh38
rs3808553 Chr.8: 103325139-103325139 on Build GRCh38
rs776494531 Chr.8: 103324727-103324727 on Build GRCh38
rs754477513 Chr.8: 103324816-103324816 on Build GRCh38
rs757364166 Chr.8: 103325120-103325120 on Build GRCh38
rs771804340 Chr.8: 103325124-103325124 on Build GRCh38

View Full Product Details