Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 108202890-108203430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3E
Entrez Gene ID
3646
Gene Name
eukaryotic translation initiation factor 3 subunit E
Gene Aliases
EIF3-P48, EIF3S6, INT6, eIF3-p46
UniGene
Hs.405590
Gene Chromosome Location
Chr.8: 108201743-108248802 on Build GRCh38
Transcripts
NM_001568.2 XM_017013398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680608 c.1163T>G p.L388* Chr.8: 108203402-108203402 on Build GRCh38
6885884 c.1231C>T p.L411F Chr.8: 108203051-108203051 on Build GRCh38
3271536 c.1184A>G p.N395S Chr.8: 108203098-108203098 on Build GRCh38
3271534 c.1234T>C p.S412P Chr.8: 108203048-108203048 on Build GRCh38
6193773 c.1211T>A p.V404E Chr.8: 108203071-108203071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773455949 Chr.8: 108203227-108203227 on Build GRCh38
rs765274205 Chr.8: 108203045-108203045 on Build GRCh38
rs376059518 Chr.8: 108202965-108202965 on Build GRCh38
rs199613801 Chr.8: 108203364-108203364 on Build GRCh38
rs771547224 Chr.8: 108203134-108203134 on Build GRCh38
rs1002822957 Chr.8: 108202993-108202993 on Build GRCh38
rs192899202 Chr.8: 108203334-108203334 on Build GRCh38
rs777767031 Chr.8: 108203068-108203068 on Build GRCh38
rs566115015 Chr.8: 108203200-108203200 on Build GRCh38
rs17851420 Chr.8: 108203076-108203076 on Build GRCh38
rs771671969 Chr.8: 108203001-108203001 on Build GRCh38
rs758575641 Chr.8: 108203048-108203048 on Build GRCh38
rs372809087 Chr.8: 108202936-108202936 on Build GRCh38
rs764232027 Chr.8: 108203151-108203151 on Build GRCh38
rs879078131 Chr.8: 108203133-108203133 on Build GRCh38
rs977136273 Chr.8: 108203256-108203256 on Build GRCh38
rs746383514 Chr.8: 108202971-108202971 on Build GRCh38
rs630492 Chr.8: 108203139-108203139 on Build GRCh38
rs768200063 Chr.8: 108202942-108202942 on Build GRCh38
rs532586768 Chr.8: 108202932-108202932 on Build GRCh38
rs779358281 Chr.8: 108202995-108202995 on Build GRCh38
rs757488671 Chr.8: 108203083-108203083 on Build GRCh38
rs201693505 Chr.8: 108203109-108203109 on Build GRCh38
rs918162744 Chr.8: 108203333-108203333 on Build GRCh38
rs770543950 Chr.8: 108203143-108203143 on Build GRCh38
rs923012262 Chr.8: 108203219-108203219 on Build GRCh38
rs141859782 Chr.8: 108203042-108203042 on Build GRCh38
rs935484989 Chr.8: 108203283-108203283 on Build GRCh38
rs1021685928 Chr.8: 108203201-108203201 on Build GRCh38
rs752879087 Chr.8: 108202956-108202956 on Build GRCh38
rs757443872 Chr.8: 108203355-108203355 on Build GRCh38
rs1025438272 Chr.8: 108203130-108203130 on Build GRCh38
rs373561944 Chr.8: 108203385-108203385 on Build GRCh38
rs536370048 Chr.8: 108203179-108203179 on Build GRCh38
rs574805645 Chr.8: 108203404-108203404 on Build GRCh38
rs751776888 Chr.8: 108203082-108203082 on Build GRCh38
rs956001795 Chr.8: 108203225-108203225 on Build GRCh38
rs779128344 Chr.8: 108202939-108202939 on Build GRCh38
rs964078140 Chr.8: 108203194-108203194 on Build GRCh38
rs766164430 Chr.8: 108203038-108203038 on Build GRCh38
rs552884998 Chr.8: 108202934-108202934 on Build GRCh38
rs538275491 Chr.8: 108203214-108203214 on Build GRCh38
rs554750746 Chr.8: 108203383-108203383 on Build GRCh38
rs201014486 Chr.8: 108203094-108203094 on Build GRCh38
rs200045166 Chr.8: 108203234-108203234 on Build GRCh38
rs781451997 Chr.8: 108202970-108202970 on Build GRCh38
rs770345209 Chr.8: 108202972-108202972 on Build GRCh38
rs758630801 Chr.8: 108202957-108202957 on Build GRCh38
rs759972588 Chr.8: 108203247-108203247 on Build GRCh38
rs534047089 Chr.8: 108202989-108202989 on Build GRCh38
rs763027718 Chr.8: 108203149-108203149 on Build GRCh38
rs995965420 Chr.8: 108203115-108203115 on Build GRCh38
rs569620347 Chr.8: 108202938-108202938 on Build GRCh38
rs188998961 Chr.8: 108203387-108203387 on Build GRCh38
rs914227353 Chr.8: 108203255-108203255 on Build GRCh38
rs536780337 Chr.8: 108203349-108203349 on Build GRCh38
rs747606340 Chr.8: 108203126-108203126 on Build GRCh38
rs202212915 Chr.8: 108202997-108202997 on Build GRCh38
rs758314339 Chr.8: 108203390-108203390 on Build GRCh38
rs184968524 Chr.8: 108203375-108203375 on Build GRCh38
rs780569016 Chr.8: 108203112-108203112 on Build GRCh38
rs771307071 Chr.8: 108202966-108202966 on Build GRCh38
rs528094721 Chr.8: 108202946-108202946 on Build GRCh38
rs773793940 Chr.8: 108202979-108202979 on Build GRCh38
rs924109542 Chr.8: 108203280-108203280 on Build GRCh38
rs1056693135 Chr.8: 108202926-108202926 on Build GRCh38
rs759439844 Chr.8: 108203145-108203145 on Build GRCh38
rs1043997118 Chr.8: 108203269-108203269 on Build GRCh38
rs760293190 Chr.8: 108203029-108203029 on Build GRCh38
rs755542322 Chr.8: 108203389-108203389 on Build GRCh38
rs201912158 Chr.8: 108203233-108203233 on Build GRCh38
rs200514892 Chr.8: 108203165-108203165 on Build GRCh38
rs765221183 Chr.8: 108203147-108203147 on Build GRCh38
rs373688259 Chr.8: 108203099-108203099 on Build GRCh38
rs567701036 Chr.8: 108203163-108203163 on Build GRCh38
rs759528132 Chr.8: 108203044-108203044 on Build GRCh38
rs778175913 Chr.8: 108202962-108202962 on Build GRCh38
rs560494740 Chr.8: 108202931-108202931 on Build GRCh38
rs781608198 Chr.8: 108203095-108203095 on Build GRCh38
rs767638985 Chr.8: 108203370-108203370 on Build GRCh38
rs559057043 Chr.8: 108203128-108203129 on Build GRCh38
rs992446720 Chr.8: 108203287-108203287 on Build GRCh38
rs756249183 Chr.8: 108203147-108203149 on Build GRCh38
rs187825004 Chr.8: 108203192-108203192 on Build GRCh38
rs988446285 Chr.8: 108203236-108203236 on Build GRCh38
rs772938662 Chr.8: 108203007-108203007 on Build GRCh38
rs370672091 Chr.8: 108203046-108203046 on Build GRCh38
rs1008137886 Chr.8: 108203184-108203184 on Build GRCh38

View Full Product Details