Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 108228065-108228572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3E
Entrez Gene ID
3646
Gene Name
eukaryotic translation initiation factor 3 subunit E
Gene Aliases
EIF3-P48, EIF3S6, INT6, eIF3-p46
UniGene
Hs.405590
Gene Chromosome Location
Chr.8: 108201743-108248802 on Build GRCh38
Transcripts
NM_001568.2 XM_017013398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
371194 c.709C>T p.L237F Chr.8: 108228280-108228280 on Build GRCh38
70587 c.614T>A p.L205H Chr.8: 108228375-108228375 on Build GRCh38
1094794 c.655C>A p.L219M Chr.8: 108228334-108228334 on Build GRCh38
1094793 c.685C>T p.R229C Chr.8: 108228304-108228304 on Build GRCh38
3665301 c.687C>A p.R229R Chr.8: 108228302-108228302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751373323 Chr.8: 108228251-108228251 on Build GRCh38
rs762916048 Chr.8: 108228325-108228328 on Build GRCh38
rs758865614 Chr.8: 108228184-108228184 on Build GRCh38
rs762396551 Chr.8: 108228216-108228216 on Build GRCh38
rs761132896 Chr.8: 108228430-108228430 on Build GRCh38
rs979115329 Chr.8: 108228123-108228123 on Build GRCh38
rs769345871 Chr.8: 108228431-108228431 on Build GRCh38
rs745996277 Chr.8: 108228540-108228540 on Build GRCh38
rs757120273 Chr.8: 108228276-108228276 on Build GRCh38
rs923587632 Chr.8: 108228131-108228131 on Build GRCh38
rs867047315 Chr.8: 108228312-108228313 on Build GRCh38
rs967751580 Chr.8: 108228148-108228149 on Build GRCh38
rs536342762 Chr.8: 108228236-108228236 on Build GRCh38
rs950387711 Chr.8: 108228203-108228203 on Build GRCh38
rs781330352 Chr.8: 108228534-108228534 on Build GRCh38
rs369275032 Chr.8: 108228396-108228396 on Build GRCh38
rs956331248 Chr.8: 108228133-108228133 on Build GRCh38
rs1030900018 Chr.8: 108228516-108228516 on Build GRCh38
rs113378357 Chr.8: 108228192-108228192 on Build GRCh38
rs1049081744 Chr.8: 108228344-108228344 on Build GRCh38
rs146377099 Chr.8: 108228494-108228494 on Build GRCh38
rs188567392 Chr.8: 108228508-108228508 on Build GRCh38
rs926253014 Chr.8: 108228162-108228162 on Build GRCh38
rs373610311 Chr.8: 108228432-108228432 on Build GRCh38
rs750423024 Chr.8: 108228306-108228306 on Build GRCh38
rs887282560 Chr.8: 108228333-108228334 on Build GRCh38
rs559634467 Chr.8: 108228472-108228472 on Build GRCh38
rs764745222 Chr.8: 108228228-108228228 on Build GRCh38
rs773789329 Chr.8: 108228416-108228416 on Build GRCh38
rs1028806610 Chr.8: 108228491-108228491 on Build GRCh38
rs779188474 Chr.8: 108228416-108228417 on Build GRCh38
rs913776787 Chr.8: 108228109-108228109 on Build GRCh38
rs967987031 Chr.8: 108228108-108228108 on Build GRCh38
rs866899086 Chr.8: 108228333-108228333 on Build GRCh38
rs781221022 Chr.8: 108228302-108228302 on Build GRCh38
rs769056791 Chr.8: 108228377-108228377 on Build GRCh38
rs775932724 Chr.8: 108228221-108228221 on Build GRCh38
rs950273052 Chr.8: 108228198-108228198 on Build GRCh38
rs57222183 Chr.8: 108228244-108228244 on Build GRCh38
rs1000797001 Chr.8: 108228504-108228504 on Build GRCh38
rs917498709 Chr.8: 108228196-108228196 on Build GRCh38
rs764158183 Chr.8: 108228411-108228412 on Build GRCh38
rs1025180132 Chr.8: 108228532-108228532 on Build GRCh38
rs202021229 Chr.8: 108228304-108228304 on Build GRCh38
rs596747 Chr.8: 108228466-108228466 on Build GRCh38
rs530365864 Chr.8: 108228520-108228520 on Build GRCh38
rs775440339 Chr.8: 108228240-108228240 on Build GRCh38
rs979189064 Chr.8: 108228150-108228150 on Build GRCh38
rs567480965 Chr.8: 108228209-108228209 on Build GRCh38
rs542305918 Chr.8: 108228523-108228523 on Build GRCh38
rs899941785 Chr.8: 108228458-108228458 on Build GRCh38
rs1020766270 Chr.8: 108228531-108228531 on Build GRCh38
rs756197860 Chr.8: 108228311-108228311 on Build GRCh38
rs991927182 Chr.8: 108228170-108228170 on Build GRCh38
rs913252608 Chr.8: 108228409-108228409 on Build GRCh38
rs927473670 Chr.8: 108228542-108228542 on Build GRCh38
rs752536890 Chr.8: 108228467-108228467 on Build GRCh38
rs199909863 Chr.8: 108228412-108228412 on Build GRCh38
rs780036432 Chr.8: 108228352-108228352 on Build GRCh38
rs749515394 Chr.8: 108228375-108228375 on Build GRCh38
rs373944451 Chr.8: 108228410-108228410 on Build GRCh38
rs759113014 Chr.8: 108228222-108228222 on Build GRCh38
rs886458327 Chr.8: 108228385-108228385 on Build GRCh38

View Full Product Details