Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109967230-109967730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV1
Entrez Gene ID
27012
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Gene Aliases
HNKA, KCNB3, KV2.3, KV8.1
UniGene
Hs.13285
Gene Chromosome Location
Chr.8: 109963645-109975847 on Build GRCh38
Transcripts
NM_014379.3

SNP Detail

SNP ID Chromosome Location
rs1005534335 Chr.8: 109967271-109967271 on Build GRCh38
rs865800591 Chr.8: 109967479-109967479 on Build GRCh38
rs1018249354 Chr.8: 109967358-109967358 on Build GRCh38
rs773108347 Chr.8: 109967629-109967629 on Build GRCh38
rs17468410 Chr.8: 109967642-109967642 on Build GRCh38
rs936434646 Chr.8: 109967466-109967466 on Build GRCh38
rs140376524 Chr.8: 109967607-109967607 on Build GRCh38
rs948496395 Chr.8: 109967604-109967604 on Build GRCh38
rs928236237 Chr.8: 109967550-109967550 on Build GRCh38
rs17468375 Chr.8: 109967616-109967616 on Build GRCh38
rs899849863 Chr.8: 109967625-109967625 on Build GRCh38
rs974641253 Chr.8: 109967366-109967366 on Build GRCh38
rs1019106011 Chr.8: 109967691-109967691 on Build GRCh38
rs908443746 Chr.8: 109967502-109967502 on Build GRCh38
rs776812055 Chr.8: 109967682-109967682 on Build GRCh38
rs867721089 Chr.8: 109967647-109967647 on Build GRCh38
rs573840336 Chr.8: 109967363-109967363 on Build GRCh38
rs773887092 Chr.8: 109967501-109967501 on Build GRCh38
rs142773024 Chr.8: 109967478-109967478 on Build GRCh38
rs573061190 Chr.8: 109967336-109967336 on Build GRCh38
rs562537644 Chr.8: 109967382-109967382 on Build GRCh38
rs1052791367 Chr.8: 109967630-109967630 on Build GRCh38
rs1009183920 Chr.8: 109967675-109967675 on Build GRCh38
rs1047703019 Chr.8: 109967588-109967588 on Build GRCh38
rs952011363 Chr.8: 109967476-109967477 on Build GRCh38
rs923693349 Chr.8: 109967402-109967402 on Build GRCh38
rs1031263446 Chr.8: 109967650-109967650 on Build GRCh38
rs192504225 Chr.8: 109967602-109967602 on Build GRCh38
rs1046679432 Chr.8: 109967531-109967531 on Build GRCh38
rs954543998 Chr.8: 109967377-109967377 on Build GRCh38
rs768228797 Chr.8: 109967410-109967410 on Build GRCh38
rs187821437 Chr.8: 109967375-109967375 on Build GRCh38
rs992660793 Chr.8: 109967521-109967521 on Build GRCh38
rs989129834 Chr.8: 109967468-109967468 on Build GRCh38
rs948244967 Chr.8: 109967488-109967488 on Build GRCh38
rs989231432 Chr.8: 109967706-109967706 on Build GRCh38
rs1031894586 Chr.8: 109967256-109967256 on Build GRCh38
rs983427695 Chr.8: 109967480-109967480 on Build GRCh38
rs964572381 Chr.8: 109967361-109967361 on Build GRCh38
rs553277776 Chr.8: 109967288-109967288 on Build GRCh38
rs957777301 Chr.8: 109967705-109967705 on Build GRCh38
rs114699150 Chr.8: 109967582-109967582 on Build GRCh38
rs999342286 Chr.8: 109967644-109967644 on Build GRCh38
rs550618282 Chr.8: 109967626-109967626 on Build GRCh38
rs1009048000 Chr.8: 109967328-109967328 on Build GRCh38

View Full Product Details