Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109967542-109968039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV1
Entrez Gene ID
27012
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Gene Aliases
HNKA, KCNB3, KV2.3, KV8.1
UniGene
Hs.13285
Gene Chromosome Location
Chr.8: 109963645-109975847 on Build GRCh38
Transcripts
NM_014379.3

SNP Detail

SNP ID Chromosome Location
rs993194406 Chr.8: 109967858-109967858 on Build GRCh38
rs17468410 Chr.8: 109967642-109967642 on Build GRCh38
rs1014906248 Chr.8: 109967804-109967804 on Build GRCh38
rs11986521 Chr.8: 109967933-109967933 on Build GRCh38
rs757536789 Chr.8: 109968006-109968006 on Build GRCh38
rs17468375 Chr.8: 109967616-109967616 on Build GRCh38
rs917177563 Chr.8: 109967868-109967868 on Build GRCh38
rs930240904 Chr.8: 109967838-109967838 on Build GRCh38
rs776812055 Chr.8: 109967682-109967682 on Build GRCh38
rs763794190 Chr.8: 109967959-109967959 on Build GRCh38
rs1047199906 Chr.8: 109967860-109967860 on Build GRCh38
rs1047703019 Chr.8: 109967588-109967588 on Build GRCh38
rs1031263446 Chr.8: 109967650-109967650 on Build GRCh38
rs948525564 Chr.8: 109967875-109967875 on Build GRCh38
rs928183749 Chr.8: 109967800-109967800 on Build GRCh38
rs981803195 Chr.8: 109967768-109967768 on Build GRCh38
rs191860698 Chr.8: 109967958-109967958 on Build GRCh38
rs952149478 Chr.8: 109967755-109967755 on Build GRCh38
rs187050897 Chr.8: 109967791-109967791 on Build GRCh38
rs114699150 Chr.8: 109967582-109967582 on Build GRCh38
rs979973494 Chr.8: 109967896-109967896 on Build GRCh38
rs1039762478 Chr.8: 109967904-109967904 on Build GRCh38
rs999342286 Chr.8: 109967644-109967644 on Build GRCh38
rs944389921 Chr.8: 109967971-109967971 on Build GRCh38
rs983398260 Chr.8: 109967778-109967778 on Build GRCh38
rs1040991764 Chr.8: 109967989-109967989 on Build GRCh38
rs773108347 Chr.8: 109967629-109967629 on Build GRCh38
rs948496395 Chr.8: 109967604-109967604 on Build GRCh38
rs140376524 Chr.8: 109967607-109967607 on Build GRCh38
rs766454434 Chr.8: 109967751-109967751 on Build GRCh38
rs890348073 Chr.8: 109967962-109967962 on Build GRCh38
rs548664944 Chr.8: 109967895-109967895 on Build GRCh38
rs899849863 Chr.8: 109967625-109967625 on Build GRCh38
rs1019106011 Chr.8: 109967691-109967691 on Build GRCh38
rs562917342 Chr.8: 109967851-109967851 on Build GRCh38
rs763140422 Chr.8: 109967861-109967861 on Build GRCh38
rs867721089 Chr.8: 109967647-109967647 on Build GRCh38
rs1052791367 Chr.8: 109967630-109967630 on Build GRCh38
rs1009183920 Chr.8: 109967675-109967675 on Build GRCh38
rs192504225 Chr.8: 109967602-109967602 on Build GRCh38
rs899963949 Chr.8: 109967931-109967931 on Build GRCh38
rs775648895 Chr.8: 109967828-109967828 on Build GRCh38
rs969034722 Chr.8: 109967750-109967750 on Build GRCh38
rs183835046 Chr.8: 109967740-109967740 on Build GRCh38
rs989231432 Chr.8: 109967706-109967706 on Build GRCh38
rs910050161 Chr.8: 109967878-109967878 on Build GRCh38
rs912937867 Chr.8: 109967961-109967961 on Build GRCh38
rs999855698 Chr.8: 109968003-109968003 on Build GRCh38
rs145463778 Chr.8: 109967893-109967893 on Build GRCh38
rs957777301 Chr.8: 109967705-109967705 on Build GRCh38
rs764329949 Chr.8: 109967954-109967954 on Build GRCh38
rs550618282 Chr.8: 109967626-109967626 on Build GRCh38

View Full Product Details