Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119583630-119584145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP2
Entrez Gene ID
5168
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 2
Gene Aliases
ATX, ATX-X, AUTOTAXIN, LysoPLD, NPP2, PD-IALPHA, PDNP2
UniGene
Hs.190977
Gene Chromosome Location
Chr.8: 119557077-119673404 on Build GRCh38
Transcripts
NM_001040092.2 NM_001130863.2 NM_006209.4 NR_045555.1 XM_006716584.1 XM_006716585.1 XM_006716587.1 XM_017013571.1 XM_017013572.1 XM_017013573.1 XM_017013574.1 XM_017013575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366269 c.1578C>A p.D526E Chr.8: 119583995-119583995 on Build GRCh38
5629257 c.1388A>G p.K463R Chr.8: 119584029-119584029 on Build GRCh38
6654194 c.1578C>T p.D526D Chr.8: 119583995-119583995 on Build GRCh38
7000627 c.1468G>A p.G490S Chr.8: 119583792-119583792 on Build GRCh38
4468884 c.1569C>T p.F523F Chr.8: 119584004-119584004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574007126 Chr.8: 119584061-119584061 on Build GRCh38
rs753940004 Chr.8: 119583945-119583945 on Build GRCh38
rs1036589299 Chr.8: 119583937-119583937 on Build GRCh38
rs144932109 Chr.8: 119584022-119584022 on Build GRCh38
rs750323126 Chr.8: 119583940-119583940 on Build GRCh38
rs376257645 Chr.8: 119583692-119583692 on Build GRCh38
rs149170905 Chr.8: 119583899-119583899 on Build GRCh38
rs552054392 Chr.8: 119583918-119583918 on Build GRCh38
rs539599614 Chr.8: 119583893-119583893 on Build GRCh38
rs761538869 Chr.8: 119583932-119583932 on Build GRCh38
rs778363813 Chr.8: 119583856-119583856 on Build GRCh38
rs367995044 Chr.8: 119583908-119583908 on Build GRCh38
rs202031268 Chr.8: 119584044-119584044 on Build GRCh38
rs772692083 Chr.8: 119583690-119583690 on Build GRCh38
rs924832584 Chr.8: 119583831-119583831 on Build GRCh38
rs549932918 Chr.8: 119583900-119583900 on Build GRCh38
rs758597135 Chr.8: 119583806-119583807 on Build GRCh38
rs771458355 Chr.8: 119584018-119584018 on Build GRCh38
rs977715411 Chr.8: 119584099-119584099 on Build GRCh38
rs747402570 Chr.8: 119583839-119583840 on Build GRCh38
rs771554668 Chr.8: 119584089-119584089 on Build GRCh38
rs761628590 Chr.8: 119584036-119584036 on Build GRCh38
rs969363232 Chr.8: 119584098-119584098 on Build GRCh38
rs747556345 Chr.8: 119584013-119584013 on Build GRCh38
rs748518937 Chr.8: 119583841-119583841 on Build GRCh38
rs138647379 Chr.8: 119584045-119584045 on Build GRCh38
rs936128139 Chr.8: 119583992-119583992 on Build GRCh38
rs759602216 Chr.8: 119584090-119584090 on Build GRCh38
rs369132467 Chr.8: 119583963-119583963 on Build GRCh38
rs766417245 Chr.8: 119583943-119583943 on Build GRCh38
rs779201526 Chr.8: 119583822-119583822 on Build GRCh38
rs923777652 Chr.8: 119583763-119583763 on Build GRCh38
rs570331954 Chr.8: 119583964-119583964 on Build GRCh38
rs368406755 Chr.8: 119583740-119583740 on Build GRCh38
rs567059238 Chr.8: 119584017-119584017 on Build GRCh38
rs976824212 Chr.8: 119583871-119583871 on Build GRCh38
rs767465637 Chr.8: 119583934-119583934 on Build GRCh38
rs768863918 Chr.8: 119583671-119583671 on Build GRCh38
rs760265854 Chr.8: 119584031-119584031 on Build GRCh38
rs374618555 Chr.8: 119583810-119583810 on Build GRCh38
rs1039356197 Chr.8: 119583869-119583869 on Build GRCh38
rs748649436 Chr.8: 119583682-119583682 on Build GRCh38
rs757256892 Chr.8: 119583753-119583753 on Build GRCh38
rs145038215 Chr.8: 119583739-119583739 on Build GRCh38
rs1035447902 Chr.8: 119584039-119584039 on Build GRCh38
rs71532464 Chr.8: 119583689-119583689 on Build GRCh38
rs770248113 Chr.8: 119583743-119583743 on Build GRCh38
rs372619923 Chr.8: 119583807-119583807 on Build GRCh38
rs200910762 Chr.8: 119583680-119583680 on Build GRCh38
rs774954814 Chr.8: 119583734-119583734 on Build GRCh38
rs944092471 Chr.8: 119583839-119583839 on Build GRCh38
rs778452030 Chr.8: 119584001-119584001 on Build GRCh38
rs561006610 Chr.8: 119583888-119583888 on Build GRCh38
rs755204781 Chr.8: 119583814-119583814 on Build GRCh38
rs753866969 Chr.8: 119584076-119584076 on Build GRCh38
rs537932287 Chr.8: 119584109-119584109 on Build GRCh38
rs774024173 Chr.8: 119583923-119583923 on Build GRCh38
rs764990239 Chr.8: 119583728-119583728 on Build GRCh38
rs768211674 Chr.8: 119584025-119584025 on Build GRCh38
rs201625824 Chr.8: 119584093-119584093 on Build GRCh38
rs921224187 Chr.8: 119583875-119583875 on Build GRCh38
rs10283100 Chr.8: 119583783-119583783 on Build GRCh38
rs759046480 Chr.8: 119583693-119583693 on Build GRCh38
rs758783776 Chr.8: 119583983-119583983 on Build GRCh38
rs748768934 Chr.8: 119584024-119584024 on Build GRCh38
rs755222090 Chr.8: 119583947-119583947 on Build GRCh38
rs772710238 Chr.8: 119583850-119583850 on Build GRCh38
rs778173219 Chr.8: 119583825-119583826 on Build GRCh38
rs1024327496 Chr.8: 119583968-119583968 on Build GRCh38
rs767379905 Chr.8: 119583768-119583768 on Build GRCh38
rs751359427 Chr.8: 119583741-119583741 on Build GRCh38
rs186998505 Chr.8: 119583848-119583848 on Build GRCh38
rs368948557 Chr.8: 119583914-119583914 on Build GRCh38
rs527239155 Chr.8: 119583790-119583790 on Build GRCh38
rs750516882 Chr.8: 119583771-119583771 on Build GRCh38
rs374504229 Chr.8: 119583665-119583665 on Build GRCh38
rs146606740 Chr.8: 119583915-119583915 on Build GRCh38
rs367612954 Chr.8: 119584097-119584097 on Build GRCh38
rs773945079 Chr.8: 119584029-119584029 on Build GRCh38
rs577451265 Chr.8: 119583663-119583663 on Build GRCh38
rs753120756 Chr.8: 119583971-119583971 on Build GRCh38

View Full Product Details