Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 126554709-126555217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM84B
Entrez Gene ID
157638
Gene Name
family with sequence similarity 84 member B
Gene Aliases
BCMP101, NSE2
UniGene
Hs.741352
Gene Chromosome Location
Chr.8: 126552438-126558499 on Build GRCh38
Transcripts
NM_174911.4 XM_017013107.1 XM_017013108.1

SNP Detail

SNP ID Chromosome Location
rs184063989 Chr.8: 126555179-126555179 on Build GRCh38
rs918535977 Chr.8: 126555119-126555119 on Build GRCh38
rs191791828 Chr.8: 126555115-126555115 on Build GRCh38
rs1006983261 Chr.8: 126555003-126555003 on Build GRCh38
rs945520647 Chr.8: 126554856-126554856 on Build GRCh38
rs773137803 Chr.8: 126554902-126554902 on Build GRCh38
rs571511289 Chr.8: 126554804-126554804 on Build GRCh38
rs759017088 Chr.8: 126555163-126555163 on Build GRCh38
rs929340960 Chr.8: 126554746-126554746 on Build GRCh38
rs965551909 Chr.8: 126555096-126555096 on Build GRCh38
rs896898057 Chr.8: 126554937-126554937 on Build GRCh38
rs1021281837 Chr.8: 126555046-126555046 on Build GRCh38
rs989971836 Chr.8: 126554810-126554810 on Build GRCh38
rs927947640 Chr.8: 126554876-126554876 on Build GRCh38
rs538075498 Chr.8: 126555043-126555043 on Build GRCh38
rs569227630 Chr.8: 126555030-126555030 on Build GRCh38
rs911060277 Chr.8: 126555095-126555095 on Build GRCh38
rs149700734 Chr.8: 126554738-126554738 on Build GRCh38
rs756906569 Chr.8: 126554952-126554952 on Build GRCh38
rs781501444 Chr.8: 126554765-126554765 on Build GRCh38
rs1047717235 Chr.8: 126554747-126554747 on Build GRCh38
rs187313201 Chr.8: 126555066-126555066 on Build GRCh38
rs929874343 Chr.8: 126555132-126555132 on Build GRCh38
rs913101239 Chr.8: 126555090-126555090 on Build GRCh38
rs146675216 Chr.8: 126555133-126555133 on Build GRCh38
rs1055353255 Chr.8: 126554887-126554887 on Build GRCh38
rs796081134 Chr.8: 126554855-126554855 on Build GRCh38
rs772978722 Chr.8: 126554792-126554793 on Build GRCh38
rs939366625 Chr.8: 126554877-126554877 on Build GRCh38
rs878901086 Chr.8: 126554986-126554986 on Build GRCh38
rs58174190 Chr.8: 126554843-126554843 on Build GRCh38
rs767087432 Chr.8: 126555194-126555194 on Build GRCh38
rs928052006 Chr.8: 126555166-126555166 on Build GRCh38
rs549038110 Chr.8: 126554886-126554886 on Build GRCh38
rs534040167 Chr.8: 126555103-126555103 on Build GRCh38
rs1044584153 Chr.8: 126554812-126554812 on Build GRCh38
rs370661467 Chr.8: 126554949-126554949 on Build GRCh38
rs773764496 Chr.8: 126555138-126555138 on Build GRCh38
rs993895172 Chr.8: 126554939-126554939 on Build GRCh38
rs1003130649 Chr.8: 126554862-126554862 on Build GRCh38
rs1041320280 Chr.8: 126554732-126554732 on Build GRCh38
rs770853030 Chr.8: 126555137-126555137 on Build GRCh38
rs1038586323 Chr.8: 126554929-126554929 on Build GRCh38
rs147791258 Chr.8: 126554859-126554859 on Build GRCh38
rs989940683 Chr.8: 126555057-126555057 on Build GRCh38
rs906004702 Chr.8: 126554846-126554846 on Build GRCh38
rs1005704049 Chr.8: 126554920-126554920 on Build GRCh38
rs183025228 Chr.8: 126555047-126555047 on Build GRCh38

View Full Product Details