Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 126555110-126555616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM84B
Entrez Gene ID
157638
Gene Name
family with sequence similarity 84 member B
Gene Aliases
BCMP101, NSE2
UniGene
Hs.741352
Gene Chromosome Location
Chr.8: 126552438-126558499 on Build GRCh38
Transcripts
NM_174911.4 XM_017013107.1 XM_017013108.1

SNP Detail

SNP ID Chromosome Location
rs941625257 Chr.8: 126555275-126555275 on Build GRCh38
rs1010480782 Chr.8: 126555241-126555241 on Build GRCh38
rs184063989 Chr.8: 126555179-126555179 on Build GRCh38
rs372455045 Chr.8: 126555296-126555296 on Build GRCh38
rs397690224 Chr.8: 126555361-126555362 on Build GRCh38
rs967208897 Chr.8: 126555465-126555465 on Build GRCh38
rs909220722 Chr.8: 126555446-126555446 on Build GRCh38
rs878960780 Chr.8: 126555220-126555220 on Build GRCh38
rs752400787 Chr.8: 126555210-126555210 on Build GRCh38
rs968333327 Chr.8: 126555250-126555250 on Build GRCh38
rs556257912 Chr.8: 126555244-126555244 on Build GRCh38
rs977012946 Chr.8: 126555328-126555328 on Build GRCh38
rs1050838840 Chr.8: 126555327-126555327 on Build GRCh38
rs906042432 Chr.8: 126555205-126555205 on Build GRCh38
rs1006540182 Chr.8: 126555336-126555336 on Build GRCh38
rs1038639053 Chr.8: 126555289-126555290 on Build GRCh38
rs560010680 Chr.8: 126555568-126555568 on Build GRCh38
rs760314173 Chr.8: 126555394-126555394 on Build GRCh38
rs9643213 Chr.8: 126555284-126555284 on Build GRCh38
rs770853030 Chr.8: 126555137-126555137 on Build GRCh38
rs1020176763 Chr.8: 126555447-126555447 on Build GRCh38
rs1024620519 Chr.8: 126555445-126555445 on Build GRCh38
rs1025461943 Chr.8: 126555382-126555382 on Build GRCh38
rs187849899 Chr.8: 126555317-126555317 on Build GRCh38
rs961114353 Chr.8: 126555522-126555522 on Build GRCh38
rs540489385 Chr.8: 126555529-126555529 on Build GRCh38
rs139351237 Chr.8: 126555469-126555469 on Build GRCh38
rs981709388 Chr.8: 126555485-126555486 on Build GRCh38
rs980121723 Chr.8: 126555583-126555583 on Build GRCh38
rs759017088 Chr.8: 126555163-126555163 on Build GRCh38
rs1017935732 Chr.8: 126555353-126555353 on Build GRCh38
rs60722538 Chr.8: 126555352-126555353 on Build GRCh38
rs576047324 Chr.8: 126555266-126555266 on Build GRCh38
rs990809623 Chr.8: 126555523-126555523 on Build GRCh38
rs929874343 Chr.8: 126555132-126555132 on Build GRCh38
rs749899340 Chr.8: 126555481-126555481 on Build GRCh38
rs1018474673 Chr.8: 126555300-126555300 on Build GRCh38
rs146675216 Chr.8: 126555133-126555133 on Build GRCh38
rs927409603 Chr.8: 126555585-126555585 on Build GRCh38
rs767087432 Chr.8: 126555194-126555194 on Build GRCh38
rs928052006 Chr.8: 126555166-126555166 on Build GRCh38
rs930157519 Chr.8: 126555325-126555325 on Build GRCh38
rs564477960 Chr.8: 126555440-126555440 on Build GRCh38
rs773764496 Chr.8: 126555138-126555138 on Build GRCh38
rs568719705 Chr.8: 126555401-126555401 on Build GRCh38
rs939353930 Chr.8: 126555238-126555238 on Build GRCh38
rs974300227 Chr.8: 126555584-126555584 on Build GRCh38
rs951311942 Chr.8: 126555430-126555430 on Build GRCh38
rs536270412 Chr.8: 126555509-126555509 on Build GRCh38
rs1003489209 Chr.8: 126555224-126555224 on Build GRCh38
rs892076185 Chr.8: 126555233-126555233 on Build GRCh38
rs780866017 Chr.8: 126555226-126555226 on Build GRCh38

View Full Product Details