Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 18950656-18951179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAXO1
Entrez Gene ID
158297
Gene Name
stabilizer of axonemal microtubules 1
Gene Aliases
C9orf138, FAM154A
UniGene
Hs.731593
Gene Chromosome Location
Chr.9: 18927660-19049354 on Build GRCh38
Transcripts
NM_001287049.1 NM_001287050.1 NM_153707.3 XM_011517744.1 XM_011517745.1 XM_011517746.2 XM_017014314.1 XM_017014315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403564 c.56A>G p.H19R Chr.9: 18950920-18950920 on Build GRCh38
6400523 c.151G>T p.E51* Chr.9: 18950825-18950825 on Build GRCh38
5900749 c.61C>T p.P21S Chr.9: 18950915-18950915 on Build GRCh38
1685423 c.189T>G p.P63P Chr.9: 18950787-18950787 on Build GRCh38
3942989 c.102C>G p.L34L Chr.9: 18950874-18950874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74341844 Chr.9: 18951038-18951038 on Build GRCh38
rs533574728 Chr.9: 18951075-18951075 on Build GRCh38
rs759828170 Chr.9: 18950924-18950924 on Build GRCh38
rs112028517 Chr.9: 18950744-18950744 on Build GRCh38
rs139678747 Chr.9: 18950879-18950879 on Build GRCh38
rs149656581 Chr.9: 18950934-18950934 on Build GRCh38
rs6475273 Chr.9: 18950789-18950789 on Build GRCh38
rs769000016 Chr.9: 18950751-18950751 on Build GRCh38
rs746698694 Chr.9: 18950738-18950738 on Build GRCh38
rs1044967250 Chr.9: 18951064-18951064 on Build GRCh38
rs944513033 Chr.9: 18950824-18950824 on Build GRCh38
rs1018452995 Chr.9: 18951134-18951134 on Build GRCh38
rs778976513 Chr.9: 18950724-18950724 on Build GRCh38
rs373188005 Chr.9: 18950815-18950815 on Build GRCh38
rs746455480 Chr.9: 18950807-18950807 on Build GRCh38
rs6475274 Chr.9: 18950982-18950982 on Build GRCh38
rs773432651 Chr.9: 18950852-18950852 on Build GRCh38
rs373939813 Chr.9: 18950935-18950935 on Build GRCh38
rs1051484785 Chr.9: 18951094-18951094 on Build GRCh38
rs142183118 Chr.9: 18951100-18951100 on Build GRCh38
rs150692117 Chr.9: 18950904-18950904 on Build GRCh38
rs763232693 Chr.9: 18950749-18950749 on Build GRCh38
rs766643681 Chr.9: 18950783-18950783 on Build GRCh38
rs538875168 Chr.9: 18950743-18950744 on Build GRCh38
rs771178814 Chr.9: 18950921-18950921 on Build GRCh38
rs75030601 Chr.9: 18950816-18950816 on Build GRCh38
rs754107981 Chr.9: 18950784-18950784 on Build GRCh38
rs745433894 Chr.9: 18951071-18951071 on Build GRCh38
rs542754967 Chr.9: 18950712-18950712 on Build GRCh38
rs192478144 Chr.9: 18950960-18950960 on Build GRCh38
rs773913326 Chr.9: 18950909-18950909 on Build GRCh38
rs765114276 Chr.9: 18950867-18950867 on Build GRCh38
rs770519044 Chr.9: 18950743-18950743 on Build GRCh38
rs776792464 Chr.9: 18950979-18950979 on Build GRCh38
rs747554175 Chr.9: 18950963-18950963 on Build GRCh38
rs567192219 Chr.9: 18951112-18951112 on Build GRCh38
rs776817030 Chr.9: 18950923-18950923 on Build GRCh38
rs947995950 Chr.9: 18950864-18950864 on Build GRCh38
rs989034734 Chr.9: 18950766-18950766 on Build GRCh38
rs918641655 Chr.9: 18950851-18950851 on Build GRCh38
rs779641510 Chr.9: 18950953-18950953 on Build GRCh38
rs892107760 Chr.9: 18951087-18951087 on Build GRCh38
rs748719736 Chr.9: 18950841-18950841 on Build GRCh38
rs1052137808 Chr.9: 18951086-18951086 on Build GRCh38
rs575777151 Chr.9: 18950788-18950788 on Build GRCh38
rs139004652 Chr.9: 18950925-18950925 on Build GRCh38
rs772403393 Chr.9: 18950842-18950842 on Build GRCh38
rs372204385 Chr.9: 18950900-18950900 on Build GRCh38
rs756037737 Chr.9: 18950954-18950954 on Build GRCh38
rs7036389 Chr.9: 18950871-18950871 on Build GRCh38
rs930048004 Chr.9: 18950859-18950859 on Build GRCh38
rs974572288 Chr.9: 18950823-18950823 on Build GRCh38
rs761768402 Chr.9: 18950851-18950851 on Build GRCh38
rs752389247 Chr.9: 18950868-18950868 on Build GRCh38
rs766388731 Chr.9: 18950888-18950889 on Build GRCh38
rs780656968 Chr.9: 18950881-18950881 on Build GRCh38
rs777119497 Chr.9: 18950737-18950737 on Build GRCh38
rs550272114 Chr.9: 18950716-18950716 on Build GRCh38
rs931914721 Chr.9: 18951026-18951026 on Build GRCh38
rs767673402 Chr.9: 18950761-18950761 on Build GRCh38
rs775628130 Chr.9: 18950986-18950986 on Build GRCh38
rs763208720 Chr.9: 18950987-18950987 on Build GRCh38
rs145470510 Chr.9: 18950937-18950937 on Build GRCh38
rs751490035 Chr.9: 18950872-18950872 on Build GRCh38
rs918573684 Chr.9: 18950999-18950999 on Build GRCh38
rs777046523 Chr.9: 18950862-18950862 on Build GRCh38
rs201015949 Chr.9: 18950965-18950965 on Build GRCh38
rs769112959 Chr.9: 18950838-18950838 on Build GRCh38
rs921785431 Chr.9: 18951016-18951016 on Build GRCh38
rs767042921 Chr.9: 18950948-18950948 on Build GRCh38
rs192818730 Chr.9: 18950709-18950709 on Build GRCh38
rs200103545 Chr.9: 18950806-18950806 on Build GRCh38
rs561020514 Chr.9: 18951013-18951013 on Build GRCh38
rs749885304 Chr.9: 18950950-18950950 on Build GRCh38
rs979329642 Chr.9: 18951036-18951036 on Build GRCh38
rs936579269 Chr.9: 18951070-18951070 on Build GRCh38
rs756905426 Chr.9: 18950813-18950813 on Build GRCh38
rs1007026267 Chr.9: 18951133-18951133 on Build GRCh38
rs956410845 Chr.9: 18950763-18950763 on Build GRCh38
rs370404136 Chr.9: 18950711-18950711 on Build GRCh38
rs7021572 Chr.9: 18950897-18950897 on Build GRCh38
rs1013852954 Chr.9: 18950705-18950705 on Build GRCh38
rs745865771 Chr.9: 18950979-18950979 on Build GRCh38
rs577515210 Chr.9: 18950926-18950926 on Build GRCh38
rs757154901 Chr.9: 18950936-18950936 on Build GRCh38
rs796256356 Chr.9: 18951005-18951005 on Build GRCh38
rs774979234 Chr.9: 18950753-18950753 on Build GRCh38
rs779355899 Chr.9: 18950901-18950901 on Build GRCh38
rs772481983 Chr.9: 18950908-18950908 on Build GRCh38
rs755796375 Chr.9: 18950952-18950952 on Build GRCh38
rs930104236 Chr.9: 18951007-18951007 on Build GRCh38
rs917765641 Chr.9: 18951009-18951009 on Build GRCh38
rs1019603230 Chr.9: 18951136-18951136 on Build GRCh38
rs775843672 Chr.9: 18950747-18950747 on Build GRCh38
rs758231016 Chr.9: 18950802-18950802 on Build GRCh38
rs774122350 Chr.9: 18950846-18950849 on Build GRCh38
rs762720753 Chr.9: 18950933-18950933 on Build GRCh38
rs183901971 Chr.9: 18951051-18951051 on Build GRCh38
rs762047455 Chr.9: 18950734-18950734 on Build GRCh38
rs201537958 Chr.9: 18950861-18950861 on Build GRCh38
rs78353825 Chr.9: 18951069-18951069 on Build GRCh38
rs551789849 Chr.9: 18950800-18950800 on Build GRCh38
rs754971353 Chr.9: 18950718-18950718 on Build GRCh38
rs200081146 Chr.9: 18950736-18950736 on Build GRCh38
rs16937198 Chr.9: 18950772-18950772 on Build GRCh38
rs762204066 Chr.9: 18950756-18950756 on Build GRCh38
rs760904247 Chr.9: 18950856-18950856 on Build GRCh38
rs760992531 Chr.9: 18950781-18950781 on Build GRCh38
rs997285818 Chr.9: 18951093-18951093 on Build GRCh38
rs754878471 Chr.9: 18950785-18950785 on Build GRCh38
rs542228263 Chr.9: 18951008-18951008 on Build GRCh38
rs1029829826 Chr.9: 18950767-18950767 on Build GRCh38
rs201872237 Chr.9: 18950839-18950839 on Build GRCh38
rs781438092 Chr.9: 18950969-18950969 on Build GRCh38
rs747497079 Chr.9: 18950910-18950910 on Build GRCh38
rs891310665 Chr.9: 18951119-18951119 on Build GRCh38
rs758366545 Chr.9: 18950870-18950870 on Build GRCh38

View Full Product Details