Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 22451776-22452288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTA1
Entrez Gene ID
63951
Gene Name
DMRT like family A1
Gene Aliases
DMO, DMRT4
UniGene
Hs.371976
Gene Chromosome Location
Chr.9: 22446841-22452473 on Build GRCh38
Transcripts
NM_022160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7134234 c.1445C>T p.S482F Chr.9: 22451841-22451841 on Build GRCh38
1674418 c.1422T>A p.F474L Chr.9: 22451818-22451818 on Build GRCh38
7112166 c.1454C>A p.S485Y Chr.9: 22451850-22451850 on Build GRCh38
7104560 c.1495C>A p.P499T Chr.9: 22451891-22451891 on Build GRCh38
293123 c.1399C>T p.R467W Chr.9: 22451795-22451795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147279220 Chr.9: 22451871-22451871 on Build GRCh38
rs983949914 Chr.9: 22452182-22452182 on Build GRCh38
rs1031306944 Chr.9: 22452045-22452045 on Build GRCh38
rs767396120 Chr.9: 22451902-22451902 on Build GRCh38
rs1052028280 Chr.9: 22451977-22451977 on Build GRCh38
rs933604119 Chr.9: 22451977-22451980 on Build GRCh38
rs748898667 Chr.9: 22451811-22451811 on Build GRCh38
rs533854325 Chr.9: 22452121-22452121 on Build GRCh38
rs761885141 Chr.9: 22452133-22452133 on Build GRCh38
rs775699664 Chr.9: 22451954-22451954 on Build GRCh38
rs572741213 Chr.9: 22452134-22452134 on Build GRCh38
rs912373599 Chr.9: 22452145-22452145 on Build GRCh38
rs1007747869 Chr.9: 22452008-22452008 on Build GRCh38
rs74311482 Chr.9: 22452052-22452054 on Build GRCh38
rs778746318 Chr.9: 22451928-22451928 on Build GRCh38
rs377188812 Chr.9: 22451944-22451945 on Build GRCh38
rs879109774 Chr.9: 22452044-22452045 on Build GRCh38
rs796542359 Chr.9: 22452053-22452054 on Build GRCh38
rs759928972 Chr.9: 22451844-22451844 on Build GRCh38
rs76681070 Chr.9: 22452086-22452086 on Build GRCh38
rs770393554 Chr.9: 22451952-22451952 on Build GRCh38
rs200448357 Chr.9: 22452043-22452043 on Build GRCh38
rs552979741 Chr.9: 22452072-22452072 on Build GRCh38
rs757793240 Chr.9: 22452102-22452102 on Build GRCh38
rs556658012 Chr.9: 22451978-22451978 on Build GRCh38
rs921006232 Chr.9: 22452119-22452119 on Build GRCh38
rs753176592 Chr.9: 22451863-22451863 on Build GRCh38
rs761825073 Chr.9: 22451962-22451962 on Build GRCh38
rs371450525 Chr.9: 22451956-22451956 on Build GRCh38
rs200626982 Chr.9: 22452086-22452086 on Build GRCh38
rs761456782 Chr.9: 22451826-22451826 on Build GRCh38
rs1013735532 Chr.9: 22451965-22451965 on Build GRCh38
rs990346875 Chr.9: 22452092-22452092 on Build GRCh38
rs374502249 Chr.9: 22452131-22452131 on Build GRCh38
rs56665855 Chr.9: 22452043-22452044 on Build GRCh38
rs939892641 Chr.9: 22451878-22451878 on Build GRCh38
rs919264232 Chr.9: 22451969-22451969 on Build GRCh38
rs771728375 Chr.9: 22451940-22451940 on Build GRCh38
rs749164588 Chr.9: 22451919-22451919 on Build GRCh38
rs62574786 Chr.9: 22452003-22452003 on Build GRCh38
rs982047600 Chr.9: 22451901-22451901 on Build GRCh38
rs76494653 Chr.9: 22451907-22451907 on Build GRCh38
rs746378786 Chr.9: 22451948-22451948 on Build GRCh38
rs751700755 Chr.9: 22451799-22451799 on Build GRCh38
rs1003582480 Chr.9: 22452116-22452116 on Build GRCh38
rs575932978 Chr.9: 22452254-22452254 on Build GRCh38
rs536827758 Chr.9: 22451914-22451914 on Build GRCh38
rs1053926575 Chr.9: 22452042-22452042 on Build GRCh38
rs1055206435 Chr.9: 22451921-22451921 on Build GRCh38
rs1009299166 Chr.9: 22452062-22452062 on Build GRCh38
rs530620208 Chr.9: 22451835-22451835 on Build GRCh38
rs986648136 Chr.9: 22452137-22452137 on Build GRCh38
rs919954591 Chr.9: 22452198-22452198 on Build GRCh38
rs773721294 Chr.9: 22451818-22451818 on Build GRCh38
rs763418548 Chr.9: 22451955-22451955 on Build GRCh38
rs771730054 Chr.9: 22451906-22451906 on Build GRCh38
rs966947067 Chr.9: 22452027-22452027 on Build GRCh38
rs564688248 Chr.9: 22452152-22452152 on Build GRCh38
rs970428138 Chr.9: 22452115-22452115 on Build GRCh38
rs746441472 Chr.9: 22451831-22451832 on Build GRCh38
rs147980569 Chr.9: 22451822-22451822 on Build GRCh38
rs763077227 Chr.9: 22451869-22451869 on Build GRCh38
rs201004574 Chr.9: 22451884-22451884 on Build GRCh38
rs768421763 Chr.9: 22451814-22451814 on Build GRCh38
rs1036914736 Chr.9: 22452171-22452171 on Build GRCh38
rs1051034178 Chr.9: 22451847-22451847 on Build GRCh38
rs745624147 Chr.9: 22451802-22451802 on Build GRCh38
rs909702832 Chr.9: 22452194-22452194 on Build GRCh38
rs562226498 Chr.9: 22452202-22452202 on Build GRCh38
rs777306115 Chr.9: 22451837-22451837 on Build GRCh38
rs111683201 Chr.9: 22451908-22451908 on Build GRCh38
rs141877034 Chr.9: 22451851-22451851 on Build GRCh38
rs747617754 Chr.9: 22451934-22451934 on Build GRCh38
rs776896935 Chr.9: 22451980-22451980 on Build GRCh38
rs959203416 Chr.9: 22451913-22451913 on Build GRCh38
rs999693407 Chr.9: 22452032-22452032 on Build GRCh38
rs374183822 Chr.9: 22451833-22451833 on Build GRCh38
rs779698916 Chr.9: 22451803-22451803 on Build GRCh38
rs770253371 Chr.9: 22451837-22451851 on Build GRCh38
rs781151246 Chr.9: 22451801-22451801 on Build GRCh38
rs773654931 Chr.9: 22452232-22452232 on Build GRCh38
rs959254144 Chr.9: 22452169-22452169 on Build GRCh38
rs35705218 Chr.9: 22451800-22451800 on Build GRCh38
rs539067523 Chr.9: 22452068-22452068 on Build GRCh38
rs754562934 Chr.9: 22451926-22451926 on Build GRCh38
rs777216280 Chr.9: 22451942-22451942 on Build GRCh38
rs957054148 Chr.9: 22452065-22452065 on Build GRCh38
rs868065888 Chr.9: 22451862-22451862 on Build GRCh38
rs765529702 Chr.9: 22451845-22451845 on Build GRCh38

View Full Product Details