Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 26860950-26861462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAAP1
Entrez Gene ID
79886
Gene Name
caspase activity and apoptosis inhibitor 1
Gene Aliases
C9orf82, CAAP
UniGene
Hs.178357
Gene Chromosome Location
Chr.9: 26840685-26892828 on Build GRCh38
Transcripts
NM_001167575.1 NM_024828.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1674420 c.682G>T p.D228Y Chr.9: 26861123-26861123 on Build GRCh38
6427269 c.670G>T p.D224Y Chr.9: 26861135-26861135 on Build GRCh38
3367678 c.674T>G p.I225S Chr.9: 26861131-26861131 on Build GRCh38
5954424 c.696C>T p.S232S Chr.9: 26861109-26861109 on Build GRCh38
1195694 c.677G>A p.C226Y Chr.9: 26861128-26861128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749149157 Chr.9: 26861044-26861044 on Build GRCh38
rs971042399 Chr.9: 26861185-26861185 on Build GRCh38
rs569828253 Chr.9: 26861197-26861197 on Build GRCh38
rs375449766 Chr.9: 26861054-26861054 on Build GRCh38
rs749407972 Chr.9: 26861109-26861109 on Build GRCh38
rs568689948 Chr.9: 26860986-26860986 on Build GRCh38
rs999841964 Chr.9: 26861395-26861395 on Build GRCh38
rs903011414 Chr.9: 26861399-26861399 on Build GRCh38
rs1038636333 Chr.9: 26861050-26861050 on Build GRCh38
rs778082435 Chr.9: 26861015-26861015 on Build GRCh38
rs768443560 Chr.9: 26861379-26861379 on Build GRCh38
rs774837408 Chr.9: 26861175-26861175 on Build GRCh38
rs372138819 Chr.9: 26861184-26861184 on Build GRCh38
rs751864465 Chr.9: 26861088-26861088 on Build GRCh38
rs188958232 Chr.9: 26861205-26861205 on Build GRCh38
rs768743791 Chr.9: 26861048-26861048 on Build GRCh38
rs201198523 Chr.9: 26861066-26861066 on Build GRCh38
rs774238098 Chr.9: 26861053-26861053 on Build GRCh38
rs556090673 Chr.9: 26861251-26861251 on Build GRCh38
rs911718375 Chr.9: 26861373-26861373 on Build GRCh38
rs765787591 Chr.9: 26861077-26861077 on Build GRCh38
rs942052953 Chr.9: 26860978-26860978 on Build GRCh38
rs12342214 Chr.9: 26861108-26861108 on Build GRCh38
rs764609163 Chr.9: 26861087-26861087 on Build GRCh38
rs904174549 Chr.9: 26861386-26861386 on Build GRCh38
rs767938374 Chr.9: 26861094-26861094 on Build GRCh38
rs1000040166 Chr.9: 26861410-26861410 on Build GRCh38
rs181801164 Chr.9: 26861300-26861300 on Build GRCh38
rs1039385618 Chr.9: 26861345-26861345 on Build GRCh38
rs574624443 Chr.9: 26861270-26861270 on Build GRCh38
rs763333218 Chr.9: 26861085-26861085 on Build GRCh38
rs771976059 Chr.9: 26861132-26861132 on Build GRCh38
rs770652765 Chr.9: 26861159-26861159 on Build GRCh38
rs139606460 Chr.9: 26861138-26861138 on Build GRCh38
rs150582572 Chr.9: 26861137-26861137 on Build GRCh38
rs754901100 Chr.9: 26861110-26861110 on Build GRCh38
rs963383982 Chr.9: 26861260-26861260 on Build GRCh38
rs750432451 Chr.9: 26861059-26861059 on Build GRCh38
rs117723134 Chr.9: 26861382-26861382 on Build GRCh38
rs368467899 Chr.9: 26861023-26861023 on Build GRCh38
rs919380526 Chr.9: 26861287-26861287 on Build GRCh38
rs919062536 Chr.9: 26861001-26861001 on Build GRCh38
rs529556369 Chr.9: 26861028-26861028 on Build GRCh38
rs982625592 Chr.9: 26861229-26861229 on Build GRCh38
rs916621889 Chr.9: 26861211-26861211 on Build GRCh38
rs900863580 Chr.9: 26861354-26861354 on Build GRCh38
rs759061339 Chr.9: 26861173-26861173 on Build GRCh38
rs184206392 Chr.9: 26861154-26861154 on Build GRCh38
rs888700901 Chr.9: 26861299-26861299 on Build GRCh38
rs141753758 Chr.9: 26861117-26861117 on Build GRCh38
rs114332590 Chr.9: 26861032-26861032 on Build GRCh38
rs949481015 Chr.9: 26861222-26861222 on Build GRCh38
rs918759739 Chr.9: 26861247-26861247 on Build GRCh38
rs752252479 Chr.9: 26861167-26861167 on Build GRCh38
rs554144076 Chr.9: 26861350-26861350 on Build GRCh38
rs750587844 Chr.9: 26861098-26861098 on Build GRCh38
rs879733418 Chr.9: 26861364-26861364 on Build GRCh38
rs749164985 Chr.9: 26861371-26861371 on Build GRCh38
rs1020103010 Chr.9: 26861123-26861123 on Build GRCh38
rs748161219 Chr.9: 26861125-26861125 on Build GRCh38
rs17694121 Chr.9: 26861435-26861435 on Build GRCh38
rs916680153 Chr.9: 26860983-26860983 on Build GRCh38
rs751954666 Chr.9: 26861021-26861021 on Build GRCh38
rs772990810 Chr.9: 26861058-26861058 on Build GRCh38
rs769760179 Chr.9: 26861030-26861030 on Build GRCh38
rs781408425 Chr.9: 26861027-26861027 on Build GRCh38
rs753432368 Chr.9: 26861084-26861084 on Build GRCh38
rs751628087 Chr.9: 26861184-26861184 on Build GRCh38
rs368505141 Chr.9: 26861174-26861174 on Build GRCh38
rs779952917 Chr.9: 26861244-26861244 on Build GRCh38

View Full Product Details