Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33265673-33266194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP5
Entrez Gene ID
51510
Gene Name
charged multivesicular body protein 5
Gene Aliases
C9orf83, CGI-34, HSPC177, PNAS-2, SNF7DC2, Vps60
UniGene
Hs.635313
Gene Chromosome Location
Chr.9: 33264879-33282069 on Build GRCh38
Transcripts
NM_001195536.1 NM_016410.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260102 c.138G>T p.K46N Chr.9: 33266078-33266078 on Build GRCh38
6183321 c.85G>C p.E29Q Chr.9: 33266025-33266025 on Build GRCh38
4842506 c.80G>C p.R27T Chr.9: 33266020-33266020 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753809757 Chr.9: 33266136-33266137 on Build GRCh38
rs1037281253 Chr.9: 33265791-33265791 on Build GRCh38
rs761246914 Chr.9: 33265821-33265821 on Build GRCh38
rs11791605 Chr.9: 33265745-33265745 on Build GRCh38
rs888627775 Chr.9: 33265771-33265771 on Build GRCh38
rs1012496077 Chr.9: 33266006-33266006 on Build GRCh38
rs111482314 Chr.9: 33265739-33265739 on Build GRCh38
rs940265825 Chr.9: 33265789-33265789 on Build GRCh38
rs1036431244 Chr.9: 33265886-33265886 on Build GRCh38
rs368164218 Chr.9: 33265796-33265796 on Build GRCh38
rs970017038 Chr.9: 33266021-33266021 on Build GRCh38
rs146626883 Chr.9: 33266077-33266077 on Build GRCh38
rs761242894 Chr.9: 33265975-33265975 on Build GRCh38
rs771726380 Chr.9: 33266139-33266139 on Build GRCh38
rs142982468 Chr.9: 33266125-33266125 on Build GRCh38
rs571137711 Chr.9: 33265921-33265921 on Build GRCh38
rs757374416 Chr.9: 33266056-33266056 on Build GRCh38
rs966608085 Chr.9: 33265851-33265851 on Build GRCh38
rs1032714906 Chr.9: 33265966-33265966 on Build GRCh38
rs1032660760 Chr.9: 33265703-33265703 on Build GRCh38
rs910035156 Chr.9: 33266100-33266100 on Build GRCh38
rs755164507 Chr.9: 33266123-33266123 on Build GRCh38
rs190478757 Chr.9: 33265809-33265809 on Build GRCh38
rs766922765 Chr.9: 33265981-33265981 on Build GRCh38
rs977994274 Chr.9: 33265936-33265936 on Build GRCh38
rs1055015679 Chr.9: 33266164-33266164 on Build GRCh38
rs916797084 Chr.9: 33265794-33265794 on Build GRCh38
rs940800419 Chr.9: 33265812-33265812 on Build GRCh38
rs921626231 Chr.9: 33266050-33266050 on Build GRCh38
rs982666160 Chr.9: 33265804-33265804 on Build GRCh38
rs182465933 Chr.9: 33266029-33266029 on Build GRCh38
rs183294815 Chr.9: 33265903-33265903 on Build GRCh38
rs974380437 Chr.9: 33266049-33266049 on Build GRCh38
rs374475946 Chr.9: 33266122-33266122 on Build GRCh38
rs1023861534 Chr.9: 33266018-33266018 on Build GRCh38
rs777234521 Chr.9: 33266142-33266142 on Build GRCh38
rs750834178 Chr.9: 33265802-33265802 on Build GRCh38
rs892494257 Chr.9: 33265828-33265828 on Build GRCh38
rs930626177 Chr.9: 33265897-33265897 on Build GRCh38
rs1008743980 Chr.9: 33265939-33265939 on Build GRCh38
rs755875918 Chr.9: 33266090-33266090 on Build GRCh38
rs760160005 Chr.9: 33266143-33266143 on Build GRCh38
rs11540561 Chr.9: 33266024-33266024 on Build GRCh38
rs138735863 Chr.9: 33265888-33265888 on Build GRCh38
rs879308381 Chr.9: 33266136-33266136 on Build GRCh38
rs1003525055 Chr.9: 33265795-33265795 on Build GRCh38
rs371624915 Chr.9: 33266133-33266133 on Build GRCh38
rs989725084 Chr.9: 33265753-33265753 on Build GRCh38
rs370192690 Chr.9: 33266119-33266119 on Build GRCh38
rs998600243 Chr.9: 33265701-33265701 on Build GRCh38
rs982372781 Chr.9: 33266054-33266054 on Build GRCh38
rs750392835 Chr.9: 33266088-33266088 on Build GRCh38
rs74883194 Chr.9: 33265958-33265958 on Build GRCh38
rs186707720 Chr.9: 33266169-33266169 on Build GRCh38
rs765727412 Chr.9: 33266015-33266015 on Build GRCh38
rs372918889 Chr.9: 33266040-33266040 on Build GRCh38
rs764219886 Chr.9: 33266037-33266037 on Build GRCh38
rs1011000554 Chr.9: 33265808-33265808 on Build GRCh38
rs186202502 Chr.9: 33265915-33265915 on Build GRCh38
rs752933348 Chr.9: 33266017-33266017 on Build GRCh38
rs191099219 Chr.9: 33265988-33265988 on Build GRCh38
rs999047683 Chr.9: 33265964-33265964 on Build GRCh38
rs1702657 Chr.9: 33266137-33266137 on Build GRCh38
rs1040638687 Chr.9: 33265940-33265940 on Build GRCh38

View Full Product Details