Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33524080-33524595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18B
Entrez Gene ID
441459
Gene Name
ankyrin repeat domain 18B
Gene Aliases
bA255A11.3, bA255A11.5
UniGene
Hs.493710
Gene Chromosome Location
Chr.9: 33524403-33578755 on Build GRCh38
Transcripts
NM_001244752.1 XM_011517881.2 XM_011517883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6588061 c.64G>C p.E22Q Chr.9: 33524553-33524553 on Build GRCh38
3686399 c.77delG p.Y28fs*27 Chr.9: 33524566-33524566 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764397820 Chr.9: 33524494-33524494 on Build GRCh38
rs572245697 Chr.9: 33524398-33524398 on Build GRCh38
rs368272098 Chr.9: 33524524-33524524 on Build GRCh38
rs780560837 Chr.9: 33524457-33524457 on Build GRCh38
rs902430781 Chr.9: 33524469-33524469 on Build GRCh38
rs927932297 Chr.9: 33524375-33524375 on Build GRCh38
rs763203854 Chr.9: 33524491-33524491 on Build GRCh38
rs143898653 Chr.9: 33524371-33524371 on Build GRCh38
rs1037100204 Chr.9: 33524192-33524192 on Build GRCh38
rs529468743 Chr.9: 33524108-33524108 on Build GRCh38
rs894937520 Chr.9: 33524236-33524236 on Build GRCh38
rs3849912 Chr.9: 33524103-33524103 on Build GRCh38
rs145182598 Chr.9: 33524367-33524367 on Build GRCh38
rs758861087 Chr.9: 33524454-33524454 on Build GRCh38
rs552368397 Chr.9: 33524565-33524566 on Build GRCh38
rs113220106 Chr.9: 33524284-33524284 on Build GRCh38
rs927986111 Chr.9: 33524570-33524570 on Build GRCh38
rs939433623 Chr.9: 33524395-33524395 on Build GRCh38
rs528733537 Chr.9: 33524191-33524191 on Build GRCh38
rs370149388 Chr.9: 33524172-33524172 on Build GRCh38
rs954208541 Chr.9: 33524308-33524308 on Build GRCh38
rs767762975 Chr.9: 33524526-33524526 on Build GRCh38
rs999175805 Chr.9: 33524268-33524268 on Build GRCh38
rs939231248 Chr.9: 33524231-33524231 on Build GRCh38
rs946559058 Chr.9: 33524437-33524437 on Build GRCh38
rs62542572 Chr.9: 33524368-33524368 on Build GRCh38
rs984302337 Chr.9: 33524544-33524544 on Build GRCh38
rs74178886 Chr.9: 33524358-33524359 on Build GRCh38
rs555569610 Chr.9: 33524389-33524389 on Build GRCh38
rs902211957 Chr.9: 33524267-33524267 on Build GRCh38
rs1055190513 Chr.9: 33524396-33524396 on Build GRCh38
rs141482639 Chr.9: 33524341-33524342 on Build GRCh38
rs577614483 Chr.9: 33524514-33524514 on Build GRCh38
rs999226560 Chr.9: 33524478-33524478 on Build GRCh38
rs1011260652 Chr.9: 33524240-33524240 on Build GRCh38
rs1055373019 Chr.9: 33524232-33524232 on Build GRCh38
rs116315226 Chr.9: 33524364-33524364 on Build GRCh38
rs549641622 Chr.9: 33524301-33524301 on Build GRCh38
rs147274232 Chr.9: 33524495-33524495 on Build GRCh38
rs549126222 Chr.9: 33524166-33524166 on Build GRCh38
rs537449259 Chr.9: 33524329-33524329 on Build GRCh38
rs571347634 Chr.9: 33524324-33524324 on Build GRCh38
rs541373523 Chr.9: 33524483-33524483 on Build GRCh38
rs1022185494 Chr.9: 33524244-33524244 on Build GRCh38
rs906621905 Chr.9: 33524205-33524205 on Build GRCh38
rs984806400 Chr.9: 33524318-33524318 on Build GRCh38
rs2380774 Chr.9: 33524307-33524307 on Build GRCh38
rs940064316 Chr.9: 33524328-33524328 on Build GRCh38
rs894987210 Chr.9: 33524420-33524420 on Build GRCh38
rs1048615666 Chr.9: 33524112-33524112 on Build GRCh38
rs974485621 Chr.9: 33524286-33524286 on Build GRCh38
rs961612226 Chr.9: 33524548-33524548 on Build GRCh38
rs373457287 Chr.9: 33524177-33524177 on Build GRCh38
rs142799809 Chr.9: 33524340-33524341 on Build GRCh38
rs891308610 Chr.9: 33524130-33524130 on Build GRCh38
rs149935885 Chr.9: 33524378-33524378 on Build GRCh38
rs45547737 Chr.9: 33524359-33524359 on Build GRCh38
rs954379561 Chr.9: 33524530-33524530 on Build GRCh38
rs535303817 Chr.9: 33524382-33524382 on Build GRCh38
rs116092427 Chr.9: 33524363-33524363 on Build GRCh38
rs921565241 Chr.9: 33524102-33524102 on Build GRCh38
rs1017172420 Chr.9: 33524546-33524546 on Build GRCh38
rs965778634 Chr.9: 33524276-33524276 on Build GRCh38
rs567708891 Chr.9: 33524381-33524381 on Build GRCh38
rs775960699 Chr.9: 33524485-33524485 on Build GRCh38

View Full Product Details