Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34958028-34958565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF24
Entrez Gene ID
23349
Gene Name
PHD finger protein 24
Gene Aliases
KIAA1045
UniGene
Hs.7989
Gene Chromosome Location
Chr.9: 34810040-34982544 on Build GRCh38
Transcripts
NM_001304333.2 NM_015297.2 NR_136307.1 NR_136308.1 XM_011517827.1 XM_011517828.2 XM_011517829.1 XM_017014553.1 XM_017014554.1 XM_017014555.1 XM_017014556.1

SNP Detail

SNP ID Chromosome Location
rs569641972 Chr.9: 34958296-34958296 on Build GRCh38
rs866171097 Chr.9: 34958275-34958275 on Build GRCh38
rs1046096312 Chr.9: 34958060-34958060 on Build GRCh38
rs924478383 Chr.9: 34958385-34958385 on Build GRCh38
rs1015776772 Chr.9: 34958270-34958270 on Build GRCh38
rs981164081 Chr.9: 34958069-34958069 on Build GRCh38
rs1028455056 Chr.9: 34958220-34958221 on Build GRCh38
rs917958226 Chr.9: 34958365-34958365 on Build GRCh38
rs549788888 Chr.9: 34958219-34958219 on Build GRCh38
rs533191672 Chr.9: 34958101-34958101 on Build GRCh38
rs959395500 Chr.9: 34958329-34958329 on Build GRCh38
rs36002106 Chr.9: 34958521-34958522 on Build GRCh38
rs893061307 Chr.9: 34958420-34958420 on Build GRCh38
rs1014171389 Chr.9: 34958055-34958055 on Build GRCh38
rs1032659397 Chr.9: 34958071-34958071 on Build GRCh38
rs1054332861 Chr.9: 34958412-34958412 on Build GRCh38
rs772771562 Chr.9: 34958255-34958256 on Build GRCh38
rs113792004 Chr.9: 34958515-34958516 on Build GRCh38
rs1035363754 Chr.9: 34958206-34958206 on Build GRCh38
rs1009842308 Chr.9: 34958542-34958542 on Build GRCh38
rs112630672 Chr.9: 34958338-34958338 on Build GRCh38
rs949891049 Chr.9: 34958169-34958169 on Build GRCh38
rs771823085 Chr.9: 34958215-34958215 on Build GRCh38
rs540130531 Chr.9: 34958139-34958140 on Build GRCh38
rs1045572725 Chr.9: 34958178-34958178 on Build GRCh38
rs533991403 Chr.9: 34958509-34958509 on Build GRCh38
rs915739573 Chr.9: 34958158-34958158 on Build GRCh38
rs957103424 Chr.9: 34958071-34958071 on Build GRCh38
rs974346039 Chr.9: 34958277-34958277 on Build GRCh38
rs998439427 Chr.9: 34958070-34958070 on Build GRCh38
rs1008787525 Chr.9: 34958235-34958235 on Build GRCh38
rs1049422210 Chr.9: 34958462-34958462 on Build GRCh38
rs1030391191 Chr.9: 34958472-34958472 on Build GRCh38
rs548142626 Chr.9: 34958363-34958363 on Build GRCh38
rs932269302 Chr.9: 34958423-34958423 on Build GRCh38
rs1055658787 Chr.9: 34958138-34958138 on Build GRCh38
rs879194374 Chr.9: 34958280-34958280 on Build GRCh38
rs961641553 Chr.9: 34958109-34958109 on Build GRCh38
rs949516426 Chr.9: 34958367-34958367 on Build GRCh38
rs955761881 Chr.9: 34958097-34958097 on Build GRCh38
rs997432645 Chr.9: 34958469-34958469 on Build GRCh38
rs7863453 Chr.9: 34958255-34958255 on Build GRCh38
rs919953660 Chr.9: 34958132-34958132 on Build GRCh38
rs1028487338 Chr.9: 34958087-34958087 on Build GRCh38
rs891850669 Chr.9: 34958540-34958540 on Build GRCh38
rs892840512 Chr.9: 34958217-34958217 on Build GRCh38
rs919922881 Chr.9: 34958301-34958301 on Build GRCh38
rs974040789 Chr.9: 34958118-34958118 on Build GRCh38
rs990845122 Chr.9: 34958364-34958364 on Build GRCh38
rs903109780 Chr.9: 34958064-34958064 on Build GRCh38
rs998743774 Chr.9: 34958216-34958216 on Build GRCh38
rs529171694 Chr.9: 34958309-34958309 on Build GRCh38
rs938058908 Chr.9: 34958136-34958136 on Build GRCh38
rs7863231 Chr.9: 34958220-34958220 on Build GRCh38
rs568122546 Chr.9: 34958368-34958368 on Build GRCh38
rs934574292 Chr.9: 34958402-34958402 on Build GRCh38
rs996573862 Chr.9: 34958085-34958085 on Build GRCh38
rs908747299 Chr.9: 34958103-34958103 on Build GRCh38
rs955152585 Chr.9: 34958223-34958223 on Build GRCh38
rs866180952 Chr.9: 34958189-34958189 on Build GRCh38

View Full Product Details