Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35609507-35610015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD72
Entrez Gene ID
971
Gene Name
CD72 molecule
Gene Aliases
CD72b, LYB2
UniGene
Hs.116481
Gene Chromosome Location
Chr.9: 35609977-35646857 on Build GRCh38
Transcripts
NM_001782.2 XM_006716893.2 XM_006716894.3 XM_011518074.2 XM_017015351.1 NM_001318230.1 NM_006285.2

Gene Symbol
TESK1
Entrez Gene ID
7016
Gene Name
testis-specific kinase 1
Gene Aliases
-
UniGene
Hs.708096
Gene Chromosome Location
Chr.9: 35605284-35610041 on Build GRCh38
Transcripts
NM_001782.2 XM_006716893.2 XM_006716894.3 XM_011518074.2 XM_017015351.1 NM_001318230.1 NM_006285.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4752689 c.1834G>A p.A612T Chr.9: 35609695-35609695 on Build GRCh38
7393546 c.1720G>A p.G574S Chr.9: 35609581-35609581 on Build GRCh38
6799997 c.1803G>A p.A601A Chr.9: 35609664-35609664 on Build GRCh38
6799998 c.1679G>A p.R560Q Chr.9: 35609540-35609540 on Build GRCh38
5835743 c.1751_1752insC p.R586fs*15 Chr.9: 35609612-35609613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771060906 Chr.9: 35609709-35609709 on Build GRCh38
rs776428805 Chr.9: 35609610-35609610 on Build GRCh38
rs769669483 Chr.9: 35609695-35609695 on Build GRCh38
rs139165251 Chr.9: 35609846-35609846 on Build GRCh38
rs764117160 Chr.9: 35609777-35609821 on Build GRCh38
rs754617394 Chr.9: 35609758-35609758 on Build GRCh38
rs775468891 Chr.9: 35609526-35609526 on Build GRCh38
rs777678872 Chr.9: 35609779-35609779 on Build GRCh38
rs555884768 Chr.9: 35609925-35609925 on Build GRCh38
rs1047762604 Chr.9: 35609844-35609844 on Build GRCh38
rs56280235 Chr.9: 35609580-35609580 on Build GRCh38
rs929469734 Chr.9: 35609918-35609918 on Build GRCh38
rs781482575 Chr.9: 35609570-35609570 on Build GRCh38
rs567619446 Chr.9: 35609920-35609920 on Build GRCh38
rs759848808 Chr.9: 35609729-35609729 on Build GRCh38
rs775307644 Chr.9: 35609598-35609598 on Build GRCh38
rs780729192 Chr.9: 35609759-35609759 on Build GRCh38
rs1002067089 Chr.9: 35609902-35609902 on Build GRCh38
rs774420657 Chr.9: 35609725-35609725 on Build GRCh38
rs1038727158 Chr.9: 35609939-35609939 on Build GRCh38
rs966366134 Chr.9: 35609715-35609715 on Build GRCh38
rs979478805 Chr.9: 35609891-35609891 on Build GRCh38
rs757378958 Chr.9: 35609644-35609644 on Build GRCh38
rs747966223 Chr.9: 35609694-35609694 on Build GRCh38
rs779506569 Chr.9: 35609673-35609673 on Build GRCh38
rs767978520 Chr.9: 35609643-35609643 on Build GRCh38
rs746745234 Chr.9: 35609674-35609674 on Build GRCh38
rs955865645 Chr.9: 35609983-35609983 on Build GRCh38
rs773145759 Chr.9: 35609704-35609704 on Build GRCh38
rs745857724 Chr.9: 35609648-35609648 on Build GRCh38
rs900181046 Chr.9: 35609949-35609949 on Build GRCh38
rs764642330 Chr.9: 35609753-35609753 on Build GRCh38
rs771812530 Chr.9: 35609595-35609595 on Build GRCh38
rs1037693105 Chr.9: 35609970-35609970 on Build GRCh38
rs781079474 Chr.9: 35609690-35609690 on Build GRCh38
rs146243679 Chr.9: 35609645-35609645 on Build GRCh38
rs763041122 Chr.9: 35609625-35609625 on Build GRCh38
rs552566995 Chr.9: 35609926-35609926 on Build GRCh38
rs1026572313 Chr.9: 35609655-35609655 on Build GRCh38
rs533441187 Chr.9: 35609639-35609639 on Build GRCh38
rs753068416 Chr.9: 35609735-35609735 on Build GRCh38
rs974861909 Chr.9: 35609797-35609797 on Build GRCh38
rs1030093520 Chr.9: 35609837-35609837 on Build GRCh38
rs374955583 Chr.9: 35609975-35609975 on Build GRCh38
rs918052224 Chr.9: 35609914-35609914 on Build GRCh38
rs923384840 Chr.9: 35609825-35609825 on Build GRCh38
rs942006166 Chr.9: 35609873-35609873 on Build GRCh38
rs529352747 Chr.9: 35609604-35609604 on Build GRCh38
rs796747598 Chr.9: 35609912-35609912 on Build GRCh38
rs373777485 Chr.9: 35609617-35609617 on Build GRCh38
rs773235606 Chr.9: 35609618-35609618 on Build GRCh38
rs1017768927 Chr.9: 35609740-35609740 on Build GRCh38
rs1056578984 Chr.9: 35609913-35609913 on Build GRCh38
rs567466527 Chr.9: 35609539-35609539 on Build GRCh38
rs111918879 Chr.9: 35609666-35609666 on Build GRCh38
rs370840118 Chr.9: 35609754-35609754 on Build GRCh38
rs765368705 Chr.9: 35609543-35609543 on Build GRCh38
rs200002717 Chr.9: 35609945-35609946 on Build GRCh38
rs926684616 Chr.9: 35609904-35609904 on Build GRCh38
rs138246140 Chr.9: 35609527-35609527 on Build GRCh38
rs544805818 Chr.9: 35609976-35609976 on Build GRCh38
rs761868564 Chr.9: 35609613-35609613 on Build GRCh38
rs974914243 Chr.9: 35609931-35609932 on Build GRCh38
rs557413044 Chr.9: 35609889-35609889 on Build GRCh38
rs769581228 Chr.9: 35609771-35609771 on Build GRCh38
rs767745383 Chr.9: 35609734-35609734 on Build GRCh38
rs200679265 Chr.9: 35609664-35609664 on Build GRCh38
rs979041322 Chr.9: 35609919-35609919 on Build GRCh38
rs768504971 Chr.9: 35609609-35609609 on Build GRCh38
rs1016821335 Chr.9: 35609554-35609554 on Build GRCh38
rs761147803 Chr.9: 35609738-35609738 on Build GRCh38
rs907923262 Chr.9: 35609950-35609950 on Build GRCh38
rs753052092 Chr.9: 35609540-35609540 on Build GRCh38
rs964916938 Chr.9: 35609928-35609928 on Build GRCh38
rs79361110 Chr.9: 35609549-35609549 on Build GRCh38
rs946704325 Chr.9: 35609875-35609875 on Build GRCh38
rs759922705 Chr.9: 35609906-35609907 on Build GRCh38
rs751151521 Chr.9: 35609757-35609757 on Build GRCh38
rs955875404 Chr.9: 35609843-35609843 on Build GRCh38
rs55673450 Chr.9: 35609581-35609581 on Build GRCh38
rs201230682 Chr.9: 35609548-35609548 on Build GRCh38
rs768324368 Chr.9: 35609684-35609684 on Build GRCh38
rs571781197 Chr.9: 35609857-35609857 on Build GRCh38
rs751820508 Chr.9: 35609557-35609557 on Build GRCh38
rs141508580 Chr.9: 35609708-35609708 on Build GRCh38
rs755380642 Chr.9: 35609559-35609559 on Build GRCh38
rs549408096 Chr.9: 35609630-35609630 on Build GRCh38
rs537640834 Chr.9: 35609877-35609877 on Build GRCh38
rs776645568 Chr.9: 35609536-35609536 on Build GRCh38
rs143842480 Chr.9: 35609547-35609547 on Build GRCh38
rs942050895 Chr.9: 35609963-35609963 on Build GRCh38
rs1005922004 Chr.9: 35609737-35609737 on Build GRCh38
rs747879770 Chr.9: 35609763-35609763 on Build GRCh38
rs749907092 Chr.9: 35609663-35609663 on Build GRCh38
rs779725799 Chr.9: 35609831-35609831 on Build GRCh38
rs751807950 Chr.9: 35609632-35609632 on Build GRCh38

View Full Product Details