Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35661415-35661945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF39
Entrez Gene ID
84904
Gene Name
Rho guanine nucleotide exchange factor 39
Gene Aliases
C9orf100
UniGene
Hs.534579
Gene Chromosome Location
Chr.9: 35658875-35665281 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015226.1 XM_017015227.1 NM_001195200.1 NM_001195201.1 NM_001195217.1 NM_174923.2

Gene Symbol
CCDC107
Entrez Gene ID
203260
Gene Name
coiled-coil domain containing 107
Gene Aliases
PSEC0222
UniGene
Hs.745107
Gene Chromosome Location
Chr.9: 35658282-35661503 on Build GRCh38
Transcripts
NM_032818.2 XM_011518057.2 XM_017015225.1 XM_017015226.1 XM_017015227.1 NM_001195200.1 NM_001195201.1 NM_001195217.1 NM_174923.2

SNP Detail

SNP ID Chromosome Location
rs183825465 Chr.9: 35661698-35661698 on Build GRCh38
rs940166329 Chr.9: 35661612-35661612 on Build GRCh38
rs964675015 Chr.9: 35661508-35661508 on Build GRCh38
rs1020693751 Chr.9: 35661439-35661440 on Build GRCh38
rs1032682955 Chr.9: 35661858-35661858 on Build GRCh38
rs531161831 Chr.9: 35661692-35661692 on Build GRCh38
rs955410351 Chr.9: 35661555-35661555 on Build GRCh38
rs940767181 Chr.9: 35661469-35661469 on Build GRCh38
rs79058295 Chr.9: 35661732-35661732 on Build GRCh38
rs112231054 Chr.9: 35661798-35661798 on Build GRCh38
rs958410728 Chr.9: 35661877-35661877 on Build GRCh38
rs565320939 Chr.9: 35661547-35661547 on Build GRCh38
rs140553174 Chr.9: 35661548-35661548 on Build GRCh38
rs896146541 Chr.9: 35661712-35661712 on Build GRCh38
rs187935173 Chr.9: 35661831-35661831 on Build GRCh38
rs56077544 Chr.9: 35661830-35661830 on Build GRCh38
rs1029118430 Chr.9: 35661529-35661529 on Build GRCh38
rs11546560 Chr.9: 35661448-35661448 on Build GRCh38
rs577712321 Chr.9: 35661862-35661862 on Build GRCh38
rs1013243166 Chr.9: 35661723-35661723 on Build GRCh38
rs375764017 Chr.9: 35661497-35661497 on Build GRCh38
rs193212801 Chr.9: 35661601-35661601 on Build GRCh38
rs1001076608 Chr.9: 35661759-35661759 on Build GRCh38
rs1000308916 Chr.9: 35661853-35661853 on Build GRCh38
rs369229378 Chr.9: 35661897-35661897 on Build GRCh38
rs529933416 Chr.9: 35661789-35661789 on Build GRCh38
rs982717607 Chr.9: 35661721-35661721 on Build GRCh38
rs978524011 Chr.9: 35661641-35661641 on Build GRCh38
rs938278755 Chr.9: 35661660-35661660 on Build GRCh38
rs76753837 Chr.9: 35661491-35661491 on Build GRCh38
rs902298628 Chr.9: 35661739-35661739 on Build GRCh38
rs986824429 Chr.9: 35661878-35661878 on Build GRCh38
rs534259608 Chr.9: 35661549-35661549 on Build GRCh38
rs991254555 Chr.9: 35661909-35661909 on Build GRCh38
rs940720269 Chr.9: 35661775-35661775 on Build GRCh38
rs949923460 Chr.9: 35661708-35661708 on Build GRCh38
rs144878718 Chr.9: 35661642-35661642 on Build GRCh38
rs1000727643 Chr.9: 35661483-35661483 on Build GRCh38
rs996338181 Chr.9: 35661512-35661512 on Build GRCh38
rs867754154 Chr.9: 35661833-35661833 on Build GRCh38
rs958562857 Chr.9: 35661663-35661663 on Build GRCh38
rs556562071 Chr.9: 35661691-35661691 on Build GRCh38
rs966511387 Chr.9: 35661449-35661449 on Build GRCh38
rs572319479 Chr.9: 35661814-35661814 on Build GRCh38
rs550987771 Chr.9: 35661813-35661813 on Build GRCh38
rs964948960 Chr.9: 35661793-35661793 on Build GRCh38
rs762587408 Chr.9: 35661887-35661887 on Build GRCh38
rs986875444 Chr.9: 35661556-35661556 on Build GRCh38
rs899203092 Chr.9: 35661807-35661807 on Build GRCh38
rs570785403 Chr.9: 35661614-35661614 on Build GRCh38
rs902841488 Chr.9: 35661849-35661849 on Build GRCh38
rs1050526849 Chr.9: 35661827-35661827 on Build GRCh38

View Full Product Details