Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 39132843-39133345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP3
Entrez Gene ID
79937
Gene Name
contactin associated protein-like 3
Gene Aliases
CASPR3, CNTNAP3A
UniGene
Hs.128474
Gene Chromosome Location
Chr.9: 39072767-39288303 on Build GRCh38
Transcripts
NM_033655.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6212064 c.1714G>A p.A572T Chr.9: 39133034-39133034 on Build GRCh38
4374438 c.1628C>T p.T543M Chr.9: 39133120-39133120 on Build GRCh38
6288090 c.1933G>T p.G645C Chr.9: 39133079-39133079 on Build GRCh38
7330534 c.1905C>T p.H635H Chr.9: 39133107-39133107 on Build GRCh38
2776253 c.1892C>T p.T631M Chr.9: 39133120-39133120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998433477 Chr.9: 39132958-39132958 on Build GRCh38
rs756606265 Chr.9: 39132982-39132982 on Build GRCh38
rs779455681 Chr.9: 39132990-39132990 on Build GRCh38
rs762041605 Chr.9: 39132885-39132885 on Build GRCh38
rs533123142 Chr.9: 39132924-39132924 on Build GRCh38
rs901466007 Chr.9: 39132955-39132955 on Build GRCh38
rs1010313709 Chr.9: 39132868-39132868 on Build GRCh38
rs765256460 Chr.9: 39132896-39132896 on Build GRCh38
rs960503231 Chr.9: 39133287-39133287 on Build GRCh38
rs138227053 Chr.9: 39132902-39132902 on Build GRCh38
rs770712561 Chr.9: 39132927-39132927 on Build GRCh38
rs1034839605 Chr.9: 39133274-39133274 on Build GRCh38
rs765932136 Chr.9: 39133156-39133156 on Build GRCh38
rs773389539 Chr.9: 39133029-39133029 on Build GRCh38
rs778705300 Chr.9: 39132984-39132984 on Build GRCh38
rs774300042 Chr.9: 39133038-39133038 on Build GRCh38
rs748123265 Chr.9: 39133093-39133093 on Build GRCh38
rs768477765 Chr.9: 39133005-39133005 on Build GRCh38
rs946038931 Chr.9: 39133080-39133080 on Build GRCh38
rs551040562 Chr.9: 39132863-39132863 on Build GRCh38
rs773012879 Chr.9: 39133095-39133095 on Build GRCh38
rs988029532 Chr.9: 39133289-39133289 on Build GRCh38
rs770529876 Chr.9: 39132906-39132907 on Build GRCh38
rs776427520 Chr.9: 39133009-39133009 on Build GRCh38
rs753324474 Chr.9: 39133060-39133060 on Build GRCh38
rs758022327 Chr.9: 39133070-39133070 on Build GRCh38
rs550440645 Chr.9: 39133062-39133062 on Build GRCh38
rs752083379 Chr.9: 39132966-39132966 on Build GRCh38
rs750051915 Chr.9: 39133069-39133069 on Build GRCh38
rs781458085 Chr.9: 39132919-39132919 on Build GRCh38
rs748029253 Chr.9: 39133179-39133179 on Build GRCh38
rs771071590 Chr.9: 39133110-39133110 on Build GRCh38
rs979261373 Chr.9: 39133089-39133089 on Build GRCh38
rs190162126 Chr.9: 39133035-39133035 on Build GRCh38
rs149597063 Chr.9: 39132977-39132977 on Build GRCh38
rs369580796 Chr.9: 39133166-39133166 on Build GRCh38
rs951242081 Chr.9: 39132890-39132890 on Build GRCh38
rs745358679 Chr.9: 39132935-39132935 on Build GRCh38
rs745716538 Chr.9: 39133039-39133039 on Build GRCh38
rs769642592 Chr.9: 39133025-39133025 on Build GRCh38
rs560148915 Chr.9: 39132954-39132954 on Build GRCh38
rs1001896185 Chr.9: 39133235-39133235 on Build GRCh38
rs763839735 Chr.9: 39132883-39132883 on Build GRCh38
rs904975087 Chr.9: 39133226-39133226 on Build GRCh38
rs778510909 Chr.9: 39132934-39132934 on Build GRCh38
rs1758272 Chr.9: 39133130-39133130 on Build GRCh38
rs373851232 Chr.9: 39133081-39133081 on Build GRCh38
rs554462365 Chr.9: 39133163-39133163 on Build GRCh38
rs1758270 Chr.9: 39133010-39133010 on Build GRCh38
rs549375588 Chr.9: 39133023-39133023 on Build GRCh38
rs760583489 Chr.9: 39132942-39132942 on Build GRCh38
rs769552862 Chr.9: 39133094-39133094 on Build GRCh38
rs554930194 Chr.9: 39133180-39133180 on Build GRCh38
rs752070599 Chr.9: 39132915-39132915 on Build GRCh38
rs909731620 Chr.9: 39132897-39132897 on Build GRCh38
rs111962901 Chr.9: 39133100-39133100 on Build GRCh38
rs754072161 Chr.9: 39132884-39132884 on Build GRCh38
rs745558298 Chr.9: 39132986-39132986 on Build GRCh38
rs879198183 Chr.9: 39132874-39132874 on Build GRCh38
rs1025314059 Chr.9: 39132889-39132889 on Build GRCh38
rs551656801 Chr.9: 39132946-39132946 on Build GRCh38
rs1020834171 Chr.9: 39133302-39133302 on Build GRCh38
rs746921118 Chr.9: 39133002-39133002 on Build GRCh38
rs1043644073 Chr.9: 39132869-39132869 on Build GRCh38
rs767677440 Chr.9: 39133119-39133119 on Build GRCh38
rs11794921 Chr.9: 39133034-39133034 on Build GRCh38
rs1758271 Chr.9: 39133111-39133111 on Build GRCh38
rs776326365 Chr.9: 39133029-39133030 on Build GRCh38
rs1017008253 Chr.9: 39133192-39133192 on Build GRCh38
rs886759823 Chr.9: 39132976-39132976 on Build GRCh38
rs1047287817 Chr.9: 39133143-39133143 on Build GRCh38
rs748930575 Chr.9: 39132920-39132920 on Build GRCh38
rs754255112 Chr.9: 39133146-39133146 on Build GRCh38
rs939268294 Chr.9: 39132903-39132903 on Build GRCh38
rs780524036 Chr.9: 39132908-39132908 on Build GRCh38
rs756175358 Chr.9: 39132892-39132892 on Build GRCh38
rs776121037 Chr.9: 39133128-39133128 on Build GRCh38
rs775470070 Chr.9: 39132941-39132941 on Build GRCh38
rs913218710 Chr.9: 39132906-39132906 on Build GRCh38
rs1692830 Chr.9: 39133171-39133171 on Build GRCh38
rs143310488 Chr.9: 39133152-39133152 on Build GRCh38
rs768452630 Chr.9: 39132943-39132943 on Build GRCh38
rs1018507111 Chr.9: 39132888-39132888 on Build GRCh38
rs934254974 Chr.9: 39133116-39133116 on Build GRCh38
rs767899158 Chr.9: 39133055-39133055 on Build GRCh38
rs749471152 Chr.9: 39133103-39133103 on Build GRCh38
rs539810470 Chr.9: 39132886-39132886 on Build GRCh38
rs755338006 Chr.9: 39132916-39132916 on Build GRCh38
rs992821979 Chr.9: 39132905-39132905 on Build GRCh38
rs573382335 Chr.9: 39132898-39132898 on Build GRCh38
rs573443441 Chr.9: 39133232-39133232 on Build GRCh38
rs764605578 Chr.9: 39133068-39133068 on Build GRCh38
rs760030789 Chr.9: 39133054-39133054 on Build GRCh38
rs960639812 Chr.9: 39133030-39133030 on Build GRCh38
rs764517924 Chr.9: 39133138-39133138 on Build GRCh38
rs561470022 Chr.9: 39132888-39132888 on Build GRCh38
rs771689905 Chr.9: 39132989-39132989 on Build GRCh38
rs754572269 Chr.9: 39133088-39133088 on Build GRCh38
rs571811737 Chr.9: 39133092-39133092 on Build GRCh38
rs779546708 Chr.9: 39133078-39133078 on Build GRCh38
rs767217792 Chr.9: 39132957-39132957 on Build GRCh38
rs544039196 Chr.9: 39132899-39132899 on Build GRCh38

View Full Product Details