Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113424076-113424510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
UniGene
Hs.632691
Gene Chromosome Location
Chr.9: 113410644-113429684 on Build GRCh38
Transcripts
NM_001278629.1 NM_001278630.1 NM_152786.2 XM_005251885.3 XM_011518477.1 XM_011518478.1 XM_011518479.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
455188 c.791A>G p.H264R Chr.9: 113424300-113424300 on Build GRCh38
5063542 c.748C>T p.R250* Chr.9: 113424257-113424257 on Build GRCh38
1104236 c.680C>T p.T227I Chr.9: 113424189-113424189 on Build GRCh38
3432788 c.729T>A p.N243K Chr.9: 113424238-113424238 on Build GRCh38
357118 c.667G>T p.G223C Chr.9: 113424176-113424176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147595352 Chr.9: 113424232-113424232 on Build GRCh38
rs148905915 Chr.9: 113424302-113424302 on Build GRCh38
rs973365549 Chr.9: 113424416-113424416 on Build GRCh38
rs776996638 Chr.9: 113424169-113424169 on Build GRCh38
rs1025134679 Chr.9: 113424332-113424332 on Build GRCh38
rs779802964 Chr.9: 113424349-113424349 on Build GRCh38
rs770388466 Chr.9: 113424242-113424242 on Build GRCh38
rs781164565 Chr.9: 113424124-113424124 on Build GRCh38
rs770152149 Chr.9: 113424364-113424364 on Build GRCh38
rs777818445 Chr.9: 113424237-113424237 on Build GRCh38
rs757203921 Chr.9: 113424317-113424317 on Build GRCh38
rs1031980741 Chr.9: 113424286-113424286 on Build GRCh38
rs772724913 Chr.9: 113424127-113424132 on Build GRCh38
rs372912068 Chr.9: 113424196-113424196 on Build GRCh38
rs749207190 Chr.9: 113424240-113424240 on Build GRCh38
rs1049547399 Chr.9: 113424171-113424171 on Build GRCh38
rs147812340 Chr.9: 113424291-113424291 on Build GRCh38
rs766500320 Chr.9: 113424273-113424273 on Build GRCh38
rs776857944 Chr.9: 113424258-113424258 on Build GRCh38
rs755255479 Chr.9: 113424214-113424214 on Build GRCh38
rs567396522 Chr.9: 113424337-113424337 on Build GRCh38
rs770865384 Chr.9: 113424342-113424343 on Build GRCh38
rs183431270 Chr.9: 113424435-113424435 on Build GRCh38
rs758017506 Chr.9: 113424341-113424341 on Build GRCh38
rs900561583 Chr.9: 113424156-113424156 on Build GRCh38
rs763827480 Chr.9: 113424314-113424314 on Build GRCh38
rs191657183 Chr.9: 113424357-113424357 on Build GRCh38
rs200435793 Chr.9: 113424132-113424132 on Build GRCh38
rs755567204 Chr.9: 113424459-113424459 on Build GRCh38
rs971333776 Chr.9: 113424350-113424350 on Build GRCh38
rs983704982 Chr.9: 113424352-113424352 on Build GRCh38
rs755891099 Chr.9: 113424303-113424303 on Build GRCh38
rs754194807 Chr.9: 113424441-113424441 on Build GRCh38
rs866292730 Chr.9: 113424218-113424218 on Build GRCh38
rs990569751 Chr.9: 113424323-113424323 on Build GRCh38
rs370448386 Chr.9: 113424119-113424119 on Build GRCh38
rs762216326 Chr.9: 113424176-113424176 on Build GRCh38
rs773945256 Chr.9: 113424155-113424155 on Build GRCh38
rs372042993 Chr.9: 113424153-113424153 on Build GRCh38
rs375387134 Chr.9: 113424194-113424194 on Build GRCh38
rs549570387 Chr.9: 113424111-113424111 on Build GRCh38
rs773715977 Chr.9: 113424267-113424267 on Build GRCh38
rs773858283 Chr.9: 113424140-113424140 on Build GRCh38
rs374079843 Chr.9: 113424131-113424131 on Build GRCh38
rs759218719 Chr.9: 113424159-113424159 on Build GRCh38
rs376222703 Chr.9: 113424367-113424367 on Build GRCh38
rs778384886 Chr.9: 113424329-113424329 on Build GRCh38
rs766661107 Chr.9: 113424431-113424431 on Build GRCh38
rs755999475 Chr.9: 113424236-113424236 on Build GRCh38
rs765821232 Chr.9: 113424465-113424465 on Build GRCh38
rs368689694 Chr.9: 113424195-113424195 on Build GRCh38
rs762276425 Chr.9: 113424260-113424260 on Build GRCh38
rs745469839 Chr.9: 113424257-113424257 on Build GRCh38
rs551801116 Chr.9: 113424213-113424213 on Build GRCh38
rs778399315 Chr.9: 113424243-113424243 on Build GRCh38
rs748871282 Chr.9: 113424141-113424141 on Build GRCh38
rs1008137038 Chr.9: 113424212-113424212 on Build GRCh38
rs942270372 Chr.9: 113424390-113424390 on Build GRCh38
rs932053103 Chr.9: 113424460-113424460 on Build GRCh38
rs767916368 Chr.9: 113424301-113424301 on Build GRCh38
rs781076077 Chr.9: 113424220-113424220 on Build GRCh38
rs548025184 Chr.9: 113424142-113424145 on Build GRCh38
rs201657547 Chr.9: 113424299-113424299 on Build GRCh38
rs762416045 Chr.9: 113424120-113424120 on Build GRCh38
rs41313331 Chr.9: 113424264-113424264 on Build GRCh38
rs1004630876 Chr.9: 113424396-113424396 on Build GRCh38
rs763396807 Chr.9: 113424268-113424268 on Build GRCh38
rs528759702 Chr.9: 113424137-113424137 on Build GRCh38

View Full Product Details