Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15749893-15750396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA5B
Entrez Gene ID
11238
Gene Name
carbonic anhydrase 5B
Gene Aliases
CA-VB
UniGene
Hs.653287
Gene Chromosome Location
Chr.X: 15738281-15787625 on Build GRCh38
Transcripts
NM_007220.3 XM_005274442.4 XM_017029249.1

SNP Detail

SNP ID Chromosome Location
rs779402154 Chr.X: 15750054-15750054 on Build GRCh38
rs150225380 Chr.X: 15750030-15750030 on Build GRCh38
rs757255721 Chr.X: 15750062-15750062 on Build GRCh38
rs12013305 Chr.X: 15750270-15750270 on Build GRCh38
rs759916192 Chr.X: 15750125-15750125 on Build GRCh38
rs755830090 Chr.X: 15750150-15750150 on Build GRCh38
rs752334253 Chr.X: 15749988-15749988 on Build GRCh38
rs923984198 Chr.X: 15750272-15750272 on Build GRCh38
rs906967930 Chr.X: 15749956-15749956 on Build GRCh38
rs1047212150 Chr.X: 15750364-15750364 on Build GRCh38
rs758160020 Chr.X: 15750201-15750201 on Build GRCh38
rs748338398 Chr.X: 15750087-15750087 on Build GRCh38
rs771799122 Chr.X: 15750080-15750080 on Build GRCh38
rs376834870 Chr.X: 15750159-15750159 on Build GRCh38
rs987070256 Chr.X: 15750182-15750182 on Build GRCh38
rs191865386 Chr.X: 15750354-15750354 on Build GRCh38
rs200179565 Chr.X: 15750083-15750083 on Build GRCh38
rs756873583 Chr.X: 15750163-15750163 on Build GRCh38
rs1053709036 Chr.X: 15750302-15750302 on Build GRCh38
rs751013469 Chr.X: 15749946-15749946 on Build GRCh38
rs780876826 Chr.X: 15749986-15749986 on Build GRCh38
rs975832461 Chr.X: 15750288-15750288 on Build GRCh38
rs748223781 Chr.X: 15750254-15750254 on Build GRCh38
rs187875359 Chr.X: 15749967-15749967 on Build GRCh38
rs745821056 Chr.X: 15750169-15750169 on Build GRCh38
rs769809997 Chr.X: 15750100-15750100 on Build GRCh38
rs777795853 Chr.X: 15750218-15750219 on Build GRCh38
rs964065779 Chr.X: 15750280-15750280 on Build GRCh38
rs749169372 Chr.X: 15750057-15750057 on Build GRCh38
rs919984695 Chr.X: 15750336-15750336 on Build GRCh38
rs778982825 Chr.X: 15750222-15750222 on Build GRCh38
rs767804694 Chr.X: 15750135-15750135 on Build GRCh38
rs1020420652 Chr.X: 15750133-15750133 on Build GRCh38
rs1012404745 Chr.X: 15749932-15749932 on Build GRCh38
rs777433637 Chr.X: 15750038-15750038 on Build GRCh38
rs748363071 Chr.X: 15750065-15750066 on Build GRCh38
rs754041531 Chr.X: 15750101-15750101 on Build GRCh38
rs766858791 Chr.X: 15750115-15750115 on Build GRCh38
rs368039748 Chr.X: 15750103-15750103 on Build GRCh38
rs1001636371 Chr.X: 15749944-15749944 on Build GRCh38
rs755418977 Chr.X: 15749982-15749982 on Build GRCh38
rs752334695 Chr.X: 15750146-15750146 on Build GRCh38
rs763906499 Chr.X: 15750154-15750154 on Build GRCh38
rs35387885 Chr.X: 15750157-15750158 on Build GRCh38
rs745649867 Chr.X: 15750073-15750073 on Build GRCh38
rs766685420 Chr.X: 15750064-15750064 on Build GRCh38
rs778958796 Chr.X: 15750168-15750168 on Build GRCh38
rs547815978 Chr.X: 15749969-15749969 on Build GRCh38
rs755612115 Chr.X: 15750032-15750032 on Build GRCh38
rs772339115 Chr.X: 15750092-15750092 on Build GRCh38
rs200064337 Chr.X: 15750116-15750116 on Build GRCh38

View Full Product Details