Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 31120299-31120846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMD
Entrez Gene ID
1756
Gene Name
dystrophin
Gene Aliases
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
UniGene
Hs.495912
Gene Chromosome Location
Chr.X: 31119219-33339609 on Build GRCh38
Transcripts
NM_000109.3 NM_004006.2 NM_004009.3 NM_004010.3 NM_004011.3 NM_004012.3 NM_004013.2 NM_004014.2 NM_004015.2 NM_004016.2 NM_004017.2 NM_004018.2 NM_004020.3 NM_004021.2 NM_004022.2 NM_004023.2 XM_006724468.2 XM_006724469.3 XM_006724470.3 XM_006724473.2 XM_006724474.3 XM_006724475.2 XM_011545467.1 XM_017029328.1 XM_017029331.1

SNP Detail

SNP ID Chromosome Location
rs1013604841 Chr.X: 31120607-31120607 on Build GRCh38
rs142110702 Chr.X: 31120634-31120634 on Build GRCh38
rs962211140 Chr.X: 31120686-31120686 on Build GRCh38
rs1022600093 Chr.X: 31120580-31120580 on Build GRCh38
rs771302445 Chr.X: 31120354-31120357 on Build GRCh38
rs758612124 Chr.X: 31120357-31120358 on Build GRCh38
rs145871126 Chr.X: 31120689-31120689 on Build GRCh38
rs777231552 Chr.X: 31120732-31120732 on Build GRCh38
rs747497461 Chr.X: 31120753-31120754 on Build GRCh38
rs933008424 Chr.X: 31120429-31120429 on Build GRCh38
rs767372350 Chr.X: 31120526-31120526 on Build GRCh38
rs774861121 Chr.X: 31120452-31120452 on Build GRCh38
rs1036051178 Chr.X: 31120478-31120478 on Build GRCh38
rs764825924 Chr.X: 31120570-31120570 on Build GRCh38
rs771473362 Chr.X: 31120798-31120799 on Build GRCh38
rs1016424213 Chr.X: 31120681-31120681 on Build GRCh38
rs993426832 Chr.X: 31120470-31120471 on Build GRCh38
rs191747923 Chr.X: 31120464-31120464 on Build GRCh38
rs200311985 Chr.X: 31120730-31120731 on Build GRCh38
rs985484114 Chr.X: 31120663-31120663 on Build GRCh38
rs767994823 Chr.X: 31120386-31120386 on Build GRCh38
rs1024272193 Chr.X: 31120629-31120629 on Build GRCh38
rs923480339 Chr.X: 31120748-31120748 on Build GRCh38
rs556300211 Chr.X: 31120649-31120649 on Build GRCh38
rs968425458 Chr.X: 31120586-31120586 on Build GRCh38
rs752222680 Chr.X: 31120591-31120591 on Build GRCh38
rs1050391634 Chr.X: 31120758-31120759 on Build GRCh38
rs978503404 Chr.X: 31120639-31120639 on Build GRCh38
rs1057515857 Chr.X: 31120490-31120491 on Build GRCh38
rs750744246 Chr.X: 31120419-31120419 on Build GRCh38
rs759171595 Chr.X: 31120432-31120432 on Build GRCh38
rs1053155218 Chr.X: 31120350-31120351 on Build GRCh38
rs750283076 Chr.X: 31120606-31120606 on Build GRCh38
rs771242421 Chr.X: 31120327-31120328 on Build GRCh38
rs752873899 Chr.X: 31120637-31120637 on Build GRCh38
rs935530958 Chr.X: 31120701-31120705 on Build GRCh38
rs1042894098 Chr.X: 31120515-31120515 on Build GRCh38
rs1057515856 Chr.X: 31120470-31120471 on Build GRCh38
rs889183676 Chr.X: 31120438-31120439 on Build GRCh38
rs942074454 Chr.X: 31120775-31120775 on Build GRCh38
rs933285670 Chr.X: 31120384-31120384 on Build GRCh38
rs182383454 Chr.X: 31120518-31120518 on Build GRCh38
rs1057515858 Chr.X: 31120580-31120580 on Build GRCh38
rs1016091957 Chr.X: 31120465-31120465 on Build GRCh38
rs187318961 Chr.X: 31120737-31120737 on Build GRCh38
rs755247077 Chr.X: 31120610-31120611 on Build GRCh38
rs923039797 Chr.X: 31120749-31120749 on Build GRCh38
rs901834311 Chr.X: 31120357-31120357 on Build GRCh38
rs897676638 Chr.X: 31120468-31120468 on Build GRCh38
rs932956201 Chr.X: 31120754-31120754 on Build GRCh38
rs935576250 Chr.X: 31120334-31120334 on Build GRCh38
rs3361 Chr.X: 31120472-31120472 on Build GRCh38
rs985817741 Chr.X: 31120777-31120777 on Build GRCh38
rs763527539 Chr.X: 31120469-31120469 on Build GRCh38
rs922925467 Chr.X: 31120420-31120420 on Build GRCh38
rs764913815 Chr.X: 31120440-31120441 on Build GRCh38
rs933608816 Chr.X: 31120754-31120755 on Build GRCh38
rs1003825974 Chr.X: 31120763-31120789 on Build GRCh38
rs1057515859 Chr.X: 31120754-31120755 on Build GRCh38
rs866894549 Chr.X: 31120353-31120353 on Build GRCh38

View Full Product Details