Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 45149149-45149645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf36
Entrez Gene ID
79742
Gene Name
chromosome X open reading frame 36
Gene Aliases
4930578C19Rik, DIA1R, EPQL1862, PRO3743, bA435K1.1
UniGene
Hs.98321
Gene Chromosome Location
Chr.X: 45148373-45200901 on Build GRCh38
Transcripts
NM_176819.3 XM_005272670.1 XM_006724559.1

SNP Detail

SNP ID Chromosome Location
rs766853307 Chr.X: 45149239-45149240 on Build GRCh38
rs946953813 Chr.X: 45149371-45149371 on Build GRCh38
rs752448195 Chr.X: 45149582-45149582 on Build GRCh38
rs777130181 Chr.X: 45149275-45149275 on Build GRCh38
rs1006113505 Chr.X: 45149550-45149550 on Build GRCh38
rs1044294897 Chr.X: 45149372-45149372 on Build GRCh38
rs551898198 Chr.X: 45149491-45149491 on Build GRCh38
rs933970104 Chr.X: 45149192-45149192 on Build GRCh38
rs1052726632 Chr.X: 45149243-45149243 on Build GRCh38
rs893107436 Chr.X: 45149416-45149416 on Build GRCh38
rs575465638 Chr.X: 45149568-45149568 on Build GRCh38
rs767266212 Chr.X: 45149438-45149438 on Build GRCh38
rs975937029 Chr.X: 45149600-45149600 on Build GRCh38
rs1002393845 Chr.X: 45149421-45149421 on Build GRCh38
rs1053151423 Chr.X: 45149406-45149406 on Build GRCh38
rs758532662 Chr.X: 45149477-45149477 on Build GRCh38
rs914158668 Chr.X: 45149329-45149329 on Build GRCh38
rs752757003 Chr.X: 45149349-45149349 on Build GRCh38
rs922626993 Chr.X: 45149174-45149174 on Build GRCh38
rs764027694 Chr.X: 45149502-45149502 on Build GRCh38
rs975503675 Chr.X: 45149173-45149173 on Build GRCh38
rs896197157 Chr.X: 45149460-45149460 on Build GRCh38
rs905334898 Chr.X: 45149400-45149400 on Build GRCh38
rs1026386943 Chr.X: 45149171-45149171 on Build GRCh38
rs754602589 Chr.X: 45149417-45149417 on Build GRCh38
rs765750319 Chr.X: 45149318-45149318 on Build GRCh38
rs751797152 Chr.X: 45149527-45149527 on Build GRCh38
rs762307217 Chr.X: 45149618-45149618 on Build GRCh38
rs944595603 Chr.X: 45149573-45149573 on Build GRCh38
rs928682485 Chr.X: 45149268-45149268 on Build GRCh38
rs981529937 Chr.X: 45149256-45149256 on Build GRCh38
rs760176446 Chr.X: 45149313-45149313 on Build GRCh38
rs915783039 Chr.X: 45149188-45149188 on Build GRCh38
rs952020719 Chr.X: 45149172-45149172 on Build GRCh38
rs1008311377 Chr.X: 45149557-45149557 on Build GRCh38
rs36053572 Chr.X: 45149388-45149389 on Build GRCh38
rs901623224 Chr.X: 45149596-45149596 on Build GRCh38

View Full Product Details