Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 45149383-45149878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf36
Entrez Gene ID
79742
Gene Name
chromosome X open reading frame 36
Gene Aliases
4930578C19Rik, DIA1R, EPQL1862, PRO3743, bA435K1.1
UniGene
Hs.98321
Gene Chromosome Location
Chr.X: 45148373-45200901 on Build GRCh38
Transcripts
NM_176819.3 XM_005272670.1 XM_006724559.1

SNP Detail

SNP ID Chromosome Location
rs752448195 Chr.X: 45149582-45149582 on Build GRCh38
rs1006113505 Chr.X: 45149550-45149550 on Build GRCh38
rs988146770 Chr.X: 45149666-45149666 on Build GRCh38
rs551898198 Chr.X: 45149491-45149491 on Build GRCh38
rs981246186 Chr.X: 45149788-45149788 on Build GRCh38
rs955402327 Chr.X: 45149662-45149662 on Build GRCh38
rs893107436 Chr.X: 45149416-45149416 on Build GRCh38
rs1022850643 Chr.X: 45149739-45149739 on Build GRCh38
rs951795078 Chr.X: 45149642-45149642 on Build GRCh38
rs575465638 Chr.X: 45149568-45149568 on Build GRCh38
rs150465715 Chr.X: 45149697-45149697 on Build GRCh38
rs767266212 Chr.X: 45149438-45149438 on Build GRCh38
rs993179905 Chr.X: 45149787-45149787 on Build GRCh38
rs926820247 Chr.X: 45149750-45149750 on Build GRCh38
rs1047403814 Chr.X: 45149802-45149802 on Build GRCh38
rs975937029 Chr.X: 45149600-45149600 on Build GRCh38
rs1002393845 Chr.X: 45149421-45149421 on Build GRCh38
rs778880384 Chr.X: 45149810-45149810 on Build GRCh38
rs1053151423 Chr.X: 45149406-45149406 on Build GRCh38
rs758532662 Chr.X: 45149477-45149477 on Build GRCh38
rs141530450 Chr.X: 45149669-45149669 on Build GRCh38
rs969899881 Chr.X: 45149775-45149775 on Build GRCh38
rs764027694 Chr.X: 45149502-45149502 on Build GRCh38
rs896197157 Chr.X: 45149460-45149460 on Build GRCh38
rs754602589 Chr.X: 45149417-45149417 on Build GRCh38
rs938193539 Chr.X: 45149761-45149761 on Build GRCh38
rs751797152 Chr.X: 45149527-45149527 on Build GRCh38
rs762307217 Chr.X: 45149618-45149618 on Build GRCh38
rs944595603 Chr.X: 45149573-45149573 on Build GRCh38
rs946975204 Chr.X: 45149675-45149675 on Build GRCh38
rs1056520074 Chr.X: 45149782-45149782 on Build GRCh38
rs1008311377 Chr.X: 45149557-45149557 on Build GRCh38
rs138260277 Chr.X: 45149786-45149786 on Build GRCh38
rs73477120 Chr.X: 45149638-45149638 on Build GRCh38
rs901623224 Chr.X: 45149596-45149596 on Build GRCh38
rs1006152336 Chr.X: 45149657-45149657 on Build GRCh38

View Full Product Details