Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46574303-46574804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST7
Entrez Gene ID
56548
Gene Name
carbohydrate sulfotransferase 7
Gene Aliases
C6ST-2, GST-5
UniGene
Hs.129955
Gene Chromosome Location
Chr.X: 46573687-46598496 on Build GRCh38
Transcripts
NM_019886.3

SNP Detail

SNP ID Chromosome Location
rs773697310 Chr.X: 46574445-46574445 on Build GRCh38
rs774849883 Chr.X: 46574579-46574579 on Build GRCh38
rs777553106 Chr.X: 46574369-46574369 on Build GRCh38
rs150774371 Chr.X: 46574691-46574691 on Build GRCh38
rs757997517 Chr.X: 46574488-46574488 on Build GRCh38
rs746500973 Chr.X: 46574534-46574534 on Build GRCh38
rs778970910 Chr.X: 46574402-46574402 on Build GRCh38
rs745409653 Chr.X: 46574407-46574407 on Build GRCh38
rs181984333 Chr.X: 46574464-46574464 on Build GRCh38
rs749031627 Chr.X: 46574372-46574372 on Build GRCh38
rs751134611 Chr.X: 46574721-46574721 on Build GRCh38
rs376196221 Chr.X: 46574387-46574387 on Build GRCh38
rs766923926 Chr.X: 46574452-46574452 on Build GRCh38
rs752917425 Chr.X: 46574476-46574476 on Build GRCh38
rs764208188 Chr.X: 46574482-46574482 on Build GRCh38
rs112707438 Chr.X: 46574584-46574584 on Build GRCh38
rs772559971 Chr.X: 46574607-46574607 on Build GRCh38
rs146831351 Chr.X: 46574729-46574729 on Build GRCh38
rs780932142 Chr.X: 46574723-46574723 on Build GRCh38
rs779724875 Chr.X: 46574715-46574715 on Build GRCh38
rs61734022 Chr.X: 46574447-46574447 on Build GRCh38
rs904597845 Chr.X: 46574742-46574742 on Build GRCh38
rs767525052 Chr.X: 46574468-46574468 on Build GRCh38
rs141240078 Chr.X: 46574648-46574648 on Build GRCh38
rs775696947 Chr.X: 46574611-46574611 on Build GRCh38
rs761724283 Chr.X: 46574666-46574666 on Build GRCh38
rs746614653 Chr.X: 46574420-46574420 on Build GRCh38
rs774424444 Chr.X: 46574483-46574483 on Build GRCh38
rs754417837 Chr.X: 46574722-46574722 on Build GRCh38
rs867218695 Chr.X: 46574733-46574733 on Build GRCh38
rs771287502 Chr.X: 46574773-46574773 on Build GRCh38
rs779640252 Chr.X: 46574533-46574533 on Build GRCh38
rs749705527 Chr.X: 46574566-46574566 on Build GRCh38
rs752864505 Chr.X: 46574350-46574350 on Build GRCh38
rs138804520 Chr.X: 46574585-46574585 on Build GRCh38
rs779313210 Chr.X: 46574779-46574779 on Build GRCh38
rs371271901 Chr.X: 46574484-46574484 on Build GRCh38
rs377476475 Chr.X: 46574542-46574542 on Build GRCh38
rs764415985 Chr.X: 46574629-46574629 on Build GRCh38
rs755870456 Chr.X: 46574365-46574365 on Build GRCh38
rs770059621 Chr.X: 46574439-46574439 on Build GRCh38
rs956147595 Chr.X: 46574340-46574340 on Build GRCh38
rs760649108 Chr.X: 46574624-46574624 on Build GRCh38
rs774811144 Chr.X: 46574453-46574453 on Build GRCh38
rs771234285 Chr.X: 46574572-46574572 on Build GRCh38
rs750391262 Chr.X: 46574675-46574675 on Build GRCh38
rs921002483 Chr.X: 46574459-46574459 on Build GRCh38
rs761449271 Chr.X: 46574335-46574335 on Build GRCh38
rs780688599 Chr.X: 46574565-46574565 on Build GRCh38
rs765069576 Chr.X: 46574670-46574670 on Build GRCh38
rs771669492 Chr.X: 46574408-46574408 on Build GRCh38
rs895751075 Chr.X: 46574627-46574627 on Build GRCh38
rs761020258 Chr.X: 46574477-46574477 on Build GRCh38
rs780129932 Chr.X: 46574700-46574700 on Build GRCh38

View Full Product Details