Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47636243-47636751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK1
Entrez Gene ID
2002
Gene Name
ELK1, ETS transcription factor
Gene Aliases
-
UniGene
Hs.181128
Gene Chromosome Location
Chr.X: 47635520-47650604 on Build GRCh38
Transcripts
NM_001114123.2 NM_001257168.1 NM_005229.4 XM_017029339.1

SNP Detail

SNP ID Chromosome Location
rs902374332 Chr.X: 47636494-47636494 on Build GRCh38
rs751606012 Chr.X: 47636592-47636592 on Build GRCh38
rs1028329600 Chr.X: 47636569-47636569 on Build GRCh38
rs373585813 Chr.X: 47636570-47636570 on Build GRCh38
rs957932578 Chr.X: 47636550-47636550 on Build GRCh38
rs777403536 Chr.X: 47636276-47636276 on Build GRCh38
rs1025117876 Chr.X: 47636437-47636437 on Build GRCh38
rs773977334 Chr.X: 47636647-47636647 on Build GRCh38
rs898731139 Chr.X: 47636325-47636325 on Build GRCh38
rs1054729404 Chr.X: 47636370-47636370 on Build GRCh38
rs986915684 Chr.X: 47636593-47636593 on Build GRCh38
rs748738498 Chr.X: 47636486-47636486 on Build GRCh38
rs766329948 Chr.X: 47636272-47636272 on Build GRCh38
rs961588535 Chr.X: 47636616-47636616 on Build GRCh38
rs1013356869 Chr.X: 47636425-47636428 on Build GRCh38
rs974623962 Chr.X: 47636566-47636566 on Build GRCh38
rs757417398 Chr.X: 47636633-47636633 on Build GRCh38
rs763300066 Chr.X: 47636555-47636555 on Build GRCh38
rs527444297 Chr.X: 47636421-47636421 on Build GRCh38
rs931569925 Chr.X: 47636668-47636668 on Build GRCh38
rs912622657 Chr.X: 47636602-47636602 on Build GRCh38
rs752922243 Chr.X: 47636554-47636554 on Build GRCh38
rs1032300376 Chr.X: 47636537-47636537 on Build GRCh38
rs376802243 Chr.X: 47636576-47636576 on Build GRCh38
rs999749286 Chr.X: 47636530-47636530 on Build GRCh38
rs894776751 Chr.X: 47636382-47636382 on Build GRCh38
rs1055281570 Chr.X: 47636669-47636669 on Build GRCh38
rs931016922 Chr.X: 47636363-47636363 on Build GRCh38
rs764168665 Chr.X: 47636559-47636559 on Build GRCh38
rs973306344 Chr.X: 47636628-47636629 on Build GRCh38

View Full Product Details