Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47637659-47638164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK1
Entrez Gene ID
2002
Gene Name
ELK1, ETS transcription factor
Gene Aliases
-
UniGene
Hs.181128
Gene Chromosome Location
Chr.X: 47635520-47650604 on Build GRCh38
Transcripts
NM_001114123.2 NM_001257168.1 NM_005229.4 XM_017029339.1

SNP Detail

SNP ID Chromosome Location
rs773489927 Chr.X: 47637990-47637990 on Build GRCh38
rs770629454 Chr.X: 47637925-47637925 on Build GRCh38
rs751772833 Chr.X: 47638001-47638001 on Build GRCh38
rs371259584 Chr.X: 47638113-47638113 on Build GRCh38
rs200141730 Chr.X: 47638124-47638124 on Build GRCh38
rs763316945 Chr.X: 47637867-47637867 on Build GRCh38
rs747375894 Chr.X: 47637962-47637962 on Build GRCh38
rs780006644 Chr.X: 47637760-47637760 on Build GRCh38
rs762419349 Chr.X: 47637973-47637973 on Build GRCh38
rs201698802 Chr.X: 47638057-47638057 on Build GRCh38
rs779320553 Chr.X: 47637716-47637716 on Build GRCh38
rs750560181 Chr.X: 47637744-47637744 on Build GRCh38
rs369027699 Chr.X: 47637967-47637967 on Build GRCh38
rs111614510 Chr.X: 47637817-47637817 on Build GRCh38
rs778250617 Chr.X: 47638110-47638110 on Build GRCh38
rs751368233 Chr.X: 47637847-47637847 on Build GRCh38
rs762609294 Chr.X: 47637794-47637794 on Build GRCh38
rs756731775 Chr.X: 47638102-47638102 on Build GRCh38
rs777167555 Chr.X: 47637806-47637806 on Build GRCh38
rs765426882 Chr.X: 47637809-47637809 on Build GRCh38
rs777459618 Chr.X: 47637941-47637941 on Build GRCh38
rs748793978 Chr.X: 47637904-47637904 on Build GRCh38
rs766899855 Chr.X: 47638123-47638123 on Build GRCh38
rs778401778 Chr.X: 47637800-47637800 on Build GRCh38
rs769100410 Chr.X: 47637965-47637965 on Build GRCh38
rs774035251 Chr.X: 47637947-47637947 on Build GRCh38
rs971394376 Chr.X: 47637907-47637907 on Build GRCh38
rs756113016 Chr.X: 47638053-47638053 on Build GRCh38
rs376201899 Chr.X: 47637865-47637865 on Build GRCh38
rs373267605 Chr.X: 47637974-47637974 on Build GRCh38
rs376822462 Chr.X: 47638112-47638112 on Build GRCh38
rs758694016 Chr.X: 47637834-47637834 on Build GRCh38
rs199829517 Chr.X: 47637872-47637872 on Build GRCh38
rs747885568 Chr.X: 47637813-47637813 on Build GRCh38
rs755425548 Chr.X: 47637787-47637787 on Build GRCh38
rs1008259950 Chr.X: 47637725-47637725 on Build GRCh38
rs752423073 Chr.X: 47638027-47638027 on Build GRCh38
rs775785466 Chr.X: 47637797-47637797 on Build GRCh38
rs772662025 Chr.X: 47637788-47637788 on Build GRCh38
rs764561959 Chr.X: 47637802-47637802 on Build GRCh38
rs754780539 Chr.X: 47637863-47637863 on Build GRCh38
rs765768273 Chr.X: 47637975-47637975 on Build GRCh38
rs371893100 Chr.X: 47637784-47637784 on Build GRCh38
rs1027014686 Chr.X: 47637798-47637798 on Build GRCh38
rs755114172 Chr.X: 47638007-47638007 on Build GRCh38
rs770504652 Chr.X: 47637795-47637795 on Build GRCh38
rs200172068 Chr.X: 47638009-47638009 on Build GRCh38
rs757341998 Chr.X: 47637707-47637707 on Build GRCh38
rs777270278 Chr.X: 47637892-47637892 on Build GRCh38
rs1023924488 Chr.X: 47637895-47637895 on Build GRCh38
rs952906295 Chr.X: 47637807-47637807 on Build GRCh38
rs987401033 Chr.X: 47637686-47637686 on Build GRCh38
rs766616373 Chr.X: 47637846-47637846 on Build GRCh38
rs772779662 Chr.X: 47637727-47637727 on Build GRCh38
rs766556938 Chr.X: 47637995-47637995 on Build GRCh38
rs376525319 Chr.X: 47637993-47637993 on Build GRCh38
rs910491693 Chr.X: 47637922-47637922 on Build GRCh38
rs762372393 Chr.X: 47637808-47637808 on Build GRCh38
rs765230225 Chr.X: 47637737-47637737 on Build GRCh38
rs746747467 Chr.X: 47637762-47637762 on Build GRCh38

View Full Product Details