Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47650342-47650832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK1
Entrez Gene ID
2002
Gene Name
ELK1, ETS transcription factor
Gene Aliases
-
UniGene
Hs.181128
Gene Chromosome Location
Chr.X: 47635520-47650604 on Build GRCh38
Transcripts
NM_001114123.2 NM_001257168.1 NM_005229.4

SNP Detail

SNP ID Chromosome Location
rs754812337 Chr.X: 47650556-47650556 on Build GRCh38
rs1042361384 Chr.X: 47650595-47650595 on Build GRCh38
rs780776015 Chr.X: 47650726-47650726 on Build GRCh38
rs758915461 Chr.X: 47650531-47650533 on Build GRCh38
rs761400530 Chr.X: 47650769-47650769 on Build GRCh38
rs769492715 Chr.X: 47650568-47650568 on Build GRCh38
rs762725923 Chr.X: 47650548-47650549 on Build GRCh38
rs761251161 Chr.X: 47650581-47650581 on Build GRCh38
rs768347522 Chr.X: 47650694-47650694 on Build GRCh38
rs966424600 Chr.X: 47650512-47650512 on Build GRCh38
rs747651388 Chr.X: 47650565-47650565 on Build GRCh38
rs747582963 Chr.X: 47650536-47650537 on Build GRCh38
rs192596929 Chr.X: 47650624-47650624 on Build GRCh38
rs769340100 Chr.X: 47650542-47650543 on Build GRCh38
rs953442948 Chr.X: 47650530-47650530 on Build GRCh38
rs776068314 Chr.X: 47650527-47650527 on Build GRCh38
rs772510344 Chr.X: 47650575-47650575 on Build GRCh38
rs906236174 Chr.X: 47650586-47650586 on Build GRCh38
rs776072843 Chr.X: 47650749-47650749 on Build GRCh38
rs764695916 Chr.X: 47650602-47650602 on Build GRCh38
rs902128990 Chr.X: 47650625-47650625 on Build GRCh38
rs774763711 Chr.X: 47650535-47650535 on Build GRCh38
rs768604125 Chr.X: 47650552-47650553 on Build GRCh38
rs776050931 Chr.X: 47650539-47650540 on Build GRCh38
rs757623390 Chr.X: 47650545-47650545 on Build GRCh38
rs778858618 Chr.X: 47650428-47650428 on Build GRCh38
rs775660827 Chr.X: 47650576-47650576 on Build GRCh38
rs758329305 Chr.X: 47650440-47650440 on Build GRCh38
rs768064079 Chr.X: 47650376-47650376 on Build GRCh38
rs1029014918 Chr.X: 47650678-47650678 on Build GRCh38
rs776097447 Chr.X: 47650538-47650538 on Build GRCh38
rs997476230 Chr.X: 47650647-47650647 on Build GRCh38
rs752777642 Chr.X: 47650384-47650384 on Build GRCh38
rs757498460 Chr.X: 47650417-47650417 on Build GRCh38
rs972626409 Chr.X: 47650740-47650740 on Build GRCh38
rs756182672 Chr.X: 47650465-47650471 on Build GRCh38
rs989611854 Chr.X: 47650716-47650716 on Build GRCh38
rs764284823 Chr.X: 47650543-47650543 on Build GRCh38
rs753906216 Chr.X: 47650544-47650544 on Build GRCh38
rs758176584 Chr.X: 47650551-47650551 on Build GRCh38
rs746986771 Chr.X: 47650555-47650555 on Build GRCh38
rs761039114 Chr.X: 47650539-47650539 on Build GRCh38
rs746537753 Chr.X: 47650459-47650459 on Build GRCh38
rs1048575626 Chr.X: 47650548-47650548 on Build GRCh38
rs1030635625 Chr.X: 47650627-47650627 on Build GRCh38
rs770414164 Chr.X: 47650720-47650720 on Build GRCh38
rs769367920 Chr.X: 47650533-47650533 on Build GRCh38
rs535762049 Chr.X: 47650766-47650766 on Build GRCh38
rs1045381801 Chr.X: 47650560-47650560 on Build GRCh38
rs973042769 Chr.X: 47650552-47650552 on Build GRCh38
rs769189512 Chr.X: 47650762-47650762 on Build GRCh38
rs184842332 Chr.X: 47650761-47650761 on Build GRCh38
rs976123442 Chr.X: 47650522-47650522 on Build GRCh38
rs756187724 Chr.X: 47650387-47650387 on Build GRCh38
rs746332406 Chr.X: 47650672-47650672 on Build GRCh38
rs754832524 Chr.X: 47650530-47650531 on Build GRCh38
rs780015299 Chr.X: 47650450-47650450 on Build GRCh38
rs745658220 Chr.X: 47650434-47650434 on Build GRCh38
rs386825120 Chr.X: 47650761-47650762 on Build GRCh38
rs747220403 Chr.X: 47650549-47650549 on Build GRCh38
rs772829588 Chr.X: 47650570-47650570 on Build GRCh38
rs375321526 Chr.X: 47650577-47650577 on Build GRCh38
rs757321217 Chr.X: 47650574-47650574 on Build GRCh38
rs747810183 Chr.X: 47650532-47650532 on Build GRCh38
rs1012531327 Chr.X: 47650572-47650572 on Build GRCh38
rs765546587 Chr.X: 47650547-47650547 on Build GRCh38
rs1003199784 Chr.X: 47650601-47650601 on Build GRCh38
rs780683384 Chr.X: 47650557-47650557 on Build GRCh38
rs755177203 Chr.X: 47650521-47650522 on Build GRCh38
rs1010429578 Chr.X: 47650589-47650589 on Build GRCh38
rs776911788 Chr.X: 47650607-47650607 on Build GRCh38
rs779670525 Chr.X: 47650554-47650554 on Build GRCh38
rs768241827 Chr.X: 47650484-47650484 on Build GRCh38
rs766410990 Chr.X: 47650465-47650466 on Build GRCh38
rs1020565824 Chr.X: 47650509-47650509 on Build GRCh38
rs775300271 Chr.X: 47650545-47650546 on Build GRCh38
rs953412167 Chr.X: 47650684-47650684 on Build GRCh38
rs759930819 Chr.X: 47650536-47650536 on Build GRCh38
rs75428830 Chr.X: 47650770-47650770 on Build GRCh38
rs754069927 Chr.X: 47650497-47650498 on Build GRCh38
rs1006296112 Chr.X: 47650689-47650689 on Build GRCh38
rs962859240 Chr.X: 47650721-47650721 on Build GRCh38
rs778408706 Chr.X: 47650533-47650534 on Build GRCh38
rs777951042 Chr.X: 47650537-47650537 on Build GRCh38
rs777675277 Chr.X: 47650388-47650388 on Build GRCh38
rs754136631 Chr.X: 47650410-47650410 on Build GRCh38
rs190242144 Chr.X: 47650786-47650786 on Build GRCh38

View Full Product Details