Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101550872-101551404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX1
Entrez Gene ID
51309
Gene Name
armadillo repeat containing, X-linked 1
Gene Aliases
ALEX1, GASP7
UniGene
Hs.9728
Gene Chromosome Location
Chr.X: 101550531-101554700 on Build GRCh38
Transcripts
NM_016608.1

SNP Detail

SNP ID Chromosome Location
rs1044616231 Chr.X: 101551265-101551265 on Build GRCh38
rs913723954 Chr.X: 101551027-101551027 on Build GRCh38
rs782063928 Chr.X: 101551241-101551241 on Build GRCh38
rs979372594 Chr.X: 101551058-101551058 on Build GRCh38
rs782487809 Chr.X: 101551360-101551360 on Build GRCh38
rs181833713 Chr.X: 101551282-101551282 on Build GRCh38
rs1034665060 Chr.X: 101551333-101551333 on Build GRCh38
rs987911280 Chr.X: 101550984-101550984 on Build GRCh38
rs185076424 Chr.X: 101550942-101550942 on Build GRCh38
rs782524835 Chr.X: 101551192-101551192 on Build GRCh38
rs782001328 Chr.X: 101550958-101550958 on Build GRCh38
rs960390948 Chr.X: 101551348-101551348 on Build GRCh38
rs887947577 Chr.X: 101551231-101551231 on Build GRCh38
rs887970893 Chr.X: 101550915-101550915 on Build GRCh38
rs782622904 Chr.X: 101551031-101551031 on Build GRCh38
rs374987676 Chr.X: 101551322-101551322 on Build GRCh38
rs1048311043 Chr.X: 101551229-101551229 on Build GRCh38
rs781826985 Chr.X: 101551166-101551166 on Build GRCh38
rs1034736866 Chr.X: 101550976-101550976 on Build GRCh38
rs370523966 Chr.X: 101551098-101551098 on Build GRCh38
rs1040367756 Chr.X: 101551146-101551146 on Build GRCh38
rs1001846295 Chr.X: 101551290-101551290 on Build GRCh38
rs960461538 Chr.X: 101550983-101550983 on Build GRCh38
rs782810245 Chr.X: 101551101-101551101 on Build GRCh38
rs12849615 Chr.X: 101551330-101551330 on Build GRCh38
rs782438475 Chr.X: 101550953-101550953 on Build GRCh38
rs189492220 Chr.X: 101551017-101551017 on Build GRCh38
rs910462438 Chr.X: 101551093-101551093 on Build GRCh38
rs1021162706 Chr.X: 101551382-101551382 on Build GRCh38
rs980439196 Chr.X: 101550974-101550974 on Build GRCh38
rs367999415 Chr.X: 101551361-101551361 on Build GRCh38
rs1006801754 Chr.X: 101551244-101551244 on Build GRCh38
rs781830589 Chr.X: 101551316-101551316 on Build GRCh38
rs781882079 Chr.X: 101551224-101551224 on Build GRCh38

View Full Product Details