Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103310904-103311394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEX2
Entrez Gene ID
84707
Gene Name
brain expressed X-linked 2
Gene Aliases
BEX1, DJ79P11.1
UniGene
Hs.398989
Gene Chromosome Location
Chr.X: 103309346-103311046 on Build GRCh38
Transcripts
NM_001168399.1 NM_001168400.1 NM_001168401.1 NM_032621.3

SNP Detail

SNP ID Chromosome Location
rs1003392636 Chr.X: 103310956-103310956 on Build GRCh38
rs919456224 Chr.X: 103311045-103311045 on Build GRCh38
rs975205195 Chr.X: 103311186-103311186 on Build GRCh38
rs956654226 Chr.X: 103310962-103310962 on Build GRCh38
rs186240357 Chr.X: 103311280-103311280 on Build GRCh38
rs765145220 Chr.X: 103311217-103311217 on Build GRCh38
rs998901612 Chr.X: 103311311-103311311 on Build GRCh38
rs963921618 Chr.X: 103311147-103311147 on Build GRCh38
rs1056927402 Chr.X: 103310963-103310963 on Build GRCh38
rs911655130 Chr.X: 103311023-103311023 on Build GRCh38
rs761900745 Chr.X: 103310974-103310974 on Build GRCh38
rs11545116 Chr.X: 103310922-103310922 on Build GRCh38
rs2273693 Chr.X: 103311087-103311087 on Build GRCh38
rs977324665 Chr.X: 103311302-103311302 on Build GRCh38
rs1022198705 Chr.X: 103310977-103310977 on Build GRCh38
rs1045735071 Chr.X: 103311223-103311223 on Build GRCh38
rs914405886 Chr.X: 103311220-103311220 on Build GRCh38
rs965732456 Chr.X: 103311035-103311035 on Build GRCh38
rs972206549 Chr.X: 103311037-103311037 on Build GRCh38
rs1012795543 Chr.X: 103310993-103310993 on Build GRCh38
rs752653370 Chr.X: 103311357-103311357 on Build GRCh38
rs985735092 Chr.X: 103311000-103311000 on Build GRCh38
rs930817422 Chr.X: 103311062-103311062 on Build GRCh38
rs760493547 Chr.X: 103311360-103311360 on Build GRCh38
rs907295591 Chr.X: 103311252-103311252 on Build GRCh38
rs951101847 Chr.X: 103311038-103311038 on Build GRCh38
rs2756056 Chr.X: 103310966-103310966 on Build GRCh38
rs759364575 Chr.X: 103311157-103311157 on Build GRCh38
rs768748935 Chr.X: 103310955-103310955 on Build GRCh38
rs916148753 Chr.X: 103311121-103311121 on Build GRCh38
rs773424059 Chr.X: 103310988-103310988 on Build GRCh38
rs1030807120 Chr.X: 103310959-103310959 on Build GRCh38
rs12559214 Chr.X: 103310978-103310978 on Build GRCh38
rs753654497 Chr.X: 103311136-103311136 on Build GRCh38
rs948749596 Chr.X: 103311206-103311206 on Build GRCh38
rs752622011 Chr.X: 103311232-103311232 on Build GRCh38
rs902509929 Chr.X: 103310946-103310946 on Build GRCh38

View Full Product Details