Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 108571526-108571958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A5
Entrez Gene ID
1287
Gene Name
collagen type IV alpha 5 chain
Gene Aliases
ASLN, ATS, CA54
UniGene
Hs.369089
Gene Chromosome Location
Chr.X: 108439844-108697545 on Build GRCh38
Transcripts
NM_000495.4 NM_033380.2 XM_011530849.2 XM_017029259.1 XM_017029260.1 XM_017029261.1 XM_017029262.1

SNP Detail

SNP ID Chromosome Location
rs368541565 Chr.X: 108571707-108571707 on Build GRCh38
rs774802858 Chr.X: 108571781-108571781 on Build GRCh38
rs953325742 Chr.X: 108571657-108571657 on Build GRCh38
rs773404950 Chr.X: 108571815-108571815 on Build GRCh38
rs368401149 Chr.X: 108571564-108571564 on Build GRCh38
rs779290459 Chr.X: 108571632-108571632 on Build GRCh38
rs780861892 Chr.X: 108571760-108571760 on Build GRCh38
rs980421633 Chr.X: 108571822-108571822 on Build GRCh38
rs939625708 Chr.X: 108571893-108571893 on Build GRCh38
rs996864226 Chr.X: 108571607-108571607 on Build GRCh38
rs1005813217 Chr.X: 108571714-108571714 on Build GRCh38
rs754205579 Chr.X: 108571905-108571905 on Build GRCh38
rs903247321 Chr.X: 108571593-108571593 on Build GRCh38
rs749713471 Chr.X: 108571778-108571778 on Build GRCh38
rs184637253 Chr.X: 108571849-108571849 on Build GRCh38
rs200719406 Chr.X: 108571771-108571771 on Build GRCh38
rs987761852 Chr.X: 108571669-108571669 on Build GRCh38
rs754027462 Chr.X: 108571850-108571850 on Build GRCh38
rs940206855 Chr.X: 108571924-108571924 on Build GRCh38
rs760774618 Chr.X: 108571827-108571827 on Build GRCh38
rs1013501954 Chr.X: 108571682-108571682 on Build GRCh38
rs773196036 Chr.X: 108571616-108571616 on Build GRCh38
rs986378173 Chr.X: 108571877-108571877 on Build GRCh38
rs745493972 Chr.X: 108571763-108571763 on Build GRCh38
rs104886054 Chr.X: 108571818-108571818 on Build GRCh38
rs1028305100 Chr.X: 108571611-108571611 on Build GRCh38
rs999770270 Chr.X: 108571596-108571596 on Build GRCh38
rs767531424 Chr.X: 108571794-108571794 on Build GRCh38
rs1043178046 Chr.X: 108571578-108571578 on Build GRCh38
rs104886053 Chr.X: 108571812-108571812 on Build GRCh38
rs748489776 Chr.X: 108571706-108571706 on Build GRCh38
rs767432766 Chr.X: 108571711-108571712 on Build GRCh38
rs367679870 Chr.X: 108571790-108571790 on Build GRCh38
rs181939097 Chr.X: 108571774-108571774 on Build GRCh38
rs955167336 Chr.X: 108571848-108571848 on Build GRCh38
rs926241127 Chr.X: 108571846-108571846 on Build GRCh38
rs606231369 Chr.X: 108571814-108571815 on Build GRCh38
rs104886419 Chr.X: 108571813-108571814 on Build GRCh38
rs769657277 Chr.X: 108571606-108571606 on Build GRCh38
rs371752800 Chr.X: 108571901-108571901 on Build GRCh38

View Full Product Details