Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 116170784-116171328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTR2
Entrez Gene ID
186
Gene Name
angiotensin II receptor type 2
Gene Aliases
AT2, ATGR2, MRX88
UniGene
Hs.405348
Gene Chromosome Location
Chr.X: 116170705-116174972 on Build GRCh38
Transcripts
NM_000686.4 XM_011537533.1

SNP Detail

SNP ID Chromosome Location
rs192844243 Chr.X: 116171286-116171286 on Build GRCh38
rs782073337 Chr.X: 116171247-116171247 on Build GRCh38
rs987397265 Chr.X: 116171245-116171245 on Build GRCh38
rs1017365861 Chr.X: 116171194-116171194 on Build GRCh38
rs781936738 Chr.X: 116171146-116171146 on Build GRCh38
rs199887971 Chr.X: 116171246-116171247 on Build GRCh38
rs145201241 Chr.X: 116171297-116171297 on Build GRCh38
rs146688001 Chr.X: 116171092-116171092 on Build GRCh38
rs904242327 Chr.X: 116170953-116170953 on Build GRCh38
rs1055513995 Chr.X: 116171074-116171074 on Build GRCh38
rs17231401 Chr.X: 116171216-116171216 on Build GRCh38
rs781921584 Chr.X: 116171199-116171199 on Build GRCh38
rs914166563 Chr.X: 116170891-116170891 on Build GRCh38
rs782802553 Chr.X: 116171270-116171271 on Build GRCh38
rs185185966 Chr.X: 116171301-116171301 on Build GRCh38
rs782716576 Chr.X: 116171236-116171236 on Build GRCh38
rs1028443988 Chr.X: 116171228-116171228 on Build GRCh38
rs935760504 Chr.X: 116171027-116171027 on Build GRCh38
rs1024214429 Chr.X: 116171145-116171145 on Build GRCh38
rs987060634 Chr.X: 116170872-116170872 on Build GRCh38
rs782163007 Chr.X: 116171235-116171236 on Build GRCh38
rs782681915 Chr.X: 116170838-116170838 on Build GRCh38
rs955510353 Chr.X: 116171236-116171236 on Build GRCh38
rs782661234 Chr.X: 116171302-116171302 on Build GRCh38
rs182182972 Chr.X: 116170839-116170839 on Build GRCh38
rs934184624 Chr.X: 116170869-116170869 on Build GRCh38
rs782387285 Chr.X: 116170876-116170876 on Build GRCh38
rs1403543 Chr.X: 116170939-116170939 on Build GRCh38
rs3736556 Chr.X: 116171082-116171082 on Build GRCh38
rs140243726 Chr.X: 116171173-116171173 on Build GRCh38
rs945712765 Chr.X: 116170923-116170923 on Build GRCh38

View Full Product Details