Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120371024-120371526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B4
Entrez Gene ID
23439
Gene Name
ATPase Na+/K+ transporting family member beta 4
Gene Aliases
-
UniGene
Hs.292599
Gene Chromosome Location
Chr.X: 120362085-120383249 on Build GRCh38
Transcripts
NM_001142447.2 NM_012069.4 XM_017029381.1

SNP Detail

SNP ID Chromosome Location
rs754852244 Chr.X: 120371073-120371073 on Build GRCh38
rs2231243 Chr.X: 120371269-120371269 on Build GRCh38
rs763182565 Chr.X: 120371153-120371153 on Build GRCh38
rs188398114 Chr.X: 120371125-120371125 on Build GRCh38
rs777639155 Chr.X: 120371213-120371213 on Build GRCh38
rs1031575416 Chr.X: 120371140-120371140 on Build GRCh38
rs778399800 Chr.X: 120371087-120371087 on Build GRCh38
rs750309234 Chr.X: 120371056-120371056 on Build GRCh38
rs945552561 Chr.X: 120371439-120371439 on Build GRCh38
rs760129981 Chr.X: 120371110-120371110 on Build GRCh38
rs750859516 Chr.X: 120371223-120371223 on Build GRCh38
rs879141427 Chr.X: 120371080-120371080 on Build GRCh38
rs770247709 Chr.X: 120371211-120371211 on Build GRCh38
rs78360050 Chr.X: 120371131-120371131 on Build GRCh38
rs770001912 Chr.X: 120371235-120371235 on Build GRCh38
rs997503393 Chr.X: 120371431-120371431 on Build GRCh38
rs898097824 Chr.X: 120371334-120371334 on Build GRCh38
rs768662642 Chr.X: 120371160-120371160 on Build GRCh38
rs747866992 Chr.X: 120371098-120371098 on Build GRCh38
rs757934363 Chr.X: 120371102-120371102 on Build GRCh38
rs753571109 Chr.X: 120371061-120371061 on Build GRCh38
rs183079208 Chr.X: 120371178-120371178 on Build GRCh38
rs751621998 Chr.X: 120371272-120371272 on Build GRCh38
rs917688858 Chr.X: 120371299-120371299 on Build GRCh38
rs1054078303 Chr.X: 120371363-120371363 on Build GRCh38
rs1049731990 Chr.X: 120371355-120371355 on Build GRCh38
rs781753966 Chr.X: 120371126-120371126 on Build GRCh38
rs948478580 Chr.X: 120371222-120371222 on Build GRCh38
rs955969946 Chr.X: 120371152-120371152 on Build GRCh38
rs376920861 Chr.X: 120371164-120371164 on Build GRCh38
rs756057773 Chr.X: 120371058-120371058 on Build GRCh38
rs980898543 Chr.X: 120371315-120371315 on Build GRCh38
rs765104057 Chr.X: 120371190-120371190 on Build GRCh38
rs929565243 Chr.X: 120371345-120371345 on Build GRCh38
rs753854230 Chr.X: 120371171-120371171 on Build GRCh38
rs773378689 Chr.X: 120371457-120371458 on Build GRCh38
rs747436925 Chr.X: 120371211-120371214 on Build GRCh38
rs1031664781 Chr.X: 120371452-120371452 on Build GRCh38
rs1017621882 Chr.X: 120371386-120371386 on Build GRCh38
rs936334586 Chr.X: 120371345-120371346 on Build GRCh38
rs775531978 Chr.X: 120371143-120371143 on Build GRCh38
rs1007612971 Chr.X: 120371369-120371369 on Build GRCh38
rs140495898 Chr.X: 120371192-120371192 on Build GRCh38
rs1036680863 Chr.X: 120371329-120371329 on Build GRCh38
rs201685187 Chr.X: 120371230-120371230 on Build GRCh38
rs780478361 Chr.X: 120371242-120371242 on Build GRCh38
rs956191236 Chr.X: 120371459-120371459 on Build GRCh38
rs755021768 Chr.X: 120371312-120371312 on Build GRCh38
rs990423839 Chr.X: 120371165-120371165 on Build GRCh38
rs187760302 Chr.X: 120371392-120371392 on Build GRCh38
rs770231709 Chr.X: 120371134-120371134 on Build GRCh38
rs774417121 Chr.X: 120371161-120371161 on Build GRCh38
rs758130473 Chr.X: 120371206-120371206 on Build GRCh38
rs781515517 Chr.X: 120371225-120371225 on Build GRCh38
rs887911523 Chr.X: 120371358-120371358 on Build GRCh38
rs995439585 Chr.X: 120371167-120371167 on Build GRCh38
rs1051952938 Chr.X: 120371439-120371439 on Build GRCh38

View Full Product Details