Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120379788-120380284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B4
Entrez Gene ID
23439
Gene Name
ATPase Na+/K+ transporting family member beta 4
Gene Aliases
-
UniGene
Hs.292599
Gene Chromosome Location
Chr.X: 120362085-120383249 on Build GRCh38
Transcripts
NM_001142447.2 NM_012069.4

SNP Detail

SNP ID Chromosome Location
rs958885186 Chr.X: 120380169-120380169 on Build GRCh38
rs753307170 Chr.X: 120380040-120380040 on Build GRCh38
rs951217526 Chr.X: 120380257-120380257 on Build GRCh38
rs112348175 Chr.X: 120380160-120380160 on Build GRCh38
rs926817664 Chr.X: 120379925-120379925 on Build GRCh38
rs981007727 Chr.X: 120379914-120379914 on Build GRCh38
rs992734361 Chr.X: 120380226-120380226 on Build GRCh38
rs778538331 Chr.X: 120380116-120380116 on Build GRCh38
rs941056583 Chr.X: 120380012-120380012 on Build GRCh38
rs949674106 Chr.X: 120379913-120379913 on Build GRCh38
rs1026679995 Chr.X: 120379820-120379820 on Build GRCh38
rs993078122 Chr.X: 120379926-120379926 on Build GRCh38
rs183451889 Chr.X: 120380121-120380121 on Build GRCh38
rs1039367449 Chr.X: 120380024-120380024 on Build GRCh38
rs1046820740 Chr.X: 120379964-120379964 on Build GRCh38
rs187428800 Chr.X: 120380149-120380149 on Build GRCh38
rs1018141452 Chr.X: 120380087-120380087 on Build GRCh38
rs1008602737 Chr.X: 120380086-120380086 on Build GRCh38
rs564716967 Chr.X: 120380228-120380228 on Build GRCh38
rs890606895 Chr.X: 120380113-120380113 on Build GRCh38
rs139049149 Chr.X: 120379844-120379844 on Build GRCh38
rs767507364 Chr.X: 120379832-120379832 on Build GRCh38
rs763633512 Chr.X: 120380007-120380007 on Build GRCh38
rs150857428 Chr.X: 120380089-120380089 on Build GRCh38
rs879074031 Chr.X: 120380231-120380231 on Build GRCh38
rs748778506 Chr.X: 120380063-120380063 on Build GRCh38
rs775208928 Chr.X: 120379839-120379839 on Build GRCh38
rs900752046 Chr.X: 120380061-120380061 on Build GRCh38
rs937272164 Chr.X: 120379903-120379903 on Build GRCh38
rs945453486 Chr.X: 120380046-120380046 on Build GRCh38
rs929745058 Chr.X: 120379949-120379949 on Build GRCh38
rs968782139 Chr.X: 120380147-120380147 on Build GRCh38
rs756077276 Chr.X: 120379895-120379895 on Build GRCh38
rs996411456 Chr.X: 120380066-120380066 on Build GRCh38
rs978772773 Chr.X: 120379842-120379842 on Build GRCh38
rs927237731 Chr.X: 120379845-120379845 on Build GRCh38
rs917051878 Chr.X: 120379945-120379945 on Build GRCh38
rs979210622 Chr.X: 120380154-120380154 on Build GRCh38

View Full Product Details