Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135032222-135032716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM127A
Entrez Gene ID
8933
Gene Name
family with sequence similarity 127 member A
Gene Aliases
CXX1, MAR8C, MART8C, Mar8, Mart8
UniGene
Hs.522789
Gene Chromosome Location
Chr.X: 135032303-135033546 on Build GRCh38
Transcripts
NM_001078171.1

SNP Detail

SNP ID Chromosome Location
rs747680888 Chr.X: 135032381-135032381 on Build GRCh38
rs753658574 Chr.X: 135032372-135032372 on Build GRCh38
rs750314248 Chr.X: 135032356-135032356 on Build GRCh38
rs999729372 Chr.X: 135032311-135032311 on Build GRCh38
rs368860960 Chr.X: 135032503-135032503 on Build GRCh38
rs777984742 Chr.X: 135032512-135032512 on Build GRCh38
rs753746466 Chr.X: 135032515-135032515 on Build GRCh38
rs771062139 Chr.X: 135032665-135032665 on Build GRCh38
rs372888607 Chr.X: 135032653-135032653 on Build GRCh38
rs1046513215 Chr.X: 135032303-135032303 on Build GRCh38
rs752338670 Chr.X: 135032548-135032548 on Build GRCh38
rs775602482 Chr.X: 135032341-135032341 on Build GRCh38
rs1010564631 Chr.X: 135032579-135032579 on Build GRCh38
rs771395950 Chr.X: 135032390-135032390 on Build GRCh38
rs757819192 Chr.X: 135032581-135032581 on Build GRCh38
rs752324016 Chr.X: 135032694-135032694 on Build GRCh38
rs766233596 Chr.X: 135032363-135032363 on Build GRCh38
rs758023574 Chr.X: 135032257-135032257 on Build GRCh38
rs770083665 Chr.X: 135032637-135032637 on Build GRCh38
rs751010226 Chr.X: 135032567-135032567 on Build GRCh38
rs763082451 Chr.X: 135032344-135032344 on Build GRCh38
rs947908417 Chr.X: 135032382-135032382 on Build GRCh38
rs112073308 Chr.X: 135032602-135032602 on Build GRCh38
rs754181993 Chr.X: 135032261-135032261 on Build GRCh38
rs770067362 Chr.X: 135032412-135032412 on Build GRCh38
rs762954047 Chr.X: 135032485-135032485 on Build GRCh38
rs370016193 Chr.X: 135032378-135032378 on Build GRCh38
rs1012829950 Chr.X: 135032296-135032296 on Build GRCh38
rs749481905 Chr.X: 135032647-135032647 on Build GRCh38
rs201161184 Chr.X: 135032488-135032488 on Build GRCh38
rs755880239 Chr.X: 135032361-135032361 on Build GRCh38
rs754729222 Chr.X: 135032373-135032373 on Build GRCh38
rs778966104 Chr.X: 135032326-135032326 on Build GRCh38
rs755255807 Chr.X: 135032289-135032289 on Build GRCh38
rs764987804 Chr.X: 135032530-135032530 on Build GRCh38
rs1031321667 Chr.X: 135032315-135032315 on Build GRCh38
rs746514642 Chr.X: 135032349-135032349 on Build GRCh38
rs759419702 Chr.X: 135032680-135032680 on Build GRCh38
rs56094073 Chr.X: 135032457-135032457 on Build GRCh38
rs202167524 Chr.X: 135032500-135032500 on Build GRCh38
rs761975571 Chr.X: 135032657-135032657 on Build GRCh38
rs757859781 Chr.X: 135032384-135032384 on Build GRCh38
rs1056983 Chr.X: 135032692-135032692 on Build GRCh38
rs779578586 Chr.X: 135032308-135032308 on Build GRCh38
rs778156525 Chr.X: 135032389-135032389 on Build GRCh38
rs773050730 Chr.X: 135032489-135032489 on Build GRCh38
rs780160006 Chr.X: 135032644-135032644 on Build GRCh38
rs746216507 Chr.X: 135032407-135032407 on Build GRCh38
rs373869933 Chr.X: 135032425-135032425 on Build GRCh38
rs776645728 Chr.X: 135032674-135032674 on Build GRCh38
rs895205765 Chr.X: 135032277-135032277 on Build GRCh38
rs746234823 Chr.X: 135032607-135032607 on Build GRCh38
rs372924234 Chr.X: 135032355-135032355 on Build GRCh38
rs1056977 Chr.X: 135032562-135032562 on Build GRCh38
rs1046615628 Chr.X: 135032476-135032476 on Build GRCh38
rs757869122 Chr.X: 135032552-135032552 on Build GRCh38

View Full Product Details