Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135546995-135547525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS6L
Entrez Gene ID
203522
Gene Name
integrator complex subunit 6 like
Gene Aliases
DDX26B
UniGene
Hs.496829
Gene Chromosome Location
Chr.X: 135520630-135582535 on Build GRCh38
Transcripts
NM_182540.4 XM_005262394.2 XM_006724740.2 XM_006724741.3 XM_011531311.2 XM_017029342.1 XM_017029343.1 XM_017029344.1 XM_017029345.1 XM_017029346.1 XM_017029347.1 XM_017029348.1

SNP Detail

SNP ID Chromosome Location
rs782099771 Chr.X: 135547144-135547144 on Build GRCh38
rs782567799 Chr.X: 135547292-135547292 on Build GRCh38
rs370756456 Chr.X: 135547237-135547237 on Build GRCh38
rs782332639 Chr.X: 135547274-135547274 on Build GRCh38
rs782252196 Chr.X: 135547308-135547308 on Build GRCh38
rs781853258 Chr.X: 135547141-135547141 on Build GRCh38
rs147342824 Chr.X: 135547199-135547199 on Build GRCh38
rs782317646 Chr.X: 135547095-135547095 on Build GRCh38
rs782082653 Chr.X: 135547198-135547198 on Build GRCh38
rs782819723 Chr.X: 135547330-135547330 on Build GRCh38
rs782603921 Chr.X: 135547427-135547428 on Build GRCh38
rs782217865 Chr.X: 135547262-135547262 on Build GRCh38
rs960814063 Chr.X: 135547482-135547482 on Build GRCh38
rs181035631 Chr.X: 135547356-135547356 on Build GRCh38
rs200150477 Chr.X: 135547097-135547097 on Build GRCh38
rs782633480 Chr.X: 135547248-135547248 on Build GRCh38
rs781886514 Chr.X: 135547228-135547228 on Build GRCh38
rs782795978 Chr.X: 135547114-135547114 on Build GRCh38
rs1050117417 Chr.X: 135547103-135547103 on Build GRCh38
rs888894171 Chr.X: 135547131-135547131 on Build GRCh38
rs576333151 Chr.X: 135547427-135547427 on Build GRCh38
rs1027585078 Chr.X: 135547381-135547381 on Build GRCh38
rs782412212 Chr.X: 135547301-135547301 on Build GRCh38
rs782721006 Chr.X: 135547190-135547190 on Build GRCh38
rs373900942 Chr.X: 135547300-135547300 on Build GRCh38
rs1033748668 Chr.X: 135547464-135547464 on Build GRCh38
rs189243646 Chr.X: 135547314-135547314 on Build GRCh38
rs782465815 Chr.X: 135547364-135547364 on Build GRCh38
rs782152837 Chr.X: 135547099-135547099 on Build GRCh38
rs782744423 Chr.X: 135547223-135547223 on Build GRCh38
rs782007254 Chr.X: 135547305-135547305 on Build GRCh38
rs993787273 Chr.X: 135547358-135547358 on Build GRCh38
rs887584551 Chr.X: 135547421-135547421 on Build GRCh38
rs781851459 Chr.X: 135547245-135547245 on Build GRCh38
rs782074459 Chr.X: 135547090-135547090 on Build GRCh38
rs781849837 Chr.X: 135547428-135547428 on Build GRCh38

View Full Product Details