Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136654281-136654777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD40LG
Entrez Gene ID
959
Gene Name
CD40 ligand
Gene Aliases
CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L
UniGene
Hs.592244
Gene Chromosome Location
Chr.X: 136648177-136660390 on Build GRCh38
Transcripts
NM_000074.2

SNP Detail

SNP ID Chromosome Location
rs764859012 Chr.X: 136654356-136654356 on Build GRCh38
rs760105946 Chr.X: 136654558-136654558 on Build GRCh38
rs981484035 Chr.X: 136654598-136654598 on Build GRCh38
rs768002105 Chr.X: 136654564-136654564 on Build GRCh38
rs757648814 Chr.X: 136654446-136654446 on Build GRCh38
rs914146816 Chr.X: 136654612-136654612 on Build GRCh38
rs751481337 Chr.X: 136654336-136654337 on Build GRCh38
rs924392631 Chr.X: 136654610-136654610 on Build GRCh38
rs1047827446 Chr.X: 136654315-136654315 on Build GRCh38
rs376582437 Chr.X: 136654398-136654398 on Build GRCh38
rs1022660766 Chr.X: 136654547-136654547 on Build GRCh38
rs1016790781 Chr.X: 136654447-136654447 on Build GRCh38
rs373042001 Chr.X: 136654482-136654482 on Build GRCh38
rs368691563 Chr.X: 136654414-136654414 on Build GRCh38
rs778398894 Chr.X: 136654399-136654399 on Build GRCh38
rs752210787 Chr.X: 136654394-136654394 on Build GRCh38
rs914734790 Chr.X: 136654742-136654742 on Build GRCh38
rs746127988 Chr.X: 136654475-136654475 on Build GRCh38
rs955943973 Chr.X: 136654672-136654672 on Build GRCh38
rs990319321 Chr.X: 136654678-136654678 on Build GRCh38
rs759052126 Chr.X: 136654352-136654352 on Build GRCh38
rs779077894 Chr.X: 136654743-136654743 on Build GRCh38
rs374355208 Chr.X: 136654366-136654366 on Build GRCh38
rs1004051141 Chr.X: 136654374-136654374 on Build GRCh38
rs1016773951 Chr.X: 136654463-136654463 on Build GRCh38
rs1012824438 Chr.X: 136654524-136654524 on Build GRCh38
rs969864941 Chr.X: 136654489-136654489 on Build GRCh38
rs781357909 Chr.X: 136654466-136654466 on Build GRCh38
rs988108115 Chr.X: 136654609-136654609 on Build GRCh38
rs886495826 Chr.X: 136654321-136654321 on Build GRCh38

View Full Product Details