Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139731543-139732041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11C
Entrez Gene ID
286410
Gene Name
ATPase phospholipid transporting 11C
Gene Aliases
ATPIG, ATPIQ
UniGene
Hs.88252
Gene Chromosome Location
Chr.X: 139726346-139933083 on Build GRCh38
Transcripts
NM_001010986.2 NM_173694.4 XM_017029439.1 XM_017029440.1 XM_017029441.1 XM_017029442.1 XM_017029443.1 XM_017029444.1 XM_017029445.1 XM_017029446.1 XM_017029447.1 XM_017029448.1 XM_017029449.1

SNP Detail

SNP ID Chromosome Location
rs1033398557 Chr.X: 139731665-139731665 on Build GRCh38
rs201171038 Chr.X: 139731716-139731716 on Build GRCh38
rs141323884 Chr.X: 139731691-139731691 on Build GRCh38
rs959304549 Chr.X: 139731671-139731671 on Build GRCh38
rs1054483758 Chr.X: 139731956-139731956 on Build GRCh38
rs781247565 Chr.X: 139731672-139731672 on Build GRCh38
rs777247327 Chr.X: 139731793-139731793 on Build GRCh38
rs756049514 Chr.X: 139731786-139731786 on Build GRCh38
rs376989480 Chr.X: 139731762-139731762 on Build GRCh38
rs200548879 Chr.X: 139731804-139731804 on Build GRCh38
rs754914745 Chr.X: 139731869-139731869 on Build GRCh38
rs752422594 Chr.X: 139731629-139731629 on Build GRCh38
rs768818596 Chr.X: 139731707-139731707 on Build GRCh38
rs925144362 Chr.X: 139731868-139731868 on Build GRCh38
rs191719186 Chr.X: 139731992-139731992 on Build GRCh38
rs749305244 Chr.X: 139731687-139731687 on Build GRCh38
rs750604730 Chr.X: 139731595-139731595 on Build GRCh38
rs1006523409 Chr.X: 139731994-139731994 on Build GRCh38
rs771332960 Chr.X: 139731780-139731780 on Build GRCh38
rs777556358 Chr.X: 139731866-139731866 on Build GRCh38
rs758494464 Chr.X: 139731611-139731611 on Build GRCh38
rs937327259 Chr.X: 139731686-139731686 on Build GRCh38
rs371274251 Chr.X: 139731846-139731847 on Build GRCh38
rs747448155 Chr.X: 139731772-139731772 on Build GRCh38
rs377283654 Chr.X: 139731957-139731957 on Build GRCh38
rs777554132 Chr.X: 139731632-139731632 on Build GRCh38
rs758940013 Chr.X: 139731796-139731796 on Build GRCh38
rs778964087 Chr.X: 139731710-139731710 on Build GRCh38
rs758137870 Chr.X: 139731630-139731630 on Build GRCh38
rs756140771 Chr.X: 139731810-139731810 on Build GRCh38
rs750314003 Chr.X: 139731737-139731737 on Build GRCh38
rs748404658 Chr.X: 139731718-139731718 on Build GRCh38
rs751428363 Chr.X: 139731642-139731642 on Build GRCh38
rs1017922929 Chr.X: 139732015-139732015 on Build GRCh38
rs977967477 Chr.X: 139731839-139731840 on Build GRCh38
rs375104383 Chr.X: 139731635-139731635 on Build GRCh38
rs945348749 Chr.X: 139731834-139731834 on Build GRCh38
rs752794960 Chr.X: 139731804-139731805 on Build GRCh38
rs765260497 Chr.X: 139731798-139731799 on Build GRCh38
rs773716821 Chr.X: 139731765-139731765 on Build GRCh38
rs778734112 Chr.X: 139732007-139732007 on Build GRCh38
rs751637397 Chr.X: 139731586-139731586 on Build GRCh38
rs113652727 Chr.X: 139731950-139731950 on Build GRCh38
rs962852673 Chr.X: 139731569-139731570 on Build GRCh38
rs902286028 Chr.X: 139731714-139731714 on Build GRCh38
rs903391293 Chr.X: 139731935-139731935 on Build GRCh38
rs929877851 Chr.X: 139731574-139731574 on Build GRCh38
rs936406837 Chr.X: 139731774-139731774 on Build GRCh38
rs909839653 Chr.X: 139731669-139731669 on Build GRCh38
rs749117607 Chr.X: 139731963-139731963 on Build GRCh38
rs774944787 Chr.X: 139731807-139731807 on Build GRCh38
rs1033276691 Chr.X: 139731987-139731987 on Build GRCh38
rs764777840 Chr.X: 139731628-139731628 on Build GRCh38

View Full Product Details