Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197056904-197057439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F13B
Entrez Gene ID
2165
Gene Name
coagulation factor XIII B chain
Gene Aliases
FXIIIB
UniGene
Hs.435782
Gene Chromosome Location
Chr.1: 197038741-197067267 on Build GRCh38
Transcripts
NM_001994.2 XM_011509283.2 XM_011509284.2 XM_011509286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5816891 c.1150A>T p.T384S Chr.1: 197057034-197057034 on Build GRCh38
4458936 c.1105C>A p.L369I Chr.1: 197057079-197057079 on Build GRCh38
6060754 c.1157C>G p.P386R Chr.1: 197057027-197057027 on Build GRCh38
5379036 c.1072G>A p.D358N Chr.1: 197057112-197057112 on Build GRCh38
3803341 c.1040C>A p.A347E Chr.1: 197057144-197057144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770212133 Chr.1: 197057241-197057241 on Build GRCh38
rs777514231 Chr.1: 197057083-197057083 on Build GRCh38
rs745474835 Chr.1: 197057387-197057387 on Build GRCh38
rs547036343 Chr.1: 197057036-197057036 on Build GRCh38
rs141627684 Chr.1: 197057347-197057347 on Build GRCh38
rs771039575 Chr.1: 197057170-197057170 on Build GRCh38
rs755966271 Chr.1: 197057039-197057039 on Build GRCh38
rs370218942 Chr.1: 197057085-197057085 on Build GRCh38
rs973248174 Chr.1: 197057026-197057026 on Build GRCh38
rs764894253 Chr.1: 197057327-197057327 on Build GRCh38
rs764387481 Chr.1: 197057031-197057031 on Build GRCh38
rs781729553 Chr.1: 197057214-197057214 on Build GRCh38
rs540644385 Chr.1: 197057010-197057010 on Build GRCh38
rs17514281 Chr.1: 197057159-197057159 on Build GRCh38
rs377192047 Chr.1: 197057275-197057275 on Build GRCh38
rs749840335 Chr.1: 197057395-197057395 on Build GRCh38
rs770416127 Chr.1: 197057103-197057103 on Build GRCh38
rs763316899 Chr.1: 197057269-197057269 on Build GRCh38
rs748779845 Chr.1: 197057065-197057065 on Build GRCh38
rs368848030 Chr.1: 197056975-197056975 on Build GRCh38
rs376295989 Chr.1: 197057124-197057124 on Build GRCh38
rs771416936 Chr.1: 197057075-197057075 on Build GRCh38
rs5991 Chr.1: 197057021-197057021 on Build GRCh38
rs753625753 Chr.1: 197057137-197057137 on Build GRCh38
rs565327540 Chr.1: 197057054-197057054 on Build GRCh38
rs372751236 Chr.1: 197057206-197057206 on Build GRCh38
rs376931227 Chr.1: 197057066-197057066 on Build GRCh38
rs531430163 Chr.1: 197056941-197056941 on Build GRCh38
rs746252205 Chr.1: 197057239-197057239 on Build GRCh38
rs1024971042 Chr.1: 197056958-197056958 on Build GRCh38
rs866833011 Chr.1: 197057027-197057028 on Build GRCh38
rs775799552 Chr.1: 197057113-197057113 on Build GRCh38
rs5999 Chr.1: 197057135-197057135 on Build GRCh38
rs866204769 Chr.1: 197057304-197057304 on Build GRCh38
rs778460584 Chr.1: 197057067-197057067 on Build GRCh38
rs747707592 Chr.1: 197057068-197057068 on Build GRCh38
rs1053903340 Chr.1: 197057194-197057194 on Build GRCh38
rs1056143305 Chr.1: 197056934-197056934 on Build GRCh38
rs573352795 Chr.1: 197057314-197057314 on Build GRCh38
rs1044976102 Chr.1: 197057230-197057230 on Build GRCh38
rs1026193608 Chr.1: 197057385-197057385 on Build GRCh38
rs745716465 Chr.1: 197057399-197057399 on Build GRCh38
rs774261325 Chr.1: 197057277-197057277 on Build GRCh38
rs777301898 Chr.1: 197056973-197056973 on Build GRCh38
rs771474972 Chr.1: 197057086-197057086 on Build GRCh38
rs939105097 Chr.1: 197057250-197057250 on Build GRCh38
rs892716537 Chr.1: 197057229-197057229 on Build GRCh38
rs117019969 Chr.1: 197057095-197057095 on Build GRCh38
rs373063254 Chr.1: 197057116-197057116 on Build GRCh38
rs140928543 Chr.1: 197057133-197057133 on Build GRCh38
rs773356629 Chr.1: 197057317-197057317 on Build GRCh38
rs766811391 Chr.1: 197056962-197056962 on Build GRCh38
rs752731148 Chr.1: 197057328-197057328 on Build GRCh38
rs778826479 Chr.1: 197057177-197057177 on Build GRCh38
rs759358030 Chr.1: 197057320-197057320 on Build GRCh38
rs150497300 Chr.1: 197057381-197057381 on Build GRCh38
rs756567129 Chr.1: 197057393-197057393 on Build GRCh38
rs370790381 Chr.1: 197057190-197057190 on Build GRCh38
rs752538346 Chr.1: 197057185-197057185 on Build GRCh38
rs1034181633 Chr.1: 197056989-197056989 on Build GRCh38
rs76405247 Chr.1: 197057202-197057202 on Build GRCh38
rs950755576 Chr.1: 197057168-197057168 on Build GRCh38
rs777127716 Chr.1: 197057191-197057191 on Build GRCh38
rs201977522 Chr.1: 197057372-197057372 on Build GRCh38
rs779773193 Chr.1: 197057044-197057044 on Build GRCh38
rs559428305 Chr.1: 197057046-197057046 on Build GRCh38
rs371807272 Chr.1: 197056979-197056979 on Build GRCh38
rs748757906 Chr.1: 197057028-197057029 on Build GRCh38
rs868256767 Chr.1: 197057045-197057045 on Build GRCh38
rs778962813 Chr.1: 197057188-197057190 on Build GRCh38
rs776091705 Chr.1: 197057249-197057249 on Build GRCh38
rs892131723 Chr.1: 197057299-197057299 on Build GRCh38
rs780257236 Chr.1: 197057394-197057394 on Build GRCh38
rs762088912 Chr.1: 197057289-197057289 on Build GRCh38
rs769037761 Chr.1: 197057399-197057399 on Build GRCh38
rs2990511 Chr.1: 197057309-197057309 on Build GRCh38
rs758239415 Chr.1: 197057345-197057345 on Build GRCh38
rs903780706 Chr.1: 197057329-197057329 on Build GRCh38
rs774547697 Chr.1: 197057413-197057413 on Build GRCh38
rs780992530 Chr.1: 197057384-197057384 on Build GRCh38
rs751826342 Chr.1: 197057129-197057129 on Build GRCh38
rs570300747 Chr.1: 197057097-197057097 on Build GRCh38
rs765451279 Chr.1: 197056986-197056986 on Build GRCh38
rs1038755060 Chr.1: 197057164-197057164 on Build GRCh38
rs147765919 Chr.1: 197057009-197057009 on Build GRCh38
rs769067366 Chr.1: 197057278-197057278 on Build GRCh38
rs1009257552 Chr.1: 197057312-197057312 on Build GRCh38
rs936840592 Chr.1: 197057192-197057192 on Build GRCh38

View Full Product Details