Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200554048-200554598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF14
Entrez Gene ID
9928
Gene Name
kinesin family member 14
Gene Aliases
MKS12
UniGene
Hs.3104
Gene Chromosome Location
Chr.1: 200551497-200620791 on Build GRCh38
Transcripts
NM_001305792.1 NM_014875.2 XM_011510231.2 XM_011510232.2 XM_011510233.2 XM_011510235.2 XM_011510236.1 XM_017003005.1 XM_017003006.1 XM_017003007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
901933 c.4474G>T p.D1492Y Chr.1: 200554561-200554561 on Build GRCh38
3481249 c.4495C>T p.R1499C Chr.1: 200554540-200554540 on Build GRCh38
4778235 c.4490T>C p.V1497A Chr.1: 200554545-200554545 on Build GRCh38
3418593 c.4533G>A p.E1511E Chr.1: 200554502-200554502 on Build GRCh38
3803386 c.4567+1G>T p.? Chr.1: 200554467-200554467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999313314 Chr.1: 200554167-200554167 on Build GRCh38
rs530095443 Chr.1: 200554352-200554352 on Build GRCh38
rs776757266 Chr.1: 200554537-200554537 on Build GRCh38
rs747148883 Chr.1: 200554535-200554535 on Build GRCh38
rs183237709 Chr.1: 200554306-200554306 on Build GRCh38
rs748716114 Chr.1: 200554529-200554529 on Build GRCh38
rs148994696 Chr.1: 200554385-200554385 on Build GRCh38
rs966440601 Chr.1: 200554130-200554130 on Build GRCh38
rs1014952708 Chr.1: 200554372-200554372 on Build GRCh38
rs946398130 Chr.1: 200554320-200554320 on Build GRCh38
rs904660259 Chr.1: 200554077-200554077 on Build GRCh38
rs767486972 Chr.1: 200554428-200554428 on Build GRCh38
rs190088969 Chr.1: 200554282-200554282 on Build GRCh38
rs921847600 Chr.1: 200554208-200554208 on Build GRCh38
rs753422206 Chr.1: 200554476-200554476 on Build GRCh38
rs759885256 Chr.1: 200554528-200554529 on Build GRCh38
rs543643219 Chr.1: 200554112-200554112 on Build GRCh38
rs774753490 Chr.1: 200554526-200554526 on Build GRCh38
rs755806407 Chr.1: 200554463-200554463 on Build GRCh38
rs1032095119 Chr.1: 200554175-200554175 on Build GRCh38
rs567743363 Chr.1: 200554539-200554539 on Build GRCh38
rs987252930 Chr.1: 200554260-200554260 on Build GRCh38
rs779374714 Chr.1: 200554517-200554517 on Build GRCh38
rs903309449 Chr.1: 200554378-200554378 on Build GRCh38
rs999040299 Chr.1: 200554103-200554103 on Build GRCh38
rs750029644 Chr.1: 200554445-200554445 on Build GRCh38
rs907749325 Chr.1: 200554280-200554280 on Build GRCh38
rs778120976 Chr.1: 200554534-200554534 on Build GRCh38
rs761685646 Chr.1: 200554426-200554426 on Build GRCh38
rs534381955 Chr.1: 200554419-200554419 on Build GRCh38
rs186968526 Chr.1: 200554395-200554395 on Build GRCh38
rs896071851 Chr.1: 200554113-200554113 on Build GRCh38
rs563427428 Chr.1: 200554374-200554374 on Build GRCh38
rs961310863 Chr.1: 200554467-200554467 on Build GRCh38
rs200142247 Chr.1: 200554390-200554391 on Build GRCh38
rs758691608 Chr.1: 200554530-200554530 on Build GRCh38
rs974827307 Chr.1: 200554189-200554189 on Build GRCh38
rs565387163 Chr.1: 200554147-200554147 on Build GRCh38
rs111780903 Chr.1: 200554481-200554481 on Build GRCh38
rs1000371823 Chr.1: 200554381-200554381 on Build GRCh38
rs1020491585 Chr.1: 200554128-200554128 on Build GRCh38
rs146881149 Chr.1: 200554106-200554106 on Build GRCh38
rs185717716 Chr.1: 200554257-200554257 on Build GRCh38
rs920689973 Chr.1: 200554305-200554305 on Build GRCh38
rs767396042 Chr.1: 200554338-200554338 on Build GRCh38
rs1014402738 Chr.1: 200554120-200554120 on Build GRCh38
rs34049962 Chr.1: 200554229-200554229 on Build GRCh38
rs957692955 Chr.1: 200554183-200554183 on Build GRCh38
rs548550733 Chr.1: 200554370-200554370 on Build GRCh38
rs115694135 Chr.1: 200554422-200554422 on Build GRCh38
rs961465829 Chr.1: 200554251-200554251 on Build GRCh38
rs909811045 Chr.1: 200554222-200554222 on Build GRCh38
rs1037711886 Chr.1: 200554301-200554301 on Build GRCh38
rs754489441 Chr.1: 200554479-200554479 on Build GRCh38
rs771402194 Chr.1: 200554521-200554522 on Build GRCh38
rs201072668 Chr.1: 200554568-200554568 on Build GRCh38
rs1022956986 Chr.1: 200554155-200554155 on Build GRCh38
rs992945271 Chr.1: 200554151-200554151 on Build GRCh38
rs937331292 Chr.1: 200554346-200554346 on Build GRCh38
rs150882705 Chr.1: 200554561-200554561 on Build GRCh38
rs7548174 Chr.1: 200554386-200554386 on Build GRCh38
rs763489301 Chr.1: 200554520-200554520 on Build GRCh38
rs762757571 Chr.1: 200554417-200554417 on Build GRCh38
rs771213081 Chr.1: 200554536-200554536 on Build GRCh38
rs765759165 Chr.1: 200554465-200554465 on Build GRCh38
rs769821801 Chr.1: 200554540-200554540 on Build GRCh38
rs374865770 Chr.1: 200554571-200554571 on Build GRCh38
rs1053281619 Chr.1: 200554377-200554377 on Build GRCh38
rs920669091 Chr.1: 200554565-200554565 on Build GRCh38
rs985146208 Chr.1: 200554556-200554556 on Build GRCh38

View Full Product Details