Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203215991-203216482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
UniGene
Hs.201688
Gene Chromosome Location
Chr.1: 203216079-203229732 on Build GRCh38
Transcripts
NM_001256125.1 NM_001270509.1 NM_003465.2 NR_045784.1 NR_045785.1

SNP Detail

SNP ID Chromosome Location
rs1036519894 Chr.1: 203216397-203216397 on Build GRCh38
rs910314287 Chr.1: 203216343-203216343 on Build GRCh38
rs944435133 Chr.1: 203216354-203216354 on Build GRCh38
rs1027615339 Chr.1: 203216452-203216452 on Build GRCh38
rs556167000 Chr.1: 203216031-203216031 on Build GRCh38
rs893036334 Chr.1: 203216393-203216393 on Build GRCh38
rs940896050 Chr.1: 203216012-203216012 on Build GRCh38
rs772189838 Chr.1: 203216041-203216041 on Build GRCh38
rs932221568 Chr.1: 203216322-203216322 on Build GRCh38
rs566193532 Chr.1: 203216381-203216381 on Build GRCh38
rs957467683 Chr.1: 203216195-203216195 on Build GRCh38
rs991335947 Chr.1: 203216249-203216249 on Build GRCh38
rs1024149256 Chr.1: 203216192-203216192 on Build GRCh38
rs751074169 Chr.1: 203216286-203216286 on Build GRCh38
rs561096298 Chr.1: 203216157-203216157 on Build GRCh38
rs920851039 Chr.1: 203216301-203216301 on Build GRCh38
rs781453122 Chr.1: 203216091-203216091 on Build GRCh38
rs773864371 Chr.1: 203216018-203216018 on Build GRCh38
rs958634508 Chr.1: 203216139-203216139 on Build GRCh38
rs563747129 Chr.1: 203216307-203216307 on Build GRCh38
rs376701295 Chr.1: 203216290-203216290 on Build GRCh38
rs189281147 Chr.1: 203216083-203216083 on Build GRCh38
rs72739582 Chr.1: 203216076-203216076 on Build GRCh38
rs147237777 Chr.1: 203216113-203216113 on Build GRCh38
rs749211669 Chr.1: 203216358-203216358 on Build GRCh38
rs148179450 Chr.1: 203216447-203216447 on Build GRCh38
rs773134603 Chr.1: 203216152-203216152 on Build GRCh38
rs920906044 Chr.1: 203216318-203216318 on Build GRCh38
rs777878004 Chr.1: 203216412-203216412 on Build GRCh38
rs756826093 Chr.1: 203216304-203216304 on Build GRCh38
rs924378303 Chr.1: 203216292-203216292 on Build GRCh38
rs935722100 Chr.1: 203216298-203216298 on Build GRCh38
rs780662076 Chr.1: 203216334-203216334 on Build GRCh38
rs541719886 Chr.1: 203216347-203216347 on Build GRCh38
rs763617985 Chr.1: 203216219-203216219 on Build GRCh38
rs771063809 Chr.1: 203216426-203216426 on Build GRCh38
rs747018211 Chr.1: 203216418-203216418 on Build GRCh38
rs1022777083 Chr.1: 203216408-203216408 on Build GRCh38
rs886045830 Chr.1: 203216160-203216160 on Build GRCh38
rs532378393 Chr.1: 203216311-203216311 on Build GRCh38
rs1024164200 Chr.1: 203216261-203216261 on Build GRCh38
rs185074413 Chr.1: 203216237-203216237 on Build GRCh38
rs1003592674 Chr.1: 203216419-203216419 on Build GRCh38
rs890588099 Chr.1: 203216053-203216053 on Build GRCh38
rs982123965 Chr.1: 203216282-203216282 on Build GRCh38
rs773222215 Chr.1: 203216065-203216065 on Build GRCh38
rs888845118 Chr.1: 203216097-203216097 on Build GRCh38
rs998524785 Chr.1: 203216388-203216388 on Build GRCh38
rs528549499 Chr.1: 203216415-203216415 on Build GRCh38
rs903125766 Chr.1: 203216141-203216141 on Build GRCh38
rs775767788 Chr.1: 203216259-203216259 on Build GRCh38
rs1036553945 Chr.1: 203216062-203216062 on Build GRCh38
rs73066394 Chr.1: 203216355-203216355 on Build GRCh38
rs971670790 Chr.1: 203216235-203216235 on Build GRCh38
rs547622776 Chr.1: 203216459-203216459 on Build GRCh38
rs543746463 Chr.1: 203216305-203216305 on Build GRCh38
rs1050602662 Chr.1: 203216319-203216319 on Build GRCh38
rs899292911 Chr.1: 203216072-203216072 on Build GRCh38
rs969449206 Chr.1: 203216274-203216274 on Build GRCh38
rs977278241 Chr.1: 203216278-203216278 on Build GRCh38
rs192633567 Chr.1: 203216107-203216107 on Build GRCh38
rs902822888 Chr.1: 203216362-203216362 on Build GRCh38
rs1051153095 Chr.1: 203216345-203216345 on Build GRCh38
rs897928376 Chr.1: 203216404-203216404 on Build GRCh38
rs541251343 Chr.1: 203216146-203216146 on Build GRCh38
rs772141496 Chr.1: 203216366-203216366 on Build GRCh38

View Full Product Details