Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206960736-206961239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCAMR
Entrez Gene ID
83953
Gene Name
Fc fragment of IgA and IgM receptor
Gene Aliases
CD351, FCA/MR, FKSG87
UniGene
Hs.145519
Gene Chromosome Location
Chr.1: 206957955-206970657 on Build GRCh38
Transcripts
NM_001122979.2 NM_001170631.1 NM_032029.4 XM_011510051.2 XM_011510052.2 XM_017002442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3803699 c.861T>C p.S287S Chr.1: 206960943-206960943 on Build GRCh38
7341295 c.754G>A p.A252T Chr.1: 206961050-206961050 on Build GRCh38
7367982 c.1117C>A p.L373I Chr.1: 206960759-206960759 on Build GRCh38
1583980 c.709G>T p.A237S Chr.1: 206961095-206961095 on Build GRCh38
5426394 c.653-2A>G p.? Chr.1: 206961225-206961225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746443820 Chr.1: 206960941-206960941 on Build GRCh38
rs1026993805 Chr.1: 206960850-206960850 on Build GRCh38
rs545651115 Chr.1: 206960763-206960763 on Build GRCh38
rs537136085 Chr.1: 206961118-206961118 on Build GRCh38
rs756875461 Chr.1: 206960935-206960935 on Build GRCh38
rs924435565 Chr.1: 206960961-206960961 on Build GRCh38
rs763664341 Chr.1: 206960955-206960955 on Build GRCh38
rs552401505 Chr.1: 206960956-206960956 on Build GRCh38
rs541213975 Chr.1: 206960841-206960841 on Build GRCh38
rs200011175 Chr.1: 206961169-206961169 on Build GRCh38
rs3813955 Chr.1: 206960868-206960868 on Build GRCh38
rs751534898 Chr.1: 206960997-206960997 on Build GRCh38
rs1028558608 Chr.1: 206960923-206960923 on Build GRCh38
rs768213670 Chr.1: 206961182-206961182 on Build GRCh38
rs914246398 Chr.1: 206960936-206960936 on Build GRCh38
rs780060252 Chr.1: 206960762-206960762 on Build GRCh38
rs763503910 Chr.1: 206961174-206961174 on Build GRCh38
rs920015755 Chr.1: 206961087-206961087 on Build GRCh38
rs759596589 Chr.1: 206960853-206960853 on Build GRCh38
rs374535427 Chr.1: 206961114-206961114 on Build GRCh38
rs977980801 Chr.1: 206960760-206960760 on Build GRCh38
rs944004647 Chr.1: 206961125-206961125 on Build GRCh38
rs745380215 Chr.1: 206960963-206960963 on Build GRCh38
rs1007881396 Chr.1: 206960944-206960944 on Build GRCh38
rs954402599 Chr.1: 206961206-206961206 on Build GRCh38
rs928686730 Chr.1: 206961034-206961034 on Build GRCh38
rs3813953 Chr.1: 206961065-206961065 on Build GRCh38
rs894415166 Chr.1: 206960840-206960840 on Build GRCh38
rs760914691 Chr.1: 206960797-206960797 on Build GRCh38
rs556798122 Chr.1: 206961138-206961138 on Build GRCh38
rs369380685 Chr.1: 206961157-206961157 on Build GRCh38
rs947124080 Chr.1: 206960938-206960938 on Build GRCh38
rs762438858 Chr.1: 206960900-206960900 on Build GRCh38
rs889695963 Chr.1: 206961124-206961124 on Build GRCh38
rs761835621 Chr.1: 206961004-206961004 on Build GRCh38
rs940097389 Chr.1: 206961052-206961052 on Build GRCh38
rs771122836 Chr.1: 206961133-206961133 on Build GRCh38
rs552971213 Chr.1: 206961219-206961219 on Build GRCh38
rs372580744 Chr.1: 206961203-206961203 on Build GRCh38
rs1038601734 Chr.1: 206961181-206961182 on Build GRCh38
rs1032512197 Chr.1: 206961013-206961013 on Build GRCh38
rs976986132 Chr.1: 206960921-206960921 on Build GRCh38
rs751883557 Chr.1: 206961204-206961204 on Build GRCh38
rs966855376 Chr.1: 206960965-206960965 on Build GRCh38
rs924840474 Chr.1: 206960771-206960771 on Build GRCh38
rs931111058 Chr.1: 206961089-206961089 on Build GRCh38
rs865944502 Chr.1: 206961037-206961037 on Build GRCh38
rs916169526 Chr.1: 206960839-206960839 on Build GRCh38
rs3813954 Chr.1: 206960881-206960881 on Build GRCh38
rs763852435 Chr.1: 206960942-206960942 on Build GRCh38
rs200461962 Chr.1: 206961020-206961020 on Build GRCh38
rs183959195 Chr.1: 206960928-206960928 on Build GRCh38
rs1041108255 Chr.1: 206961143-206961143 on Build GRCh38
rs565464662 Chr.1: 206960770-206960770 on Build GRCh38
rs1032024961 Chr.1: 206961188-206961188 on Build GRCh38
rs548403472 Chr.1: 206961031-206961031 on Build GRCh38
rs893573373 Chr.1: 206961211-206961211 on Build GRCh38
rs938493686 Chr.1: 206960968-206960968 on Build GRCh38
rs565990158 Chr.1: 206961015-206961015 on Build GRCh38
rs901277909 Chr.1: 206961170-206961170 on Build GRCh38
rs376426554 Chr.1: 206961185-206961185 on Build GRCh38
rs999324287 Chr.1: 206960991-206960991 on Build GRCh38
rs757629990 Chr.1: 206961218-206961218 on Build GRCh38
rs747912949 Chr.1: 206961120-206961120 on Build GRCh38
rs17018337 Chr.1: 206960847-206960847 on Build GRCh38
rs375134753 Chr.1: 206960786-206960786 on Build GRCh38
rs530110722 Chr.1: 206960849-206960849 on Build GRCh38
rs201258954 Chr.1: 206961208-206961208 on Build GRCh38
rs539751187 Chr.1: 206960778-206960778 on Build GRCh38
rs376163382 Chr.1: 206960877-206960877 on Build GRCh38
rs779946834 Chr.1: 206961027-206961027 on Build GRCh38
rs761593099 Chr.1: 206961032-206961032 on Build GRCh38
rs61729352 Chr.1: 206960759-206960759 on Build GRCh38
rs751890654 Chr.1: 206960931-206960931 on Build GRCh38
rs373309798 Chr.1: 206961153-206961153 on Build GRCh38

View Full Product Details