Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207468668-207469208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR2
Entrez Gene ID
1380
Gene Name
complement component 3d receptor 2
Gene Aliases
C3DR, CD21, CR, CVID7, SLEB9
UniGene
Hs.445757
Gene Chromosome Location
Chr.1: 207454300-207489895 on Build GRCh38
Transcripts
NM_001006658.2 NM_001877.4 XM_011509206.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997814 c.620C>T p.P207L Chr.1: 207468701-207468701 on Build GRCh38
5039782 c.784G>A p.G262R Chr.1: 207469199-207469199 on Build GRCh38
3730831 c.602G>A p.G201E Chr.1: 207468683-207468683 on Build GRCh38
4027604 c.788T>C p.V263A Chr.1: 207469203-207469203 on Build GRCh38
6541657 c.725G>T p.C242F Chr.1: 207468890-207468890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950554487 Chr.1: 207468724-207468724 on Build GRCh38
rs749077626 Chr.1: 207468875-207468875 on Build GRCh38
rs745918801 Chr.1: 207469150-207469150 on Build GRCh38
rs745407153 Chr.1: 207468850-207468850 on Build GRCh38
rs149087222 Chr.1: 207468863-207468863 on Build GRCh38
rs755524953 Chr.1: 207468720-207468720 on Build GRCh38
rs776548804 Chr.1: 207468907-207468907 on Build GRCh38
rs776118230 Chr.1: 207468941-207468941 on Build GRCh38
rs759180891 Chr.1: 207469182-207469182 on Build GRCh38
rs200947637 Chr.1: 207469120-207469120 on Build GRCh38
rs542606446 Chr.1: 207468927-207468927 on Build GRCh38
rs779082480 Chr.1: 207468731-207468731 on Build GRCh38
rs966437607 Chr.1: 207468793-207468793 on Build GRCh38
rs17045039 Chr.1: 207468780-207468780 on Build GRCh38
rs150622293 Chr.1: 207469155-207469155 on Build GRCh38
rs996675765 Chr.1: 207468972-207468972 on Build GRCh38
rs747170469 Chr.1: 207468895-207468895 on Build GRCh38
rs564550690 Chr.1: 207469046-207469046 on Build GRCh38
rs927776538 Chr.1: 207468836-207468836 on Build GRCh38
rs771024465 Chr.1: 207469142-207469142 on Build GRCh38
rs199889815 Chr.1: 207468792-207468792 on Build GRCh38
rs774149171 Chr.1: 207468894-207468894 on Build GRCh38
rs1028968402 Chr.1: 207469077-207469077 on Build GRCh38
rs747982668 Chr.1: 207469132-207469132 on Build GRCh38
rs141831783 Chr.1: 207468806-207468806 on Build GRCh38
rs1049036054 Chr.1: 207468939-207468939 on Build GRCh38
rs200630015 Chr.1: 207469123-207469123 on Build GRCh38
rs371733599 Chr.1: 207468830-207468830 on Build GRCh38
rs766055159 Chr.1: 207468979-207468981 on Build GRCh38
rs373992562 Chr.1: 207468786-207468786 on Build GRCh38
rs779846456 Chr.1: 207469119-207469119 on Build GRCh38
rs61759494 Chr.1: 207468705-207468705 on Build GRCh38
rs368201764 Chr.1: 207468754-207468754 on Build GRCh38
rs562373961 Chr.1: 207468944-207468944 on Build GRCh38
rs749871372 Chr.1: 207469106-207469106 on Build GRCh38
rs767250244 Chr.1: 207469100-207469100 on Build GRCh38
rs546816560 Chr.1: 207469115-207469115 on Build GRCh38
rs377469210 Chr.1: 207468697-207468697 on Build GRCh38
rs923624124 Chr.1: 207468820-207468820 on Build GRCh38
rs61754519 Chr.1: 207469178-207469178 on Build GRCh38
rs939101643 Chr.1: 207468839-207468839 on Build GRCh38
rs573721416 Chr.1: 207468812-207468812 on Build GRCh38
rs780740575 Chr.1: 207468698-207468699 on Build GRCh38
rs758097858 Chr.1: 207468749-207468749 on Build GRCh38
rs771175429 Chr.1: 207468901-207468901 on Build GRCh38
rs749158154 Chr.1: 207468787-207468787 on Build GRCh38
rs754918026 Chr.1: 207469122-207469122 on Build GRCh38
rs201760344 Chr.1: 207468805-207468805 on Build GRCh38
rs776916652 Chr.1: 207468968-207468968 on Build GRCh38
rs763512398 Chr.1: 207469166-207469166 on Build GRCh38
rs781540705 Chr.1: 207469145-207469145 on Build GRCh38
rs769307763 Chr.1: 207469167-207469167 on Build GRCh38
rs374710389 Chr.1: 207468833-207468833 on Build GRCh38
rs778177265 Chr.1: 207468788-207468788 on Build GRCh38
rs894817260 Chr.1: 207468911-207468911 on Build GRCh38
rs994207068 Chr.1: 207469032-207469032 on Build GRCh38
rs778221461 Chr.1: 207468831-207468831 on Build GRCh38
rs771819232 Chr.1: 207468715-207468715 on Build GRCh38
rs985348012 Chr.1: 207468772-207468772 on Build GRCh38
rs769372920 Chr.1: 207468699-207468699 on Build GRCh38
rs80315759 Chr.1: 207469081-207469081 on Build GRCh38
rs544807460 Chr.1: 207469001-207469001 on Build GRCh38
rs760733654 Chr.1: 207468801-207468801 on Build GRCh38
rs747832403 Chr.1: 207468699-207468700 on Build GRCh38
rs147633291 Chr.1: 207468824-207468824 on Build GRCh38
rs966119787 Chr.1: 207468969-207468969 on Build GRCh38
rs370988625 Chr.1: 207468716-207468716 on Build GRCh38
rs573346582 Chr.1: 207468702-207468702 on Build GRCh38
rs759698195 Chr.1: 207468923-207468923 on Build GRCh38
rs766392071 Chr.1: 207468804-207468804 on Build GRCh38
rs745724633 Chr.1: 207468778-207468778 on Build GRCh38
rs746566723 Chr.1: 207468693-207468693 on Build GRCh38
rs141472681 Chr.1: 207468823-207468823 on Build GRCh38
rs754364730 Chr.1: 207468719-207468719 on Build GRCh38
rs755984980 Chr.1: 207468862-207468862 on Build GRCh38
rs746509584 Chr.1: 207468771-207468771 on Build GRCh38
rs369546456 Chr.1: 207469152-207469152 on Build GRCh38
rs756884222 Chr.1: 207468774-207468774 on Build GRCh38
rs1048415960 Chr.1: 207469042-207469042 on Build GRCh38
rs780861723 Chr.1: 207468775-207468775 on Build GRCh38
rs774836892 Chr.1: 207469175-207469175 on Build GRCh38
rs749636258 Chr.1: 207468882-207468883 on Build GRCh38
rs897223483 Chr.1: 207468991-207468991 on Build GRCh38
rs769415969 Chr.1: 207468703-207468703 on Build GRCh38
rs1042114463 Chr.1: 207468834-207468834 on Build GRCh38
rs772252200 Chr.1: 207468791-207468791 on Build GRCh38
rs752969830 Chr.1: 207468815-207468815 on Build GRCh38
rs1507763 Chr.1: 207468888-207468888 on Build GRCh38
rs368883636 Chr.1: 207468893-207468893 on Build GRCh38
rs763567541 Chr.1: 207468942-207468942 on Build GRCh38
rs1507764 Chr.1: 207468950-207468950 on Build GRCh38
rs375253183 Chr.1: 207468732-207468732 on Build GRCh38

View Full Product Details