Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207470655-207471163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR2
Entrez Gene ID
1380
Gene Name
complement component 3d receptor 2
Gene Aliases
C3DR, CD21, CR, CVID7, SLEB9
UniGene
Hs.445757
Gene Chromosome Location
Chr.1: 207454300-207489895 on Build GRCh38
Transcripts
NM_001006658.2 NM_001877.4 XM_011509206.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1748123 c.1385C>G p.P462R Chr.1: 207470899-207470899 on Build GRCh38
7078556 c.1405G>T p.A469S Chr.1: 207470999-207470999 on Build GRCh38
903122 c.1291C>T p.P431S Chr.1: 207470805-207470805 on Build GRCh38
5794378 c.1470T>A p.D490E Chr.1: 207471064-207471064 on Build GRCh38
1718502 c.1349_1350CC>TT p.S450F Chr.1: 207470863-207470864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764972735 Chr.1: 207470752-207470752 on Build GRCh38
rs765653703 Chr.1: 207470886-207470886 on Build GRCh38
rs760851517 Chr.1: 207470861-207470861 on Build GRCh38
rs761478463 Chr.1: 207471108-207471108 on Build GRCh38
rs752327442 Chr.1: 207470776-207470776 on Build GRCh38
rs35264911 Chr.1: 207470899-207470900 on Build GRCh38
rs753628062 Chr.1: 207471019-207471019 on Build GRCh38
rs770418433 Chr.1: 207471070-207471070 on Build GRCh38
rs761376121 Chr.1: 207470983-207470983 on Build GRCh38
rs770988364 Chr.1: 207470958-207470958 on Build GRCh38
rs953097565 Chr.1: 207470709-207470709 on Build GRCh38
rs756518877 Chr.1: 207470920-207470920 on Build GRCh38
rs773257775 Chr.1: 207470847-207470847 on Build GRCh38
rs776195185 Chr.1: 207471084-207471084 on Build GRCh38
rs748950155 Chr.1: 207470924-207470924 on Build GRCh38
rs767518977 Chr.1: 207470712-207470712 on Build GRCh38
rs370217966 Chr.1: 207470959-207470959 on Build GRCh38
rs977189143 Chr.1: 207470781-207470781 on Build GRCh38
rs748329891 Chr.1: 207470842-207470842 on Build GRCh38
rs771075787 Chr.1: 207471064-207471064 on Build GRCh38
rs756077508 Chr.1: 207470918-207470918 on Build GRCh38
rs770469733 Chr.1: 207470974-207470974 on Build GRCh38
rs545091003 Chr.1: 207471003-207471003 on Build GRCh38
rs376685974 Chr.1: 207470845-207470845 on Build GRCh38
rs761039132 Chr.1: 207470726-207470726 on Build GRCh38
rs773313660 Chr.1: 207470713-207470713 on Build GRCh38
rs1033642945 Chr.1: 207470875-207470875 on Build GRCh38
rs1021531705 Chr.1: 207470714-207470714 on Build GRCh38
rs775269251 Chr.1: 207470836-207470836 on Build GRCh38
rs955457959 Chr.1: 207470777-207470778 on Build GRCh38
rs183067414 Chr.1: 207470797-207470797 on Build GRCh38
rs201838281 Chr.1: 207470896-207470896 on Build GRCh38
rs751165557 Chr.1: 207470981-207470981 on Build GRCh38
rs755441915 Chr.1: 207470750-207470750 on Build GRCh38
rs1038638960 Chr.1: 207470695-207470695 on Build GRCh38
rs916035659 Chr.1: 207471129-207471129 on Build GRCh38
rs765794765 Chr.1: 207470771-207470771 on Build GRCh38
rs778039241 Chr.1: 207470949-207470949 on Build GRCh38
rs143872741 Chr.1: 207471062-207471062 on Build GRCh38
rs773747334 Chr.1: 207470910-207470910 on Build GRCh38
rs368025734 Chr.1: 207470767-207470767 on Build GRCh38
rs565397037 Chr.1: 207470788-207470788 on Build GRCh38
rs777257410 Chr.1: 207470880-207470880 on Build GRCh38
rs780027591 Chr.1: 207470950-207470950 on Build GRCh38
rs41313740 Chr.1: 207470904-207470904 on Build GRCh38
rs753132648 Chr.1: 207470890-207470890 on Build GRCh38
rs201585844 Chr.1: 207470810-207470810 on Build GRCh38
rs751627502 Chr.1: 207470760-207470760 on Build GRCh38
rs374531845 Chr.1: 207470713-207470714 on Build GRCh38
rs188078143 Chr.1: 207470909-207470909 on Build GRCh38
rs142212472 Chr.1: 207471069-207471069 on Build GRCh38
rs985999522 Chr.1: 207470710-207470710 on Build GRCh38
rs749132096 Chr.1: 207470933-207470933 on Build GRCh38
rs368563787 Chr.1: 207470976-207470976 on Build GRCh38
rs376278193 Chr.1: 207470975-207470975 on Build GRCh38
rs771341490 Chr.1: 207470867-207470867 on Build GRCh38
rs371167100 Chr.1: 207470984-207470984 on Build GRCh38
rs763823010 Chr.1: 207470980-207470980 on Build GRCh38
rs41274754 Chr.1: 207470922-207470922 on Build GRCh38
rs774611422 Chr.1: 207471105-207471105 on Build GRCh38
rs762670047 Chr.1: 207470894-207470894 on Build GRCh38
rs749924559 Chr.1: 207470997-207470997 on Build GRCh38
rs373176986 Chr.1: 207470796-207470796 on Build GRCh38
rs538276366 Chr.1: 207470732-207470732 on Build GRCh38
rs375667838 Chr.1: 207471098-207471098 on Build GRCh38
rs565317532 Chr.1: 207471004-207471004 on Build GRCh38
rs377088084 Chr.1: 207470953-207470953 on Build GRCh38
rs147378770 Chr.1: 207471052-207471052 on Build GRCh38
rs767126107 Chr.1: 207471121-207471121 on Build GRCh38
rs959926708 Chr.1: 207471122-207471122 on Build GRCh38
rs372838534 Chr.1: 207470917-207470917 on Build GRCh38
rs781307709 Chr.1: 207471066-207471066 on Build GRCh38
rs531242388 Chr.1: 207470712-207470713 on Build GRCh38
rs756848982 Chr.1: 207470905-207470905 on Build GRCh38
rs781255614 Chr.1: 207470809-207470809 on Build GRCh38
rs754830128 Chr.1: 207471026-207471026 on Build GRCh38
rs760035052 Chr.1: 207470970-207470970 on Build GRCh38
rs200000368 Chr.1: 207470956-207470956 on Build GRCh38
rs983227869 Chr.1: 207470925-207470925 on Build GRCh38
rs749825341 Chr.1: 207471097-207471097 on Build GRCh38
rs768417104 Chr.1: 207470940-207470940 on Build GRCh38
rs891263800 Chr.1: 207471072-207471072 on Build GRCh38
rs756071188 Chr.1: 207470694-207470694 on Build GRCh38
rs762037340 Chr.1: 207470701-207470701 on Build GRCh38
rs772749374 Chr.1: 207471123-207471123 on Build GRCh38
rs150464766 Chr.1: 207470897-207470897 on Build GRCh38

View Full Product Details