Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212035617-212036114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTL
Entrez Gene ID
51514
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases
CDT2, DCAF2, L2DTL, RAMP
UniGene
Hs.656473
Gene Chromosome Location
Chr.1: 212035553-212105006 on Build GRCh38
Transcripts
NM_001286229.1 NM_001286230.1 NM_016448.3 XM_011509614.1 NM_001199809.1 NM_001199811.1 NM_001199812.1 NM_015434.3 NR_037667.1

Gene Symbol
INTS7
Entrez Gene ID
25896
Gene Name
integrator complex subunit 7
Gene Aliases
C1orf73, INT7
UniGene
Hs.369285
Gene Chromosome Location
Chr.1: 211940328-212035660 on Build GRCh38
Transcripts
NM_001286229.1 NM_001286230.1 NM_016448.3 XM_011509614.1 NM_001199809.1 NM_001199811.1 NM_001199812.1 NM_015434.3 NR_037667.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5969137 c.21C>T p.L7L Chr.1: 212035911-212035911 on Build GRCh38
4978830 c.42C>T p.V14V Chr.1: 212035932-212035932 on Build GRCh38
6649941 c.41T>A p.V14D Chr.1: 212035931-212035931 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199630708 Chr.1: 212035959-212035959 on Build GRCh38
rs141451521 Chr.1: 212035901-212035901 on Build GRCh38
rs762734785 Chr.1: 212035957-212035979 on Build GRCh38
rs371950899 Chr.1: 212035834-212035834 on Build GRCh38
rs879824749 Chr.1: 212035815-212035815 on Build GRCh38
rs755847733 Chr.1: 212035910-212035910 on Build GRCh38
rs964030360 Chr.1: 212035798-212035798 on Build GRCh38
rs769664179 Chr.1: 212035852-212035854 on Build GRCh38
rs562016206 Chr.1: 212035762-212035762 on Build GRCh38
rs779261579 Chr.1: 212035911-212035911 on Build GRCh38
rs374285656 Chr.1: 212035968-212035968 on Build GRCh38
rs748324270 Chr.1: 212035913-212035913 on Build GRCh38
rs3215984 Chr.1: 212036025-212036026 on Build GRCh38
rs143100007 Chr.1: 212035941-212035941 on Build GRCh38
rs17018373 Chr.1: 212035692-212035692 on Build GRCh38
rs201537406 Chr.1: 212035780-212035780 on Build GRCh38
rs988078370 Chr.1: 212035727-212035727 on Build GRCh38
rs781581481 Chr.1: 212035771-212035771 on Build GRCh38
rs759790328 Chr.1: 212035880-212035880 on Build GRCh38
rs937441393 Chr.1: 212035814-212035814 on Build GRCh38
rs377204418 Chr.1: 212036019-212036019 on Build GRCh38
rs752282827 Chr.1: 212035883-212035883 on Build GRCh38
rs35155238 Chr.1: 212036056-212036057 on Build GRCh38
rs201116095 Chr.1: 212035888-212035888 on Build GRCh38
rs757288264 Chr.1: 212035893-212035893 on Build GRCh38
rs551746376 Chr.1: 212035823-212035823 on Build GRCh38
rs773091965 Chr.1: 212035881-212035882 on Build GRCh38
rs762531187 Chr.1: 212035961-212035961 on Build GRCh38
rs896044853 Chr.1: 212035650-212035650 on Build GRCh38
rs368919987 Chr.1: 212035806-212035806 on Build GRCh38
rs1029481463 Chr.1: 212035969-212035969 on Build GRCh38
rs1027184327 Chr.1: 212035803-212035803 on Build GRCh38
rs756013063 Chr.1: 212035989-212035989 on Build GRCh38
rs747143700 Chr.1: 212035994-212035994 on Build GRCh38
rs767619478 Chr.1: 212035985-212035985 on Build GRCh38
rs960125424 Chr.1: 212035841-212035841 on Build GRCh38
rs368167607 Chr.1: 212035991-212035991 on Build GRCh38
rs397844077 Chr.1: 212036027-212036028 on Build GRCh38
rs745865952 Chr.1: 212035997-212035998 on Build GRCh38
rs765519365 Chr.1: 212035957-212035957 on Build GRCh38
rs773209187 Chr.1: 212035851-212035851 on Build GRCh38
rs756804386 Chr.1: 212035971-212035971 on Build GRCh38
rs138210630 Chr.1: 212035933-212035933 on Build GRCh38
rs777102905 Chr.1: 212035947-212035947 on Build GRCh38
rs1026609550 Chr.1: 212036083-212036083 on Build GRCh38
rs908589327 Chr.1: 212035693-212035693 on Build GRCh38
rs377222218 Chr.1: 212035773-212035773 on Build GRCh38
rs968002402 Chr.1: 212036037-212036037 on Build GRCh38
rs547937414 Chr.1: 212035778-212035778 on Build GRCh38
rs879158994 Chr.1: 212035782-212035782 on Build GRCh38
rs567798044 Chr.1: 212036069-212036069 on Build GRCh38
rs913216819 Chr.1: 212035731-212035731 on Build GRCh38
rs1018479578 Chr.1: 212035686-212035686 on Build GRCh38
rs867345623 Chr.1: 212035668-212035668 on Build GRCh38
rs975623294 Chr.1: 212035680-212035680 on Build GRCh38
rs1008405954 Chr.1: 212035787-212035787 on Build GRCh38
rs372592921 Chr.1: 212035703-212035703 on Build GRCh38
rs760008177 Chr.1: 212035951-212035951 on Build GRCh38
rs1019387339 Chr.1: 212035789-212035789 on Build GRCh38
rs920201032 Chr.1: 212035691-212035691 on Build GRCh38
rs903812795 Chr.1: 212035758-212035758 on Build GRCh38
rs1044986485 Chr.1: 212035730-212035730 on Build GRCh38
rs186783778 Chr.1: 212035679-212035679 on Build GRCh38
rs1000694212 Chr.1: 212035761-212035761 on Build GRCh38
rs367698385 Chr.1: 212035695-212035695 on Build GRCh38
rs952940456 Chr.1: 212036087-212036087 on Build GRCh38
rs772042132 Chr.1: 212035844-212035844 on Build GRCh38
rs908719635 Chr.1: 212035830-212035830 on Build GRCh38
rs772279926 Chr.1: 212035924-212035924 on Build GRCh38
rs763797805 Chr.1: 212035962-212035962 on Build GRCh38
rs200995195 Chr.1: 212035879-212035879 on Build GRCh38
rs778572459 Chr.1: 212035992-212035992 on Build GRCh38
rs979145066 Chr.1: 212036080-212036080 on Build GRCh38
rs191900771 Chr.1: 212035793-212035793 on Build GRCh38
rs935210383 Chr.1: 212035708-212035708 on Build GRCh38
rs915180478 Chr.1: 212036015-212036015 on Build GRCh38
rs986449795 Chr.1: 212035654-212035654 on Build GRCh38
rs771471470 Chr.1: 212035945-212035945 on Build GRCh38
rs763521434 Chr.1: 212035892-212035892 on Build GRCh38
rs747284312 Chr.1: 212035847-212035847 on Build GRCh38
rs745705190 Chr.1: 212035904-212035904 on Build GRCh38
rs529680452 Chr.1: 212035675-212035675 on Build GRCh38
rs544900414 Chr.1: 212035677-212035677 on Build GRCh38
rs369778039 Chr.1: 212035990-212035990 on Build GRCh38
rs533405137 Chr.1: 212035801-212035801 on Build GRCh38
rs878883711 Chr.1: 212035653-212035653 on Build GRCh38
rs766824045 Chr.1: 212035873-212035873 on Build GRCh38
rs1025291820 Chr.1: 212035660-212035660 on Build GRCh38
rs370109409 Chr.1: 212035863-212035863 on Build GRCh38

View Full Product Details