Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212883056-212883544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR1
Entrez Gene ID
28982
Gene Name
feline leukemia virus subgroup C cellular receptor 1
Gene Aliases
AXPC1, FLVCR, MFSD7B, PCA, PCARP
UniGene
Hs.7055
Gene Chromosome Location
Chr.1: 212858255-212899363 on Build GRCh38
Transcripts
NM_014053.3 XM_011509446.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7311767 c.1051G>T p.V351F Chr.1: 212883397-212883397 on Build GRCh38
1748144 c.1078T>C p.L360L Chr.1: 212883424-212883424 on Build GRCh38
6567188 c.1055C>T p.S352L Chr.1: 212883401-212883401 on Build GRCh38
344957 c.1090G>C p.E364Q Chr.1: 212883436-212883436 on Build GRCh38
1162863 c.1073T>C p.M358T Chr.1: 212883419-212883419 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs189319527 Chr.1: 212883165-212883165 on Build GRCh38
rs896464023 Chr.1: 212883177-212883177 on Build GRCh38
rs1026384424 Chr.1: 212883209-212883209 on Build GRCh38
rs12750027 Chr.1: 212883085-212883085 on Build GRCh38
rs761553036 Chr.1: 212883381-212883381 on Build GRCh38
rs778581914 Chr.1: 212883420-212883420 on Build GRCh38
rs775559523 Chr.1: 212883373-212883375 on Build GRCh38
rs904095087 Chr.1: 212883126-212883126 on Build GRCh38
rs4951649 Chr.1: 212883151-212883151 on Build GRCh38
rs2644547 Chr.1: 212883477-212883477 on Build GRCh38
rs766231335 Chr.1: 212883200-212883200 on Build GRCh38
rs767302510 Chr.1: 212883383-212883383 on Build GRCh38
rs964799808 Chr.1: 212883230-212883230 on Build GRCh38
rs946805892 Chr.1: 212883162-212883162 on Build GRCh38
rs1044309561 Chr.1: 212883114-212883114 on Build GRCh38
rs746384895 Chr.1: 212883322-212883322 on Build GRCh38
rs774455543 Chr.1: 212883374-212883374 on Build GRCh38
rs974592236 Chr.1: 212883371-212883371 on Build GRCh38
rs775629662 Chr.1: 212883346-212883346 on Build GRCh38
rs140925894 Chr.1: 212883465-212883465 on Build GRCh38
rs1002420662 Chr.1: 212883149-212883149 on Build GRCh38
rs1029517048 Chr.1: 212883120-212883120 on Build GRCh38
rs754495724 Chr.1: 212883419-212883419 on Build GRCh38
rs939591772 Chr.1: 212883474-212883474 on Build GRCh38
rs920718884 Chr.1: 212883295-212883295 on Build GRCh38
rs752305026 Chr.1: 212883354-212883357 on Build GRCh38
rs369868359 Chr.1: 212883351-212883351 on Build GRCh38
rs550517136 Chr.1: 212883405-212883405 on Build GRCh38
rs999231233 Chr.1: 212883108-212883108 on Build GRCh38
rs762035273 Chr.1: 212883490-212883490 on Build GRCh38
rs950886489 Chr.1: 212883262-212883262 on Build GRCh38
rs372728084 Chr.1: 212883404-212883404 on Build GRCh38
rs1006051421 Chr.1: 212883327-212883327 on Build GRCh38
rs878856637 Chr.1: 212883478-212883478 on Build GRCh38
rs750300828 Chr.1: 212883403-212883403 on Build GRCh38
rs763296311 Chr.1: 212883361-212883361 on Build GRCh38
rs973907251 Chr.1: 212883258-212883258 on Build GRCh38
rs780471756 Chr.1: 212883453-212883453 on Build GRCh38
rs199842709 Chr.1: 212883431-212883431 on Build GRCh38
rs369619291 Chr.1: 212883328-212883328 on Build GRCh38
rs879413997 Chr.1: 212883274-212883274 on Build GRCh38
rs753358647 Chr.1: 212883408-212883408 on Build GRCh38
rs752325722 Chr.1: 212883424-212883424 on Build GRCh38
rs912963443 Chr.1: 212883504-212883504 on Build GRCh38
rs556788423 Chr.1: 212883443-212883443 on Build GRCh38
rs757631822 Chr.1: 212883428-212883428 on Build GRCh38
rs746297154 Chr.1: 212883435-212883435 on Build GRCh38

View Full Product Details