Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 216506423-216506955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRG
Entrez Gene ID
2104
Gene Name
estrogen related receptor gamma
Gene Aliases
ERR3, ERRgamma, NR3B3
UniGene
Hs.444225
Gene Chromosome Location
Chr.1: 216503246-217137757 on Build GRCh38
Transcripts
NM_001134285.2 NM_001243505.1 NM_001243506.1 NM_001243507.1 NM_001243509.1 NM_001243510.2 NM_001243511.2 NM_001243512.1 NM_001243513.1 NM_001243514.1 NM_001243515.1 NM_001243518.1 NM_001243519.1 NM_001438.3 NM_206594.2 NM_206595.2 XM_011509265.2 XM_011509266.2 XM_011509267.1 XM_011509268.2 XM_011509269.2 XM_011509270.1 XM_011509271.2 XM_011509274.1 XM_011509275.1 XM_011509276.1 XM_011509277.1 XM_011509278.1 XM_011509279.1 XM_011509280.2 XM_017000621.1 XM_017000622.1 XM_017000623.1 XM_017000624.1 XM_017000625.1 XM_017000626.1 XM_017000627.1 XM_017000628.1 XM_017000629.1 XM_017000630.1 XM_017000631.1 XM_017000632.1 XM_017000633.1 XM_017000634.1 XM_017000635.1 XM_017000636.1 XM_017000637.1 XM_017000638.1 XM_017000639.1 XM_017000640.1 XM_017000641.1 XM_017000642.1 XM_017000643.1 XM_017000644.1 XM_017000645.1 XM_017000646.1 XM_017000647.1 XM_017000648.1 XM_017000649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1338888 c.1370A>C p.K457T Chr.1: 216506946-216506946 on Build GRCh38
4028249 c.1322A>G p.K441R Chr.1: 216506946-216506946 on Build GRCh38
374752 c.1371G>T p.K457N Chr.1: 216506945-216506945 on Build GRCh38
4784964 c.1322A>C p.K441T Chr.1: 216506946-216506946 on Build GRCh38
381773 c.1371G>C p.K457N Chr.1: 216506945-216506945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866187110 Chr.1: 216506652-216506652 on Build GRCh38
rs1012808865 Chr.1: 216506785-216506785 on Build GRCh38
rs186983451 Chr.1: 216506859-216506859 on Build GRCh38
rs777925244 Chr.1: 216506894-216506894 on Build GRCh38
rs370166585 Chr.1: 216506932-216506932 on Build GRCh38
rs11572833 Chr.1: 216506575-216506575 on Build GRCh38
rs150035363 Chr.1: 216506473-216506473 on Build GRCh38
rs747409872 Chr.1: 216506565-216506565 on Build GRCh38
rs996942113 Chr.1: 216506730-216506730 on Build GRCh38
rs576163918 Chr.1: 216506822-216506822 on Build GRCh38
rs982063044 Chr.1: 216506492-216506492 on Build GRCh38
rs398103695 Chr.1: 216506525-216506526 on Build GRCh38
rs1027983774 Chr.1: 216506842-216506842 on Build GRCh38
rs1012449328 Chr.1: 216506476-216506476 on Build GRCh38
rs747894987 Chr.1: 216506620-216506620 on Build GRCh38
rs535690445 Chr.1: 216506588-216506588 on Build GRCh38
rs775010762 Chr.1: 216506666-216506668 on Build GRCh38
rs973254299 Chr.1: 216506582-216506582 on Build GRCh38
rs867335631 Chr.1: 216506488-216506488 on Build GRCh38
rs987578279 Chr.1: 216506479-216506479 on Build GRCh38
rs1054008284 Chr.1: 216506779-216506779 on Build GRCh38
rs1044068423 Chr.1: 216506705-216506708 on Build GRCh38
rs11117604 Chr.1: 216506465-216506465 on Build GRCh38
rs754031489 Chr.1: 216506577-216506577 on Build GRCh38
rs796363914 Chr.1: 216506728-216506728 on Build GRCh38
rs533200351 Chr.1: 216506716-216506716 on Build GRCh38
rs373580120 Chr.1: 216506876-216506876 on Build GRCh38
rs1023831503 Chr.1: 216506799-216506799 on Build GRCh38
rs962284313 Chr.1: 216506537-216506537 on Build GRCh38
rs773344558 Chr.1: 216506872-216506872 on Build GRCh38
rs993754386 Chr.1: 216506798-216506798 on Build GRCh38
rs144848582 Chr.1: 216506516-216506517 on Build GRCh38
rs945311244 Chr.1: 216506627-216506627 on Build GRCh38
rs3768022 Chr.1: 216506743-216506743 on Build GRCh38
rs11572832 Chr.1: 216506628-216506628 on Build GRCh38
rs190937299 Chr.1: 216506617-216506617 on Build GRCh38
rs911613533 Chr.1: 216506693-216506693 on Build GRCh38
rs770676279 Chr.1: 216506467-216506468 on Build GRCh38
rs776154929 Chr.1: 216506921-216506921 on Build GRCh38
rs770395096 Chr.1: 216506920-216506920 on Build GRCh38
rs1024134966 Chr.1: 216506478-216506478 on Build GRCh38
rs759650508 Chr.1: 216506898-216506898 on Build GRCh38
rs879497632 Chr.1: 216506592-216506592 on Build GRCh38
rs543865024 Chr.1: 216506773-216506773 on Build GRCh38
rs868130728 Chr.1: 216506922-216506922 on Build GRCh38
rs962561109 Chr.1: 216506796-216506796 on Build GRCh38
rs200351838 Chr.1: 216506463-216506463 on Build GRCh38
rs781500087 Chr.1: 216506707-216506707 on Build GRCh38
rs201367502 Chr.1: 216506526-216506526 on Build GRCh38
rs920519408 Chr.1: 216506586-216506586 on Build GRCh38
rs72735136 Chr.1: 216506489-216506489 on Build GRCh38
rs961984203 Chr.1: 216506835-216506835 on Build GRCh38
rs906705830 Chr.1: 216506821-216506821 on Build GRCh38
rs1014848967 Chr.1: 216506525-216506525 on Build GRCh38
rs1007941410 Chr.1: 216506781-216506781 on Build GRCh38
rs994832935 Chr.1: 216506838-216506838 on Build GRCh38
rs544931443 Chr.1: 216506560-216506560 on Build GRCh38
rs200205260 Chr.1: 216506890-216506890 on Build GRCh38
rs556807719 Chr.1: 216506707-216506708 on Build GRCh38
rs769365768 Chr.1: 216506930-216506930 on Build GRCh38
rs1019085075 Chr.1: 216506531-216506531 on Build GRCh38
rs916224536 Chr.1: 216506455-216506466 on Build GRCh38
rs748058972 Chr.1: 216506511-216506511 on Build GRCh38
rs755632986 Chr.1: 216506837-216506837 on Build GRCh38
rs146420215 Chr.1: 216506864-216506864 on Build GRCh38
rs749817803 Chr.1: 216506867-216506867 on Build GRCh38
rs3768021 Chr.1: 216506929-216506929 on Build GRCh38
rs534696933 Chr.1: 216506701-216506701 on Build GRCh38
rs182373024 Chr.1: 216506783-216506783 on Build GRCh38
rs185243950 Chr.1: 216506556-216506556 on Build GRCh38
rs890848703 Chr.1: 216506776-216506776 on Build GRCh38
rs557686086 Chr.1: 216506739-216506739 on Build GRCh38
rs980745069 Chr.1: 216506815-216506815 on Build GRCh38
rs952175984 Chr.1: 216506569-216506570 on Build GRCh38
rs746931549 Chr.1: 216506903-216506903 on Build GRCh38
rs181362709 Chr.1: 216506487-216506487 on Build GRCh38
rs771998273 Chr.1: 216506516-216506516 on Build GRCh38
rs952061135 Chr.1: 216506801-216506801 on Build GRCh38
rs763233614 Chr.1: 216506494-216506495 on Build GRCh38
rs775113591 Chr.1: 216506931-216506931 on Build GRCh38
rs903556670 Chr.1: 216506461-216506465 on Build GRCh38
rs77094478 Chr.1: 216506618-216506618 on Build GRCh38
rs1014963562 Chr.1: 216506828-216506828 on Build GRCh38

View Full Product Details