Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223394030-223394532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC185
Entrez Gene ID
164127
Gene Name
coiled-coil domain containing 185
Gene Aliases
C1orf65
UniGene
Hs.192090
Gene Chromosome Location
Chr.1: 223393373-223395470 on Build GRCh38
Transcripts
NM_152610.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750903 c.584T>C p.V195A Chr.1: 223394059-223394059 on Build GRCh38
1339122 c.713C>T p.P238L Chr.1: 223394188-223394188 on Build GRCh38
6433736 c.899G>A p.R300Q Chr.1: 223394374-223394374 on Build GRCh38
3418804 c.739C>T p.L247L Chr.1: 223394214-223394214 on Build GRCh38
3966297 c.574C>T p.R192W Chr.1: 223394049-223394049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761569431 Chr.1: 223394334-223394334 on Build GRCh38
rs1020729031 Chr.1: 223394266-223394266 on Build GRCh38
rs765942457 Chr.1: 223394430-223394430 on Build GRCh38
rs773266713 Chr.1: 223394471-223394471 on Build GRCh38
rs199694908 Chr.1: 223394344-223394344 on Build GRCh38
rs779449042 Chr.1: 223394128-223394128 on Build GRCh38
rs780231317 Chr.1: 223394093-223394093 on Build GRCh38
rs753180931 Chr.1: 223394243-223394243 on Build GRCh38
rs201754034 Chr.1: 223394507-223394507 on Build GRCh38
rs757701072 Chr.1: 223394264-223394264 on Build GRCh38
rs763821566 Chr.1: 223394105-223394105 on Build GRCh38
rs755615384 Chr.1: 223394116-223394116 on Build GRCh38
rs573566361 Chr.1: 223394393-223394393 on Build GRCh38
rs374835925 Chr.1: 223394382-223394382 on Build GRCh38
rs780877557 Chr.1: 223394112-223394112 on Build GRCh38
rs991530032 Chr.1: 223394232-223394232 on Build GRCh38
rs376868687 Chr.1: 223394201-223394201 on Build GRCh38
rs150761495 Chr.1: 223394147-223394147 on Build GRCh38
rs780384449 Chr.1: 223394394-223394394 on Build GRCh38
rs756539010 Chr.1: 223394464-223394464 on Build GRCh38
rs746039667 Chr.1: 223394383-223394383 on Build GRCh38
rs757846365 Chr.1: 223394458-223394458 on Build GRCh38
rs375462609 Chr.1: 223394467-223394467 on Build GRCh38
rs758947736 Chr.1: 223394441-223394441 on Build GRCh38
rs962124709 Chr.1: 223394358-223394358 on Build GRCh38
rs373455542 Chr.1: 223394143-223394143 on Build GRCh38
rs751003401 Chr.1: 223394464-223394465 on Build GRCh38
rs761008873 Chr.1: 223394075-223394075 on Build GRCh38
rs750036613 Chr.1: 223394342-223394342 on Build GRCh38
rs202172237 Chr.1: 223394099-223394099 on Build GRCh38
rs779461493 Chr.1: 223394242-223394242 on Build GRCh38
rs750121015 Chr.1: 223394233-223394233 on Build GRCh38
rs182459489 Chr.1: 223394271-223394271 on Build GRCh38
rs751373210 Chr.1: 223394216-223394216 on Build GRCh38
rs369291712 Chr.1: 223394453-223394453 on Build GRCh38
rs562241616 Chr.1: 223394459-223394459 on Build GRCh38
rs1054742146 Chr.1: 223394345-223394345 on Build GRCh38
rs141894100 Chr.1: 223394100-223394100 on Build GRCh38
rs773451734 Chr.1: 223394146-223394146 on Build GRCh38
rs776862680 Chr.1: 223394084-223394084 on Build GRCh38
rs2298063 Chr.1: 223394189-223394189 on Build GRCh38
rs777062819 Chr.1: 223394181-223394181 on Build GRCh38
rs762894193 Chr.1: 223394322-223394322 on Build GRCh38
rs369679518 Chr.1: 223394509-223394509 on Build GRCh38
rs1010649111 Chr.1: 223394069-223394069 on Build GRCh38
rs545156576 Chr.1: 223394465-223394465 on Build GRCh38
rs1004066240 Chr.1: 223394457-223394457 on Build GRCh38
rs757107911 Chr.1: 223394217-223394217 on Build GRCh38
rs756911411 Chr.1: 223394110-223394110 on Build GRCh38
rs767318649 Chr.1: 223394341-223394341 on Build GRCh38
rs543098268 Chr.1: 223394326-223394326 on Build GRCh38
rs1051276807 Chr.1: 223394131-223394131 on Build GRCh38
rs267598389 Chr.1: 223394164-223394164 on Build GRCh38
rs752161726 Chr.1: 223394380-223394380 on Build GRCh38
rs765854757 Chr.1: 223394359-223394359 on Build GRCh38
rs1018061266 Chr.1: 223394094-223394094 on Build GRCh38
rs143126549 Chr.1: 223394229-223394229 on Build GRCh38
rs6682552 Chr.1: 223394466-223394466 on Build GRCh38
rs771266668 Chr.1: 223394162-223394162 on Build GRCh38
rs772895833 Chr.1: 223394402-223394402 on Build GRCh38
rs764863446 Chr.1: 223394445-223394445 on Build GRCh38
rs768770797 Chr.1: 223394397-223394397 on Build GRCh38
rs774220858 Chr.1: 223394399-223394399 on Build GRCh38
rs10907376 Chr.1: 223394461-223394461 on Build GRCh38
rs549370625 Chr.1: 223394494-223394494 on Build GRCh38
rs762833739 Chr.1: 223394193-223394193 on Build GRCh38
rs200353990 Chr.1: 223394286-223394286 on Build GRCh38
rs1022923790 Chr.1: 223394472-223394472 on Build GRCh38
rs759749551 Chr.1: 223394097-223394097 on Build GRCh38
rs141042142 Chr.1: 223394510-223394510 on Build GRCh38
rs922962218 Chr.1: 223394384-223394386 on Build GRCh38
rs768137339 Chr.1: 223394502-223394502 on Build GRCh38
rs774310490 Chr.1: 223394333-223394333 on Build GRCh38
rs751175288 Chr.1: 223394107-223394107 on Build GRCh38
rs766794599 Chr.1: 223394508-223394508 on Build GRCh38
rs757647945 Chr.1: 223394381-223394381 on Build GRCh38
rs533107933 Chr.1: 223394145-223394145 on Build GRCh38
rs761767460 Chr.1: 223394401-223394401 on Build GRCh38
rs757786997 Chr.1: 223394297-223394298 on Build GRCh38
rs763885685 Chr.1: 223394195-223394195 on Build GRCh38
rs764777306 Chr.1: 223394497-223394497 on Build GRCh38
rs376448310 Chr.1: 223394488-223394488 on Build GRCh38
rs886973069 Chr.1: 223394439-223394439 on Build GRCh38
rs751916392 Chr.1: 223394240-223394240 on Build GRCh38
rs924822575 Chr.1: 223394340-223394340 on Build GRCh38
rs567585826 Chr.1: 223394498-223394498 on Build GRCh38
rs772367435 Chr.1: 223394050-223394050 on Build GRCh38
rs573008518 Chr.1: 223394314-223394314 on Build GRCh38
rs200958819 Chr.1: 223394114-223394114 on Build GRCh38
rs765283214 Chr.1: 223394185-223394185 on Build GRCh38
rs771984467 Chr.1: 223394470-223394470 on Build GRCh38
rs766498404 Chr.1: 223394079-223394079 on Build GRCh38
rs556677048 Chr.1: 223394244-223394244 on Build GRCh38
rs753568204 Chr.1: 223394431-223394431 on Build GRCh38
rs6689850 Chr.1: 223394059-223394059 on Build GRCh38
rs781542528 Chr.1: 223394291-223394291 on Build GRCh38
rs544003933 Chr.1: 223394454-223394454 on Build GRCh38
rs537916148 Chr.1: 223394234-223394234 on Build GRCh38
rs144076109 Chr.1: 223394273-223394273 on Build GRCh38
rs749293228 Chr.1: 223394395-223394395 on Build GRCh38
rs981563098 Chr.1: 223394489-223394489 on Build GRCh38
rs775439837 Chr.1: 223394319-223394319 on Build GRCh38
rs186875984 Chr.1: 223394374-223394374 on Build GRCh38
rs770808438 Chr.1: 223394493-223394493 on Build GRCh38
rs746100656 Chr.1: 223394304-223394304 on Build GRCh38
rs778967444 Chr.1: 223394469-223394469 on Build GRCh38
rs759656949 Chr.1: 223394182-223394182 on Build GRCh38
rs779591245 Chr.1: 223394419-223394421 on Build GRCh38
rs760574515 Chr.1: 223394420-223394420 on Build GRCh38
rs868481392 Chr.1: 223394180-223394181 on Build GRCh38

View Full Product Details