Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 224929554-224930084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH14
Entrez Gene ID
127602
Gene Name
dynein axonemal heavy chain 14
Gene Aliases
C1orf67, Dnahc14, HL-18, HL18
UniGene
Hs.133977
Gene Chromosome Location
Chr.1: 224928422-225399294 on Build GRCh38
Transcripts
NM_001145154.1 NM_001373.1 NM_144989.2 XM_006711735.3 XM_011544057.2 XM_011544058.2 XM_011544059.2 XM_011544061.2 XM_011544062.2 XM_011544063.2 XM_011544064.2 XM_011544065.2 XM_011544066.2 XM_011544067.2 XM_011544069.2 XM_011544070.2 XM_011544071.2 XM_011544072.2 XM_017000295.1 XM_017000296.1 XM_017000297.1 XM_017000298.1 XM_017000300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6571829 c.389C>T p.S130L Chr.1: 224929701-224929701 on Build GRCh38
5509772 c.476G>A p.R159H Chr.1: 224929788-224929788 on Build GRCh38
5949479 c.517G>A p.A173T Chr.1: 224929829-224929829 on Build GRCh38
6037255 c.275A>G p.H92R Chr.1: 224929587-224929587 on Build GRCh38
5462196 c.331G>A p.A111T Chr.1: 224929643-224929643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923427172 Chr.1: 224929706-224929706 on Build GRCh38
rs1056898414 Chr.1: 224930039-224930039 on Build GRCh38
rs1030530557 Chr.1: 224929919-224929919 on Build GRCh38
rs746318504 Chr.1: 224929684-224929684 on Build GRCh38
rs180984440 Chr.1: 224930009-224930009 on Build GRCh38
rs891751218 Chr.1: 224929779-224929779 on Build GRCh38
rs879444791 Chr.1: 224929704-224929704 on Build GRCh38
rs986215915 Chr.1: 224929712-224929712 on Build GRCh38
rs373318095 Chr.1: 224929741-224929741 on Build GRCh38
rs558251988 Chr.1: 224929786-224929786 on Build GRCh38
rs918482025 Chr.1: 224929988-224929988 on Build GRCh38
rs1054781573 Chr.1: 224929740-224929740 on Build GRCh38
rs776958550 Chr.1: 224929922-224929922 on Build GRCh38
rs546856756 Chr.1: 224929649-224929649 on Build GRCh38
rs988134097 Chr.1: 224930026-224930026 on Build GRCh38
rs1000546087 Chr.1: 224929613-224929613 on Build GRCh38
rs1001256143 Chr.1: 224929833-224929833 on Build GRCh38
rs1026757689 Chr.1: 224929826-224929826 on Build GRCh38
rs746064908 Chr.1: 224929579-224929579 on Build GRCh38
rs953549796 Chr.1: 224929924-224929924 on Build GRCh38
rs1045599299 Chr.1: 224930041-224930041 on Build GRCh38
rs905626978 Chr.1: 224929828-224929828 on Build GRCh38
rs937156544 Chr.1: 224930056-224930056 on Build GRCh38
rs921994044 Chr.1: 224929711-224929711 on Build GRCh38
rs765980744 Chr.1: 224929692-224929692 on Build GRCh38
rs911931992 Chr.1: 224929939-224929939 on Build GRCh38
rs913400180 Chr.1: 224929701-224929701 on Build GRCh38
rs986635696 Chr.1: 224929944-224929944 on Build GRCh38
rs962323853 Chr.1: 224929898-224929898 on Build GRCh38
rs897021364 Chr.1: 224929780-224929780 on Build GRCh38
rs995735065 Chr.1: 224929823-224929823 on Build GRCh38
rs781537970 Chr.1: 224929675-224929675 on Build GRCh38
rs34258633 Chr.1: 224929752-224929752 on Build GRCh38
rs774085574 Chr.1: 224929643-224929643 on Build GRCh38
rs936659173 Chr.1: 224929739-224929739 on Build GRCh38
rs932046052 Chr.1: 224929717-224929717 on Build GRCh38
rs548770312 Chr.1: 224929636-224929636 on Build GRCh38
rs943961676 Chr.1: 224929748-224929748 on Build GRCh38
rs1043492556 Chr.1: 224929593-224929593 on Build GRCh38
rs1005846915 Chr.1: 224929889-224929889 on Build GRCh38
rs147586265 Chr.1: 224930048-224930048 on Build GRCh38
rs573399214 Chr.1: 224929799-224929799 on Build GRCh38
rs931220771 Chr.1: 224929989-224929989 on Build GRCh38
rs931965637 Chr.1: 224929625-224929625 on Build GRCh38
rs778358092 Chr.1: 224929973-224929973 on Build GRCh38
rs113525871 Chr.1: 224930028-224930028 on Build GRCh38
rs1020946602 Chr.1: 224929894-224929894 on Build GRCh38
rs761874474 Chr.1: 224929644-224929644 on Build GRCh38
rs987480196 Chr.1: 224929934-224929934 on Build GRCh38
rs951720967 Chr.1: 224929667-224929667 on Build GRCh38
rs778492003 Chr.1: 224929699-224929699 on Build GRCh38
rs903841484 Chr.1: 224929610-224929610 on Build GRCh38
rs932052910 Chr.1: 224929595-224929595 on Build GRCh38
rs1034074193 Chr.1: 224929852-224929852 on Build GRCh38
rs945958421 Chr.1: 224929784-224929784 on Build GRCh38
rs540779667 Chr.1: 224929834-224929834 on Build GRCh38
rs979359445 Chr.1: 224929902-224929902 on Build GRCh38
rs922699575 Chr.1: 224929837-224929837 on Build GRCh38
rs1034453152 Chr.1: 224929658-224929658 on Build GRCh38
rs936746339 Chr.1: 224929974-224929974 on Build GRCh38
rs922096786 Chr.1: 224929913-224929913 on Build GRCh38
rs778248923 Chr.1: 224929653-224929653 on Build GRCh38
rs978901718 Chr.1: 224929707-224929707 on Build GRCh38
rs989099444 Chr.1: 224929885-224929885 on Build GRCh38
rs945989107 Chr.1: 224930032-224930032 on Build GRCh38
rs535055095 Chr.1: 224929671-224929671 on Build GRCh38
rs1050891360 Chr.1: 224929604-224929604 on Build GRCh38
rs557075854 Chr.1: 224929651-224929651 on Build GRCh38
rs544934446 Chr.1: 224929912-224929912 on Build GRCh38
rs960291133 Chr.1: 224929688-224929688 on Build GRCh38
rs890423828 Chr.1: 224929729-224929729 on Build GRCh38
rs752007195 Chr.1: 224929713-224929713 on Build GRCh38
rs369979710 Chr.1: 224929897-224929897 on Build GRCh38
rs189391711 Chr.1: 224929668-224929668 on Build GRCh38
rs932078530 Chr.1: 224929915-224929915 on Build GRCh38
rs1051730219 Chr.1: 224929726-224929726 on Build GRCh38
rs979745384 Chr.1: 224929682-224929682 on Build GRCh38
rs1015391862 Chr.1: 224929650-224929650 on Build GRCh38
rs905660289 Chr.1: 224930015-224930015 on Build GRCh38
rs754389285 Chr.1: 224929698-224929698 on Build GRCh38
rs2501078 Chr.1: 224929859-224929859 on Build GRCh38
rs907026232 Chr.1: 224929825-224929825 on Build GRCh38
rs35169586 Chr.1: 224930043-224930044 on Build GRCh38
rs567183299 Chr.1: 224929645-224929645 on Build GRCh38
rs1009891939 Chr.1: 224930062-224930062 on Build GRCh38
rs746191108 Chr.1: 224929879-224929879 on Build GRCh38
rs533703934 Chr.1: 224929635-224929635 on Build GRCh38
rs940687890 Chr.1: 224929953-224929953 on Build GRCh38
rs12090519 Chr.1: 224929578-224929578 on Build GRCh38
rs566314987 Chr.1: 224929623-224929623 on Build GRCh38
rs539535975 Chr.1: 224929703-224929703 on Build GRCh38
rs1036980963 Chr.1: 224929986-224929986 on Build GRCh38
rs896941666 Chr.1: 224929654-224929654 on Build GRCh38
rs1036907096 Chr.1: 224929767-224929767 on Build GRCh38
rs753511041 Chr.1: 224929693-224929693 on Build GRCh38
rs766225478 Chr.1: 224930038-224930039 on Build GRCh38
rs770179358 Chr.1: 224929583-224929583 on Build GRCh38
rs757829115 Chr.1: 224929803-224929803 on Build GRCh38
rs947500409 Chr.1: 224929588-224929588 on Build GRCh38

View Full Product Details