Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235442747-235443153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALNT2
Entrez Gene ID
148789
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
B3GalNAc-T2, MDDGA11
UniGene
Hs.498143
Gene Chromosome Location
Chr.1: 235440654-235504481 on Build GRCh38
Transcripts
XM_006711749.3 NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

Gene Symbol
TBCE
Entrez Gene ID
6905
Gene Name
tubulin folding cofactor E
Gene Aliases
HRD, KCS, KCS1, pac2
UniGene
Hs.744998
Gene Chromosome Location
Chr.1: 235367360-235448968 on Build GRCh38
Transcripts
XM_006711749.3 NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203834 c.1383G>T p.L461L Chr.1: 235442895-235442895 on Build GRCh38
6326741 c.1534C>T p.L512L Chr.1: 235442893-235442893 on Build GRCh38
6203835 c.1536G>T p.L512L Chr.1: 235442895-235442895 on Build GRCh38
2000119 c.1551G>A p.P517P Chr.1: 235442910-235442910 on Build GRCh38
5600628 c.1531G>A p.V511I Chr.1: 235442890-235442890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746647958 Chr.1: 235442881-235442881 on Build GRCh38
rs114972093 Chr.1: 235442909-235442909 on Build GRCh38
rs779754010 Chr.1: 235442843-235442843 on Build GRCh38
rs1026677845 Chr.1: 235442829-235442829 on Build GRCh38
rs886323993 Chr.1: 235443091-235443091 on Build GRCh38
rs1051681540 Chr.1: 235442985-235442985 on Build GRCh38
rs548228406 Chr.1: 235443024-235443024 on Build GRCh38
rs1056199629 Chr.1: 235442993-235442993 on Build GRCh38
rs374630204 Chr.1: 235442817-235442817 on Build GRCh38
rs565010373 Chr.1: 235442818-235442818 on Build GRCh38
rs992469074 Chr.1: 235442882-235442882 on Build GRCh38
rs570627275 Chr.1: 235443110-235443110 on Build GRCh38
rs566329297 Chr.1: 235443026-235443026 on Build GRCh38
rs558610356 Chr.1: 235443094-235443094 on Build GRCh38
rs902401517 Chr.1: 235442966-235442966 on Build GRCh38
rs757577259 Chr.1: 235442958-235442958 on Build GRCh38
rs759698442 Chr.1: 235442922-235442922 on Build GRCh38
rs762799965 Chr.1: 235442910-235442910 on Build GRCh38
rs1023417051 Chr.1: 235443063-235443063 on Build GRCh38
rs756235800 Chr.1: 235442954-235442954 on Build GRCh38
rs761513705 Chr.1: 235442840-235442843 on Build GRCh38
rs778291075 Chr.1: 235442823-235442823 on Build GRCh38
rs969760597 Chr.1: 235442900-235442900 on Build GRCh38
rs373874343 Chr.1: 235442892-235442892 on Build GRCh38
rs370166627 Chr.1: 235442826-235442826 on Build GRCh38
rs368077100 Chr.1: 235442955-235442955 on Build GRCh38
rs756644526 Chr.1: 235442821-235442821 on Build GRCh38
rs749062134 Chr.1: 235442813-235442813 on Build GRCh38
rs995535487 Chr.1: 235443090-235443090 on Build GRCh38
rs370623097 Chr.1: 235442838-235442838 on Build GRCh38
rs752741900 Chr.1: 235442948-235442948 on Build GRCh38
rs1023936472 Chr.1: 235443104-235443104 on Build GRCh38
rs774572209 Chr.1: 235442921-235442921 on Build GRCh38
rs950395312 Chr.1: 235442850-235442850 on Build GRCh38
rs767545421 Chr.1: 235442926-235442926 on Build GRCh38
rs36123030 Chr.1: 235443071-235443071 on Build GRCh38
rs892456260 Chr.1: 235442791-235442791 on Build GRCh38
rs550575040 Chr.1: 235442860-235442860 on Build GRCh38
rs970076632 Chr.1: 235443087-235443087 on Build GRCh38
rs764983659 Chr.1: 235442932-235442933 on Build GRCh38
rs757621678 Chr.1: 235442831-235442831 on Build GRCh38
rs191932606 Chr.1: 235442802-235442802 on Build GRCh38
rs999325169 Chr.1: 235442987-235442987 on Build GRCh38
rs768158218 Chr.1: 235442891-235442891 on Build GRCh38
rs979836855 Chr.1: 235443109-235443109 on Build GRCh38
rs1051735023 Chr.1: 235442816-235442816 on Build GRCh38
rs371774114 Chr.1: 235442804-235442804 on Build GRCh38
rs748063082 Chr.1: 235442905-235442905 on Build GRCh38
rs112235573 Chr.1: 235443087-235443088 on Build GRCh38
rs969749174 Chr.1: 235443106-235443106 on Build GRCh38
rs983578769 Chr.1: 235442807-235442807 on Build GRCh38
rs779162506 Chr.1: 235442961-235442961 on Build GRCh38
rs936865670 Chr.1: 235442928-235442928 on Build GRCh38
rs533767497 Chr.1: 235443032-235443032 on Build GRCh38
rs950910677 Chr.1: 235443119-235443119 on Build GRCh38
rs184180199 Chr.1: 235443118-235443118 on Build GRCh38
rs367666825 Chr.1: 235442819-235442819 on Build GRCh38

View Full Product Details