Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235447435-235447934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALNT2
Entrez Gene ID
148789
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
B3GalNAc-T2, MDDGA11
UniGene
Hs.498143
Gene Chromosome Location
Chr.1: 235440654-235504481 on Build GRCh38
Transcripts
NM_152490.4 XM_006711749.3 XM_017000394.1 XM_017000395.1 NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

Gene Symbol
TBCE
Entrez Gene ID
6905
Gene Name
tubulin folding cofactor E
Gene Aliases
HRD, KCS, KCS1, pac2
UniGene
Hs.744998
Gene Chromosome Location
Chr.1: 235367360-235448968 on Build GRCh38
Transcripts
NM_152490.4 XM_006711749.3 XM_017000394.1 XM_017000395.1 NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

SNP Detail

SNP ID Chromosome Location
rs192751881 Chr.1: 235447741-235447741 on Build GRCh38
rs375263038 Chr.1: 235447546-235447546 on Build GRCh38
rs150091674 Chr.1: 235447657-235447657 on Build GRCh38
rs986791082 Chr.1: 235447819-235447819 on Build GRCh38
rs546786721 Chr.1: 235447724-235447724 on Build GRCh38
rs555667689 Chr.1: 235447521-235447521 on Build GRCh38
rs140675942 Chr.1: 235447620-235447620 on Build GRCh38
rs974146416 Chr.1: 235447713-235447713 on Build GRCh38
rs575533212 Chr.1: 235447683-235447683 on Build GRCh38
rs929975871 Chr.1: 235447780-235447780 on Build GRCh38
rs954554685 Chr.1: 235447662-235447662 on Build GRCh38
rs949388614 Chr.1: 235447778-235447780 on Build GRCh38
rs930205794 Chr.1: 235447708-235447709 on Build GRCh38
rs962816177 Chr.1: 235447704-235447704 on Build GRCh38
rs775257108 Chr.1: 235447833-235447833 on Build GRCh38
rs1011269706 Chr.1: 235447576-235447576 on Build GRCh38
rs898715462 Chr.1: 235447494-235447494 on Build GRCh38
rs1036021764 Chr.1: 235447643-235447643 on Build GRCh38
rs930275987 Chr.1: 235447526-235447526 on Build GRCh38
rs1001076561 Chr.1: 235447549-235447549 on Build GRCh38
rs910266723 Chr.1: 235447677-235447678 on Build GRCh38
rs1051786545 Chr.1: 235447542-235447542 on Build GRCh38
rs1032132527 Chr.1: 235447877-235447877 on Build GRCh38
rs546931264 Chr.1: 235447884-235447884 on Build GRCh38
rs771592523 Chr.1: 235447785-235447785 on Build GRCh38
rs1000907397 Chr.1: 235447835-235447835 on Build GRCh38
rs868821534 Chr.1: 235447811-235447811 on Build GRCh38
rs892333634 Chr.1: 235447887-235447888 on Build GRCh38
rs778376848 Chr.1: 235447703-235447703 on Build GRCh38
rs1038561864 Chr.1: 235447485-235447485 on Build GRCh38
rs892912988 Chr.1: 235447567-235447567 on Build GRCh38
rs187517435 Chr.1: 235447629-235447629 on Build GRCh38
rs772488235 Chr.1: 235447823-235447823 on Build GRCh38
rs774845934 Chr.1: 235447656-235447656 on Build GRCh38
rs551030877 Chr.1: 235447893-235447894 on Build GRCh38
rs770367698 Chr.1: 235447625-235447625 on Build GRCh38
rs565134431 Chr.1: 235447896-235447896 on Build GRCh38
rs985833833 Chr.1: 235447662-235447663 on Build GRCh38
rs183385984 Chr.1: 235447543-235447543 on Build GRCh38
rs918570205 Chr.1: 235447522-235447522 on Build GRCh38
rs533015778 Chr.1: 235447522-235447523 on Build GRCh38
rs371610478 Chr.1: 235447466-235447466 on Build GRCh38
rs967448467 Chr.1: 235447635-235447635 on Build GRCh38
rs373250426 Chr.1: 235447660-235447661 on Build GRCh38
rs566541760 Chr.1: 235447461-235447461 on Build GRCh38
rs190052220 Chr.1: 235447504-235447504 on Build GRCh38
rs1024161013 Chr.1: 235447545-235447545 on Build GRCh38
rs540681084 Chr.1: 235447652-235447653 on Build GRCh38
rs1047375470 Chr.1: 235447714-235447714 on Build GRCh38
rs1002501343 Chr.1: 235447637-235447637 on Build GRCh38
rs1032142019 Chr.1: 235447594-235447595 on Build GRCh38
rs528770533 Chr.1: 235447824-235447824 on Build GRCh38
rs150925518 Chr.1: 235447560-235447560 on Build GRCh38
rs187514713 Chr.1: 235447467-235447467 on Build GRCh38
rs920133447 Chr.1: 235447701-235447701 on Build GRCh38
rs941798152 Chr.1: 235447680-235447680 on Build GRCh38
rs532768784 Chr.1: 235447909-235447909 on Build GRCh38
rs942939580 Chr.1: 235447887-235447887 on Build GRCh38
rs994426492 Chr.1: 235447499-235447499 on Build GRCh38
rs769131445 Chr.1: 235447638-235447638 on Build GRCh38
rs958618794 Chr.1: 235447648-235447648 on Build GRCh38
rs909026620 Chr.1: 235447684-235447684 on Build GRCh38

View Full Product Details