Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 245083446-245083965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB2
Entrez Gene ID
84288
Gene Name
EF-hand calcium binding domain 2
Gene Aliases
CFAP200, DRC8
UniGene
Hs.134857
Gene Chromosome Location
Chr.1: 244969739-245125228 on Build GRCh38
Transcripts
NM_001143943.1 NM_001290327.1 NM_032328.3 NR_026587.1 XM_011544300.2 XM_011544301.1 XM_011544302.2 XM_017002538.1 XM_017002539.1 XM_017002540.1 XM_017002541.1 XM_017002542.1 XM_017002543.1 XM_017002544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2051563 c.357G>A p.K119K Chr.1: 245083672-245083672 on Build GRCh38
7316344 c.762C>T p.I254I Chr.1: 245083669-245083669 on Build GRCh38
4577105 c.346G>A p.E116K Chr.1: 245083661-245083661 on Build GRCh38
2051561 c.325C>T p.R109C Chr.1: 245083640-245083640 on Build GRCh38
7316343 c.354C>T p.I118I Chr.1: 245083669-245083669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202024615 Chr.1: 245083605-245083605 on Build GRCh38
rs188250034 Chr.1: 245083763-245083763 on Build GRCh38
rs903078968 Chr.1: 245083808-245083808 on Build GRCh38
rs200554796 Chr.1: 245083673-245083673 on Build GRCh38
rs770810575 Chr.1: 245083500-245083500 on Build GRCh38
rs754162244 Chr.1: 245083719-245083719 on Build GRCh38
rs753576183 Chr.1: 245083585-245083585 on Build GRCh38
rs955002262 Chr.1: 245083548-245083548 on Build GRCh38
rs765544922 Chr.1: 245083592-245083592 on Build GRCh38
rs552295066 Chr.1: 245083542-245083542 on Build GRCh38
rs11541381 Chr.1: 245083470-245083470 on Build GRCh38
rs373412586 Chr.1: 245083660-245083660 on Build GRCh38
rs527797639 Chr.1: 245083597-245083597 on Build GRCh38
rs762050759 Chr.1: 245083588-245083588 on Build GRCh38
rs760081034 Chr.1: 245083685-245083685 on Build GRCh38
rs750806554 Chr.1: 245083595-245083595 on Build GRCh38
rs1000433235 Chr.1: 245083514-245083514 on Build GRCh38
rs1032867877 Chr.1: 245083540-245083540 on Build GRCh38
rs10754849 Chr.1: 245083878-245083878 on Build GRCh38
rs756868742 Chr.1: 245083826-245083826 on Build GRCh38
rs754985035 Chr.1: 245083734-245083734 on Build GRCh38
rs751283277 Chr.1: 245083823-245083823 on Build GRCh38
rs1029384505 Chr.1: 245083547-245083547 on Build GRCh38
rs772067994 Chr.1: 245083672-245083672 on Build GRCh38
rs761475638 Chr.1: 245083704-245083704 on Build GRCh38
rs780540073 Chr.1: 245083603-245083603 on Build GRCh38
rs894295002 Chr.1: 245083693-245083693 on Build GRCh38
rs781253010 Chr.1: 245083627-245083627 on Build GRCh38
rs774403512 Chr.1: 245083501-245083501 on Build GRCh38
rs759891606 Chr.1: 245083720-245083720 on Build GRCh38
rs768250433 Chr.1: 245083695-245083695 on Build GRCh38
rs569014161 Chr.1: 245083634-245083634 on Build GRCh38
rs984567711 Chr.1: 245083898-245083898 on Build GRCh38
rs568234706 Chr.1: 245083756-245083756 on Build GRCh38
rs761115089 Chr.1: 245083753-245083753 on Build GRCh38
rs752743084 Chr.1: 245083746-245083746 on Build GRCh38
rs1029202458 Chr.1: 245083867-245083867 on Build GRCh38
rs759507659 Chr.1: 245083502-245083502 on Build GRCh38
rs576965038 Chr.1: 245083853-245083853 on Build GRCh38
rs200608574 Chr.1: 245083714-245083714 on Build GRCh38
rs866748785 Chr.1: 245083723-245083723 on Build GRCh38
rs575340420 Chr.1: 245083939-245083939 on Build GRCh38
rs371783937 Chr.1: 245083849-245083849 on Build GRCh38
rs76577803 Chr.1: 245083793-245083793 on Build GRCh38
rs149676779 Chr.1: 245083473-245083473 on Build GRCh38
rs775564411 Chr.1: 245083674-245083674 on Build GRCh38
rs781295516 Chr.1: 245083745-245083745 on Build GRCh38
rs763371255 Chr.1: 245083724-245083724 on Build GRCh38
rs917342896 Chr.1: 245083763-245083764 on Build GRCh38
rs376793032 Chr.1: 245083726-245083726 on Build GRCh38
rs764603706 Chr.1: 245083755-245083755 on Build GRCh38
rs764881049 Chr.1: 245083717-245083717 on Build GRCh38
rs539288976 Chr.1: 245083877-245083877 on Build GRCh38
rs776034855 Chr.1: 245083702-245083702 on Build GRCh38
rs758316509 Chr.1: 245083757-245083757 on Build GRCh38
rs963749578 Chr.1: 245083541-245083544 on Build GRCh38
rs975100758 Chr.1: 245083541-245083541 on Build GRCh38
rs760237391 Chr.1: 245083509-245083509 on Build GRCh38
rs752161128 Chr.1: 245083729-245083729 on Build GRCh38
rs368983163 Chr.1: 245083635-245083635 on Build GRCh38
rs745672916 Chr.1: 245083664-245083664 on Build GRCh38
rs765167044 Chr.1: 245083922-245083922 on Build GRCh38
rs928284051 Chr.1: 245083925-245083925 on Build GRCh38
rs1036241957 Chr.1: 245083528-245083528 on Build GRCh38
rs375498220 Chr.1: 245083857-245083857 on Build GRCh38
rs928524825 Chr.1: 245083565-245083565 on Build GRCh38
rs372474589 Chr.1: 245083513-245083513 on Build GRCh38
rs532779256 Chr.1: 245083533-245083533 on Build GRCh38
rs778803377 Chr.1: 245083661-245083661 on Build GRCh38
rs1016389606 Chr.1: 245083916-245083916 on Build GRCh38
rs961707293 Chr.1: 245083917-245083917 on Build GRCh38
rs775082442 Chr.1: 245083507-245083507 on Build GRCh38
rs369597079 Chr.1: 245083684-245083684 on Build GRCh38
rs200585506 Chr.1: 245083620-245083620 on Build GRCh38
rs145465501 Chr.1: 245083474-245083474 on Build GRCh38
rs938586448 Chr.1: 245083573-245083573 on Build GRCh38
rs755147570 Chr.1: 245083616-245083616 on Build GRCh38
rs902126891 Chr.1: 245083786-245083786 on Build GRCh38
rs772159938 Chr.1: 245083503-245083503 on Build GRCh38

View Full Product Details